Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1337791-1338283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL1
Entrez Gene ID
1855
Gene Name
dishevelled segment polarity protein 1
Gene Aliases
DRS2, DVL, DVL1L1, DVL1P1
UniGene
Hs.731450
Gene Chromosome Location
Chr.1: 1335278-1349142 on Build GRCh38
Transcripts
NM_004421.2 XM_005244731.3 XM_005244732.3 XM_005244733.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1600674 c.1574C>T p.P525L Chr.1: 1338042-1338042 on Build GRCh38
4873556 c.1536C>T p.G512G Chr.1: 1338155-1338155 on Build GRCh38
5422063 c.1593delC p.W532fs*142 Chr.1: 1338098-1338098 on Build GRCh38
5563814 c.1623G>A p.Q541Q Chr.1: 1338068-1338068 on Build GRCh38
2180918 c.1508C>A p.P503Q Chr.1: 1338108-1338108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781424239 Chr.1: 1338245-1338245 on Build GRCh38
rs747297839 Chr.1: 1338173-1338173 on Build GRCh38
rs924762290 Chr.1: 1337950-1337950 on Build GRCh38
rs951394649 Chr.1: 1337916-1337916 on Build GRCh38
rs770137374 Chr.1: 1338041-1338041 on Build GRCh38
rs577875311 Chr.1: 1337847-1337847 on Build GRCh38
rs745919268 Chr.1: 1338226-1338226 on Build GRCh38
rs754145019 Chr.1: 1337891-1337891 on Build GRCh38
rs557766838 Chr.1: 1337841-1337841 on Build GRCh38
rs771502796 Chr.1: 1337886-1337886 on Build GRCh38
rs748931737 Chr.1: 1337883-1337883 on Build GRCh38
rs761153617 Chr.1: 1337888-1337888 on Build GRCh38
rs770453846 Chr.1: 1337987-1337987 on Build GRCh38
rs751295575 Chr.1: 1338140-1338140 on Build GRCh38
rs200411366 Chr.1: 1338242-1338242 on Build GRCh38
rs1049411951 Chr.1: 1338216-1338216 on Build GRCh38
rs373206650 Chr.1: 1338224-1338224 on Build GRCh38
rs906263591 Chr.1: 1337857-1337857 on Build GRCh38
rs797044834 Chr.1: 1338099-1338111 on Build GRCh38
rs745839316 Chr.1: 1337877-1337877 on Build GRCh38
rs760842641 Chr.1: 1338081-1338081 on Build GRCh38
rs746357673 Chr.1: 1338039-1338039 on Build GRCh38
rs751532000 Chr.1: 1337965-1337965 on Build GRCh38
rs745945473 Chr.1: 1338188-1338188 on Build GRCh38
rs372744736 Chr.1: 1338253-1338253 on Build GRCh38
rs748714505 Chr.1: 1337856-1337857 on Build GRCh38
rs201374747 Chr.1: 1337985-1337985 on Build GRCh38
rs747796678 Chr.1: 1337939-1337939 on Build GRCh38
rs978008683 Chr.1: 1337947-1337947 on Build GRCh38
rs764152783 Chr.1: 1338003-1338003 on Build GRCh38
rs752711097 Chr.1: 1337863-1337863 on Build GRCh38
rs761596141 Chr.1: 1338141-1338141 on Build GRCh38
rs939106565 Chr.1: 1338193-1338193 on Build GRCh38
rs756958598 Chr.1: 1338101-1338101 on Build GRCh38
rs797044836 Chr.1: 1338108-1338108 on Build GRCh38
rs559830305 Chr.1: 1337940-1337940 on Build GRCh38
rs768365981 Chr.1: 1337884-1337884 on Build GRCh38
rs531679178 Chr.1: 1338014-1338014 on Build GRCh38
rs771354674 Chr.1: 1337895-1337927 on Build GRCh38
rs753824562 Chr.1: 1338010-1338010 on Build GRCh38
rs183585991 Chr.1: 1337840-1337840 on Build GRCh38
rs797044839 Chr.1: 1338054-1338054 on Build GRCh38
rs759901637 Chr.1: 1337892-1337892 on Build GRCh38
rs763780144 Chr.1: 1338139-1338139 on Build GRCh38
rs375584920 Chr.1: 1338042-1338042 on Build GRCh38
rs779653558 Chr.1: 1338155-1338155 on Build GRCh38
rs797044835 Chr.1: 1338097-1338097 on Build GRCh38
rs760441838 Chr.1: 1338209-1338209 on Build GRCh38
rs763385815 Chr.1: 1338002-1338002 on Build GRCh38
rs531247891 Chr.1: 1338119-1338119 on Build GRCh38
rs750367344 Chr.1: 1337927-1337927 on Build GRCh38
rs762638741 Chr.1: 1338094-1338094 on Build GRCh38
rs797044833 Chr.1: 1338045-1338046 on Build GRCh38
rs199505104 Chr.1: 1338099-1338099 on Build GRCh38
rs767999045 Chr.1: 1337832-1337832 on Build GRCh38
rs191759160 Chr.1: 1338189-1338189 on Build GRCh38
rs779568971 Chr.1: 1338162-1338162 on Build GRCh38
rs765407601 Chr.1: 1337901-1337901 on Build GRCh38
rs780150258 Chr.1: 1337930-1337930 on Build GRCh38
rs368987984 Chr.1: 1338168-1338168 on Build GRCh38
rs759782304 Chr.1: 1337900-1337901 on Build GRCh38
rs939780908 Chr.1: 1337949-1337949 on Build GRCh38
rs376184953 Chr.1: 1338238-1338238 on Build GRCh38
rs770136685 Chr.1: 1337998-1337998 on Build GRCh38
rs369598586 Chr.1: 1338211-1338211 on Build GRCh38
rs936240718 Chr.1: 1337838-1337838 on Build GRCh38
rs761241500 Chr.1: 1337831-1337831 on Build GRCh38
rs748121596 Chr.1: 1338232-1338232 on Build GRCh38
rs746121596 Chr.1: 1338247-1338247 on Build GRCh38
rs767263562 Chr.1: 1338254-1338255 on Build GRCh38
rs1003816393 Chr.1: 1337859-1337859 on Build GRCh38
rs762939816 Chr.1: 1337960-1337960 on Build GRCh38
rs771365185 Chr.1: 1337948-1337948 on Build GRCh38
rs757439870 Chr.1: 1338223-1338223 on Build GRCh38
rs367666036 Chr.1: 1337966-1337966 on Build GRCh38
rs777158095 Chr.1: 1337951-1337951 on Build GRCh38
rs752929560 Chr.1: 1337959-1337959 on Build GRCh38
rs774249249 Chr.1: 1338231-1338231 on Build GRCh38
rs373781378 Chr.1: 1337934-1337934 on Build GRCh38
rs768600968 Chr.1: 1337933-1337933 on Build GRCh38
rs773576186 Chr.1: 1337868-1337869 on Build GRCh38
rs755252173 Chr.1: 1337854-1337854 on Build GRCh38
rs534278648 Chr.1: 1338212-1338212 on Build GRCh38
rs551168455 Chr.1: 1338120-1338120 on Build GRCh38
rs567839626 Chr.1: 1338128-1338128 on Build GRCh38
rs926561391 Chr.1: 1337910-1337910 on Build GRCh38
rs777603609 Chr.1: 1338261-1338261 on Build GRCh38
rs370012293 Chr.1: 1338239-1338239 on Build GRCh38
rs752706599 Chr.1: 1338089-1338089 on Build GRCh38
rs775680911 Chr.1: 1337946-1337946 on Build GRCh38
rs1033578505 Chr.1: 1338109-1338109 on Build GRCh38
rs797044837 Chr.1: 1338001-1338001 on Build GRCh38
rs758485540 Chr.1: 1337921-1337921 on Build GRCh38
rs375866434 Chr.1: 1338254-1338254 on Build GRCh38
rs774149938 Chr.1: 1337885-1337885 on Build GRCh38
rs766379009 Chr.1: 1337968-1337968 on Build GRCh38
rs545744942 Chr.1: 1337865-1337865 on Build GRCh38
rs757930549 Chr.1: 1337976-1337976 on Build GRCh38
rs780744557 Chr.1: 1338031-1338031 on Build GRCh38
rs747188657 Chr.1: 1338219-1338219 on Build GRCh38
rs990434682 Chr.1: 1337962-1337962 on Build GRCh38
rs1037968088 Chr.1: 1338234-1338234 on Build GRCh38
rs765642883 Chr.1: 1337958-1337958 on Build GRCh38
rs763149026 Chr.1: 1337961-1337961 on Build GRCh38
rs1046846094 Chr.1: 1338204-1338204 on Build GRCh38
rs780834416 Chr.1: 1338108-1338108 on Build GRCh38
rs764216100 Chr.1: 1337963-1337963 on Build GRCh38
rs772700354 Chr.1: 1337866-1337898 on Build GRCh38
rs760247378 Chr.1: 1337946-1337946 on Build GRCh38
rs767347338 Chr.1: 1338191-1338191 on Build GRCh38
rs1041874225 Chr.1: 1337825-1337825 on Build GRCh38
rs797044838 Chr.1: 1338087-1338087 on Build GRCh38
rs755026350 Chr.1: 1338015-1338015 on Build GRCh38
rs748486151 Chr.1: 1338163-1338163 on Build GRCh38
rs751395065 Chr.1: 1338235-1338235 on Build GRCh38
rs765532309 Chr.1: 1337923-1337934 on Build GRCh38
rs758442067 Chr.1: 1337873-1337873 on Build GRCh38
rs749850446 Chr.1: 1338113-1338113 on Build GRCh38
rs759095836 Chr.1: 1338240-1338240 on Build GRCh38
rs372304219 Chr.1: 1338005-1338005 on Build GRCh38
rs774664222 Chr.1: 1338004-1338004 on Build GRCh38
rs544336961 Chr.1: 1338008-1338008 on Build GRCh38
rs61766209 Chr.1: 1337974-1337974 on Build GRCh38
rs141914568 Chr.1: 1338076-1338076 on Build GRCh38
rs375337359 Chr.1: 1338251-1338251 on Build GRCh38
rs755677662 Chr.1: 1338112-1338112 on Build GRCh38
rs773740343 Chr.1: 1337887-1337887 on Build GRCh38
rs769086399 Chr.1: 1338058-1338058 on Build GRCh38
rs370632814 Chr.1: 1337969-1337969 on Build GRCh38
rs757084322 Chr.1: 1338234-1338235 on Build GRCh38
rs751211931 Chr.1: 1338100-1338100 on Build GRCh38
rs777752538 Chr.1: 1337924-1337924 on Build GRCh38
rs778353686 Chr.1: 1338218-1338218 on Build GRCh38
rs755556180 Chr.1: 1338153-1338153 on Build GRCh38
rs752887884 Chr.1: 1337903-1337903 on Build GRCh38
rs747825850 Chr.1: 1337973-1337973 on Build GRCh38
rs770915923 Chr.1: 1338179-1338179 on Build GRCh38
rs771096309 Chr.1: 1338130-1338130 on Build GRCh38
rs137945460 Chr.1: 1338241-1338241 on Build GRCh38
rs563155947 Chr.1: 1337928-1337928 on Build GRCh38
rs753323723 Chr.1: 1338210-1338210 on Build GRCh38
rs779479020 Chr.1: 1338125-1338125 on Build GRCh38
rs869025220 Chr.1: 1338094-1338094 on Build GRCh38
rs755456412 Chr.1: 1338161-1338161 on Build GRCh38
rs996833746 Chr.1: 1338072-1338072 on Build GRCh38
rs1049961984 Chr.1: 1337995-1337995 on Build GRCh38
rs772817718 Chr.1: 1338196-1338196 on Build GRCh38
rs750922389 Chr.1: 1337839-1337839 on Build GRCh38
rs769881531 Chr.1: 1338227-1338227 on Build GRCh38
rs769597295 Chr.1: 1337879-1337879 on Build GRCh38
rs777024094 Chr.1: 1337899-1337931 on Build GRCh38
rs780480611 Chr.1: 1338144-1338144 on Build GRCh38
rs763017084 Chr.1: 1338138-1338138 on Build GRCh38
rs144724672 Chr.1: 1338022-1338022 on Build GRCh38
rs756731811 Chr.1: 1338024-1338024 on Build GRCh38
rs754295240 Chr.1: 1338156-1338156 on Build GRCh38
rs573767085 Chr.1: 1337932-1337932 on Build GRCh38
rs766717720 Chr.1: 1337890-1337890 on Build GRCh38
rs775261386 Chr.1: 1338135-1338135 on Build GRCh38
rs754055328 Chr.1: 1337852-1337852 on Build GRCh38
rs759848205 Chr.1: 1337954-1337954 on Build GRCh38
rs372765398 Chr.1: 1338057-1338057 on Build GRCh38
rs941025089 Chr.1: 1338229-1338229 on Build GRCh38
rs752368823 Chr.1: 1338018-1338018 on Build GRCh38
rs778641019 Chr.1: 1337972-1337972 on Build GRCh38
rs764152283 Chr.1: 1337962-1337963 on Build GRCh38
rs797044840 Chr.1: 1338033-1338040 on Build GRCh38
rs775773160 Chr.1: 1338000-1338000 on Build GRCh38
rs886913922 Chr.1: 1338029-1338029 on Build GRCh38
rs307370 Chr.1: 1337898-1337898 on Build GRCh38
rs746715259 Chr.1: 1338023-1338023 on Build GRCh38
rs767693311 Chr.1: 1338080-1338080 on Build GRCh38
rs1016847346 Chr.1: 1338054-1338054 on Build GRCh38
rs1036678401 Chr.1: 1337952-1337952 on Build GRCh38
rs765046286 Chr.1: 1338082-1338082 on Build GRCh38
rs747410001 Chr.1: 1337890-1337890 on Build GRCh38
rs370256175 Chr.1: 1338250-1338250 on Build GRCh38
rs780057928 Chr.1: 1337881-1337881 on Build GRCh38
rs756494911 Chr.1: 1338129-1338129 on Build GRCh38
rs749864968 Chr.1: 1338111-1338111 on Build GRCh38
rs781138454 Chr.1: 1337874-1337874 on Build GRCh38
rs746416838 Chr.1: 1337994-1337994 on Build GRCh38
rs750098261 Chr.1: 1338145-1338145 on Build GRCh38
rs765234696 Chr.1: 1338017-1338017 on Build GRCh38

View Full Product Details