Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1374232-1374730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKAIP1
Entrez Gene ID
54998
Gene Name
aurora kinase A interacting protein 1
Gene Aliases
AIP, AKIP, MRP-S38
UniGene
Hs.632515
Gene Chromosome Location
Chr.1: 1373730-1375438 on Build GRCh38
Transcripts
NM_001127229.1 NM_001127230.1 NM_017900.2 XM_011541670.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4021501 c.171G>A p.Q57Q Chr.1: 1374327-1374327 on Build GRCh38
4664753 c.164G>A p.G55E Chr.1: 1374334-1374334 on Build GRCh38
5461966 c.220C>A p.L74M Chr.1: 1374278-1374278 on Build GRCh38
4639327 c.102C>T p.C34C Chr.1: 1374396-1374396 on Build GRCh38
4494805 c.43C>T p.P15S Chr.1: 1374714-1374714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776156349 Chr.1: 1374332-1374332 on Build GRCh38
rs35745967 Chr.1: 1374505-1374505 on Build GRCh38
rs749208474 Chr.1: 1374302-1374302 on Build GRCh38
rs1051616605 Chr.1: 1374515-1374515 on Build GRCh38
rs777456556 Chr.1: 1374420-1374420 on Build GRCh38
rs1031144999 Chr.1: 1374632-1374632 on Build GRCh38
rs763392452 Chr.1: 1374439-1374439 on Build GRCh38
rs765790127 Chr.1: 1374344-1374344 on Build GRCh38
rs768764353 Chr.1: 1374303-1374303 on Build GRCh38
rs761231104 Chr.1: 1374419-1374420 on Build GRCh38
rs370802222 Chr.1: 1374666-1374666 on Build GRCh38
rs989581705 Chr.1: 1374491-1374491 on Build GRCh38
rs533870285 Chr.1: 1374509-1374509 on Build GRCh38
rs762082992 Chr.1: 1374474-1374474 on Build GRCh38
rs774329986 Chr.1: 1374256-1374256 on Build GRCh38
rs754290379 Chr.1: 1374281-1374281 on Build GRCh38
rs555752171 Chr.1: 1374581-1374581 on Build GRCh38
rs377520465 Chr.1: 1374262-1374262 on Build GRCh38
rs1037423069 Chr.1: 1374587-1374587 on Build GRCh38
rs938504006 Chr.1: 1374561-1374561 on Build GRCh38
rs940418090 Chr.1: 1374586-1374586 on Build GRCh38
rs771854901 Chr.1: 1374414-1374414 on Build GRCh38
rs1031490710 Chr.1: 1374466-1374466 on Build GRCh38
rs143351909 Chr.1: 1374298-1374298 on Build GRCh38
rs1031815420 Chr.1: 1374585-1374585 on Build GRCh38
rs774776478 Chr.1: 1374693-1374693 on Build GRCh38
rs980423955 Chr.1: 1374637-1374637 on Build GRCh38
rs1010960091 Chr.1: 1374648-1374648 on Build GRCh38
rs201853353 Chr.1: 1374433-1374433 on Build GRCh38
rs996912201 Chr.1: 1374368-1374368 on Build GRCh38
rs753105916 Chr.1: 1374290-1374290 on Build GRCh38
rs763310367 Chr.1: 1374357-1374357 on Build GRCh38
rs776103208 Chr.1: 1374426-1374426 on Build GRCh38
rs1029736968 Chr.1: 1374394-1374394 on Build GRCh38
rs770536101 Chr.1: 1374425-1374425 on Build GRCh38
rs745532255 Chr.1: 1374688-1374688 on Build GRCh38
rs980964172 Chr.1: 1374442-1374442 on Build GRCh38
rs751766763 Chr.1: 1374369-1374369 on Build GRCh38
rs770430238 Chr.1: 1374331-1374331 on Build GRCh38
rs927816694 Chr.1: 1374280-1374280 on Build GRCh38
rs769296320 Chr.1: 1374434-1374434 on Build GRCh38
rs778840388 Chr.1: 1374306-1374306 on Build GRCh38
rs947760709 Chr.1: 1374258-1374258 on Build GRCh38
rs956798438 Chr.1: 1374490-1374490 on Build GRCh38
rs748114170 Chr.1: 1374401-1374401 on Build GRCh38
rs774137353 Chr.1: 1374379-1374379 on Build GRCh38
rs3736373 Chr.1: 1374400-1374400 on Build GRCh38
rs868461849 Chr.1: 1374428-1374428 on Build GRCh38
rs768807674 Chr.1: 1374539-1374539 on Build GRCh38
rs11548463 Chr.1: 1374562-1374562 on Build GRCh38
rs775021531 Chr.1: 1374690-1374690 on Build GRCh38
rs200673616 Chr.1: 1374288-1374288 on Build GRCh38
rs969083685 Chr.1: 1374617-1374617 on Build GRCh38
rs753052826 Chr.1: 1374354-1374354 on Build GRCh38
rs758605382 Chr.1: 1374295-1374295 on Build GRCh38
rs760169640 Chr.1: 1374271-1374271 on Build GRCh38
rs527525962 Chr.1: 1374680-1374680 on Build GRCh38
rs756209362 Chr.1: 1374657-1374657 on Build GRCh38
rs761474608 Chr.1: 1374664-1374664 on Build GRCh38
rs757311906 Chr.1: 1374300-1374300 on Build GRCh38
rs11548465 Chr.1: 1374312-1374312 on Build GRCh38
rs576921931 Chr.1: 1374603-1374604 on Build GRCh38
rs567480343 Chr.1: 1374599-1374599 on Build GRCh38
rs775146202 Chr.1: 1374360-1374360 on Build GRCh38
rs980827829 Chr.1: 1374669-1374669 on Build GRCh38
rs1038233937 Chr.1: 1374499-1374499 on Build GRCh38
rs767285152 Chr.1: 1374298-1374299 on Build GRCh38
rs2765035 Chr.1: 1374694-1374694 on Build GRCh38
rs868006729 Chr.1: 1374322-1374322 on Build GRCh38
rs775917502 Chr.1: 1374438-1374438 on Build GRCh38
rs764487926 Chr.1: 1374462-1374462 on Build GRCh38
rs374922558 Chr.1: 1374598-1374598 on Build GRCh38
rs1046861150 Chr.1: 1374484-1374484 on Build GRCh38
rs576894508 Chr.1: 1374528-1374528 on Build GRCh38
rs553614602 Chr.1: 1374510-1374510 on Build GRCh38
rs778930104 Chr.1: 1374399-1374399 on Build GRCh38
rs780177298 Chr.1: 1374297-1374297 on Build GRCh38
rs760011627 Chr.1: 1374335-1374335 on Build GRCh38
rs754965138 Chr.1: 1374397-1374397 on Build GRCh38
rs955042573 Chr.1: 1374461-1374461 on Build GRCh38
rs927269434 Chr.1: 1374538-1374538 on Build GRCh38
rs1029650730 Chr.1: 1374600-1374600 on Build GRCh38
rs749781154 Chr.1: 1374691-1374691 on Build GRCh38
rs377379487 Chr.1: 1374323-1374323 on Build GRCh38
rs772595313 Chr.1: 1374270-1374270 on Build GRCh38
rs766592469 Chr.1: 1374658-1374658 on Build GRCh38
rs9554 Chr.1: 1374423-1374423 on Build GRCh38
rs1023215502 Chr.1: 1374605-1374605 on Build GRCh38
rs777599781 Chr.1: 1374671-1374671 on Build GRCh38
rs771772525 Chr.1: 1374318-1374318 on Build GRCh38
rs781161693 Chr.1: 1374301-1374301 on Build GRCh38
rs767920663 Chr.1: 1374482-1374482 on Build GRCh38
rs148649999 Chr.1: 1374327-1374327 on Build GRCh38
rs767260581 Chr.1: 1374269-1374269 on Build GRCh38
rs780754921 Chr.1: 1374687-1374687 on Build GRCh38
rs774629915 Chr.1: 1374463-1374463 on Build GRCh38
rs983042216 Chr.1: 1374686-1374686 on Build GRCh38
rs996853466 Chr.1: 1374507-1374507 on Build GRCh38
rs750589750 Chr.1: 1374383-1374383 on Build GRCh38
rs183326210 Chr.1: 1374692-1374692 on Build GRCh38
rs927734008 Chr.1: 1374451-1374451 on Build GRCh38
rs781311924 Chr.1: 1374373-1374373 on Build GRCh38
rs879327355 Chr.1: 1374422-1374423 on Build GRCh38
rs750681399 Chr.1: 1374488-1374488 on Build GRCh38
rs1035190902 Chr.1: 1374641-1374641 on Build GRCh38
rs908481637 Chr.1: 1374493-1374493 on Build GRCh38
rs941260468 Chr.1: 1374494-1374494 on Build GRCh38
rs760062990 Chr.1: 1374287-1374287 on Build GRCh38
rs769245026 Chr.1: 1374689-1374689 on Build GRCh38
rs999441880 Chr.1: 1374573-1374573 on Build GRCh38
rs111374341 Chr.1: 1374606-1374606 on Build GRCh38
rs2765036 Chr.1: 1374597-1374597 on Build GRCh38
rs746836043 Chr.1: 1374684-1374684 on Build GRCh38
rs570329874 Chr.1: 1374320-1374320 on Build GRCh38
rs914933701 Chr.1: 1374437-1374437 on Build GRCh38
rs905656646 Chr.1: 1374349-1374349 on Build GRCh38

View Full Product Details