Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1374574-1375066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKAIP1
Entrez Gene ID
54998
Gene Name
aurora kinase A interacting protein 1
Gene Aliases
AIP, AKIP, MRP-S38
UniGene
Hs.632515
Gene Chromosome Location
Chr.1: 1373730-1375438 on Build GRCh38
Transcripts
NM_001127229.1 NM_001127230.1 NM_017900.2 XM_011541670.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363846 c.20C>T p.T7I Chr.1: 1374737-1374737 on Build GRCh38
4494805 c.43C>T p.P15S Chr.1: 1374714-1374714 on Build GRCh38
5434034 c.13C>T p.R5C Chr.1: 1374744-1374744 on Build GRCh38
6268383 c.13C>A p.R5S Chr.1: 1374744-1374744 on Build GRCh38
3801545 c.10G>C p.G4R Chr.1: 1374747-1374747 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs527525962 Chr.1: 1374680-1374680 on Build GRCh38
rs756209362 Chr.1: 1374657-1374657 on Build GRCh38
rs761474608 Chr.1: 1374664-1374664 on Build GRCh38
rs1050612554 Chr.1: 1375041-1375041 on Build GRCh38
rs1051074407 Chr.1: 1374781-1374781 on Build GRCh38
rs1002291060 Chr.1: 1374851-1374851 on Build GRCh38
rs576921931 Chr.1: 1374603-1374604 on Build GRCh38
rs932560682 Chr.1: 1374967-1374967 on Build GRCh38
rs567480343 Chr.1: 1374599-1374599 on Build GRCh38
rs1031144999 Chr.1: 1374632-1374632 on Build GRCh38
rs112533928 Chr.1: 1374801-1374801 on Build GRCh38
rs920379436 Chr.1: 1374786-1374786 on Build GRCh38
rs947546193 Chr.1: 1375034-1375034 on Build GRCh38
rs796766468 Chr.1: 1374968-1374980 on Build GRCh38
rs1053234797 Chr.1: 1374991-1374991 on Build GRCh38
rs761013046 Chr.1: 1374734-1374734 on Build GRCh38
rs980827829 Chr.1: 1374669-1374669 on Build GRCh38
rs962518996 Chr.1: 1374897-1374897 on Build GRCh38
rs748818794 Chr.1: 1374808-1374808 on Build GRCh38
rs141604295 Chr.1: 1374870-1374870 on Build GRCh38
rs370802222 Chr.1: 1374666-1374666 on Build GRCh38
rs1002719732 Chr.1: 1374957-1374962 on Build GRCh38
rs2765035 Chr.1: 1374694-1374694 on Build GRCh38
rs191693192 Chr.1: 1374978-1374978 on Build GRCh38
rs973239541 Chr.1: 1374997-1374997 on Build GRCh38
rs374922558 Chr.1: 1374598-1374598 on Build GRCh38
rs931726124 Chr.1: 1375039-1375039 on Build GRCh38
rs529608234 Chr.1: 1374816-1374816 on Build GRCh38
rs546742050 Chr.1: 1374966-1374966 on Build GRCh38
rs756987148 Chr.1: 1374767-1374767 on Build GRCh38
rs780986023 Chr.1: 1374769-1374769 on Build GRCh38
rs1029650730 Chr.1: 1374600-1374600 on Build GRCh38
rs371071779 Chr.1: 1374758-1374758 on Build GRCh38
rs749781154 Chr.1: 1374691-1374691 on Build GRCh38
rs963048027 Chr.1: 1374968-1374978 on Build GRCh38
rs1044555454 Chr.1: 1375043-1375043 on Build GRCh38
rs1015290794 Chr.1: 1374895-1374895 on Build GRCh38
rs779604187 Chr.1: 1374807-1374807 on Build GRCh38
rs766592469 Chr.1: 1374658-1374658 on Build GRCh38
rs1023215502 Chr.1: 1374605-1374605 on Build GRCh38
rs768010516 Chr.1: 1374713-1374713 on Build GRCh38
rs777599781 Chr.1: 1374671-1374671 on Build GRCh38
rs774776478 Chr.1: 1374693-1374693 on Build GRCh38
rs765980402 Chr.1: 1375032-1375032 on Build GRCh38
rs961846167 Chr.1: 1374992-1374992 on Build GRCh38
rs980423955 Chr.1: 1374637-1374637 on Build GRCh38
rs1023226922 Chr.1: 1374840-1374840 on Build GRCh38
rs893186653 Chr.1: 1375021-1375021 on Build GRCh38
rs751517131 Chr.1: 1374763-1374763 on Build GRCh38
rs1011117158 Chr.1: 1374896-1374896 on Build GRCh38
rs1010960091 Chr.1: 1374648-1374648 on Build GRCh38
rs536418752 Chr.1: 1374911-1374911 on Build GRCh38
rs780754921 Chr.1: 1374687-1374687 on Build GRCh38
rs1034670887 Chr.1: 1374865-1374865 on Build GRCh38
rs983042216 Chr.1: 1374686-1374686 on Build GRCh38
rs1011749967 Chr.1: 1374826-1374826 on Build GRCh38
rs377661377 Chr.1: 1374757-1374757 on Build GRCh38
rs934649083 Chr.1: 1374989-1374989 on Build GRCh38
rs183326210 Chr.1: 1374692-1374692 on Build GRCh38
rs745532255 Chr.1: 1374688-1374688 on Build GRCh38
rs369927790 Chr.1: 1374811-1374811 on Build GRCh38
rs969525728 Chr.1: 1374920-1374920 on Build GRCh38
rs766531279 Chr.1: 1374751-1374751 on Build GRCh38
rs1035190902 Chr.1: 1374641-1374641 on Build GRCh38
rs753910088 Chr.1: 1374753-1374753 on Build GRCh38
rs187632027 Chr.1: 1374788-1374788 on Build GRCh38
rs769245026 Chr.1: 1374689-1374689 on Build GRCh38
rs111374341 Chr.1: 1374606-1374606 on Build GRCh38
rs982583534 Chr.1: 1374883-1374883 on Build GRCh38
rs183589252 Chr.1: 1375036-1375036 on Build GRCh38
rs11548462 Chr.1: 1375049-1375049 on Build GRCh38
rs2765036 Chr.1: 1374597-1374597 on Build GRCh38
rs551967457 Chr.1: 1374980-1374980 on Build GRCh38
rs892597853 Chr.1: 1374887-1374887 on Build GRCh38
rs746836043 Chr.1: 1374684-1374684 on Build GRCh38
rs775021531 Chr.1: 1374690-1374690 on Build GRCh38
rs893342044 Chr.1: 1374821-1374821 on Build GRCh38
rs969083685 Chr.1: 1374617-1374617 on Build GRCh38
rs906081779 Chr.1: 1374841-1374841 on Build GRCh38
rs773522689 Chr.1: 1374731-1374731 on Build GRCh38

View Full Product Details