Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1785900-1786443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB1
Entrez Gene ID
2782
Gene Name
G protein subunit beta 1
Gene Aliases
MRD42
UniGene
Hs.430425
Gene Chromosome Location
Chr.1: 1785285-1891117 on Build GRCh38
Transcripts
NM_001282538.1 NM_001282539.1 NM_002074.4 XM_017001059.1 XM_017001060.1 XM_017001061.1

SNP Detail

SNP ID Chromosome Location
rs1015671632 Chr.1: 1785989-1785989 on Build GRCh38
rs562904611 Chr.1: 1786094-1786094 on Build GRCh38
rs544153544 Chr.1: 1785966-1785966 on Build GRCh38
rs913685265 Chr.1: 1786246-1786246 on Build GRCh38
rs376875581 Chr.1: 1786207-1786207 on Build GRCh38
rs540987144 Chr.1: 1786038-1786038 on Build GRCh38
rs937938399 Chr.1: 1786313-1786313 on Build GRCh38
rs777554737 Chr.1: 1785925-1785925 on Build GRCh38
rs1050086967 Chr.1: 1786360-1786373 on Build GRCh38
rs542859443 Chr.1: 1786187-1786187 on Build GRCh38
rs547054673 Chr.1: 1786260-1786260 on Build GRCh38
rs896293428 Chr.1: 1785947-1785947 on Build GRCh38
rs954300957 Chr.1: 1786277-1786277 on Build GRCh38
rs535701218 Chr.1: 1786377-1786377 on Build GRCh38
rs569843778 Chr.1: 1786326-1786326 on Build GRCh38
rs985301288 Chr.1: 1786194-1786194 on Build GRCh38
rs1043807318 Chr.1: 1786288-1786288 on Build GRCh38
rs759147259 Chr.1: 1786384-1786384 on Build GRCh38
rs892989837 Chr.1: 1786002-1786002 on Build GRCh38
rs960724452 Chr.1: 1786116-1786116 on Build GRCh38
rs1004037033 Chr.1: 1786373-1786373 on Build GRCh38
rs1028758001 Chr.1: 1786393-1786393 on Build GRCh38
rs181158679 Chr.1: 1786316-1786316 on Build GRCh38
rs1002108486 Chr.1: 1785929-1785929 on Build GRCh38
rs1028309080 Chr.1: 1786080-1786080 on Build GRCh38
rs578047957 Chr.1: 1785924-1785924 on Build GRCh38
rs1015648587 Chr.1: 1786192-1786192 on Build GRCh38
rs934321695 Chr.1: 1786289-1786292 on Build GRCh38
rs1011325317 Chr.1: 1786062-1786062 on Build GRCh38
rs369223270 Chr.1: 1786383-1786383 on Build GRCh38
rs755746405 Chr.1: 1786109-1786109 on Build GRCh38
rs139184852 Chr.1: 1786297-1786297 on Build GRCh38
rs532083518 Chr.1: 1786306-1786306 on Build GRCh38
rs573903747 Chr.1: 1785940-1785940 on Build GRCh38
rs780925923 Chr.1: 1785999-1785999 on Build GRCh38
rs1004717403 Chr.1: 1786149-1786149 on Build GRCh38
rs555045136 Chr.1: 1785937-1785937 on Build GRCh38
rs932884099 Chr.1: 1785939-1785939 on Build GRCh38
rs141773285 Chr.1: 1786098-1786098 on Build GRCh38
rs529530296 Chr.1: 1786107-1786107 on Build GRCh38
rs756651439 Chr.1: 1785992-1785992 on Build GRCh38
rs185713603 Chr.1: 1786398-1786398 on Build GRCh38
rs1035453061 Chr.1: 1786110-1786110 on Build GRCh38
rs987057463 Chr.1: 1786287-1786287 on Build GRCh38
rs146251869 Chr.1: 1786279-1786279 on Build GRCh38
rs974274166 Chr.1: 1786214-1786214 on Build GRCh38
rs962761773 Chr.1: 1785990-1785990 on Build GRCh38
rs28479643 Chr.1: 1786165-1786165 on Build GRCh38
rs912745142 Chr.1: 1786131-1786131 on Build GRCh38
rs772627323 Chr.1: 1786378-1786378 on Build GRCh38
rs6659884 Chr.1: 1786409-1786409 on Build GRCh38
rs1057017916 Chr.1: 1786395-1786395 on Build GRCh38
rs974509001 Chr.1: 1786067-1786067 on Build GRCh38
rs1051242773 Chr.1: 1785946-1785946 on Build GRCh38
rs1062363 Chr.1: 1786179-1786179 on Build GRCh38
rs559850838 Chr.1: 1786200-1786200 on Build GRCh38
rs921432044 Chr.1: 1786250-1786250 on Build GRCh38

View Full Product Details