Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1790262-1790785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB1
Entrez Gene ID
2782
Gene Name
G protein subunit beta 1
Gene Aliases
MRD42
UniGene
Hs.430425
Gene Chromosome Location
Chr.1: 1785285-1891117 on Build GRCh38
Transcripts
NM_001282538.1 NM_001282539.1 NM_002074.4 XM_017001059.1 XM_017001060.1 XM_017001061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2075068 c.699C>T p.C233C Chr.1: 1790395-1790395 on Build GRCh38
2075070 c.543C>T p.T181T Chr.1: 1790551-1790551 on Build GRCh38
6990611 c.583G>C p.D195H Chr.1: 1790511-1790511 on Build GRCh38
1336288 c.594G>A p.L198L Chr.1: 1790500-1790500 on Build GRCh38
4025106 c.537G>A p.T179T Chr.1: 1790557-1790557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035297234 Chr.1: 1790472-1790472 on Build GRCh38
rs368538489 Chr.1: 1790603-1790603 on Build GRCh38
rs766426530 Chr.1: 1790395-1790395 on Build GRCh38
rs1039233661 Chr.1: 1790317-1790317 on Build GRCh38
rs149471150 Chr.1: 1790357-1790357 on Build GRCh38
rs767751820 Chr.1: 1790371-1790371 on Build GRCh38
rs768039402 Chr.1: 1790551-1790551 on Build GRCh38
rs760941619 Chr.1: 1790617-1790617 on Build GRCh38
rs1051870035 Chr.1: 1790759-1790759 on Build GRCh38
rs371280820 Chr.1: 1790426-1790426 on Build GRCh38
rs759029020 Chr.1: 1790336-1790336 on Build GRCh38
rs763840033 Chr.1: 1790438-1790438 on Build GRCh38
rs772464654 Chr.1: 1790621-1790622 on Build GRCh38
rs767405075 Chr.1: 1790638-1790638 on Build GRCh38
rs998057803 Chr.1: 1790756-1790756 on Build GRCh38
rs752638794 Chr.1: 1790410-1790410 on Build GRCh38
rs748544570 Chr.1: 1790602-1790602 on Build GRCh38
rs573084848 Chr.1: 1790368-1790368 on Build GRCh38
rs760751848 Chr.1: 1790392-1790392 on Build GRCh38
rs755635323 Chr.1: 1790563-1790563 on Build GRCh38
rs377225069 Chr.1: 1790343-1790343 on Build GRCh38
rs1012399618 Chr.1: 1790724-1790724 on Build GRCh38
rs773385139 Chr.1: 1790376-1790376 on Build GRCh38
rs200569590 Chr.1: 1790564-1790564 on Build GRCh38
rs1002519501 Chr.1: 1790454-1790454 on Build GRCh38
rs759481718 Chr.1: 1790623-1790623 on Build GRCh38
rs950679231 Chr.1: 1790496-1790496 on Build GRCh38
rs900669853 Chr.1: 1790754-1790754 on Build GRCh38
rs773438465 Chr.1: 1790610-1790610 on Build GRCh38
rs1039504140 Chr.1: 1790723-1790723 on Build GRCh38
rs935605355 Chr.1: 1790639-1790639 on Build GRCh38
rs780992762 Chr.1: 1790513-1790513 on Build GRCh38
rs752350823 Chr.1: 1790386-1790386 on Build GRCh38
rs779612138 Chr.1: 1790574-1790574 on Build GRCh38
rs562934218 Chr.1: 1790304-1790304 on Build GRCh38
rs757075718 Chr.1: 1790508-1790508 on Build GRCh38
rs538188811 Chr.1: 1790497-1790497 on Build GRCh38
rs776746204 Chr.1: 1790621-1790621 on Build GRCh38
rs370763835 Chr.1: 1790628-1790628 on Build GRCh38
rs966354864 Chr.1: 1790340-1790340 on Build GRCh38
rs374082962 Chr.1: 1790575-1790575 on Build GRCh38
rs755759467 Chr.1: 1790644-1790644 on Build GRCh38
rs750789639 Chr.1: 1790693-1790693 on Build GRCh38
rs900640979 Chr.1: 1790318-1790318 on Build GRCh38
rs745399405 Chr.1: 1790345-1790345 on Build GRCh38
rs138058014 Chr.1: 1790344-1790344 on Build GRCh38
rs774748088 Chr.1: 1790364-1790364 on Build GRCh38
rs762104024 Chr.1: 1790455-1790455 on Build GRCh38
rs1012414788 Chr.1: 1790369-1790369 on Build GRCh38
rs769280339 Chr.1: 1790346-1790346 on Build GRCh38
rs1051752774 Chr.1: 1790335-1790335 on Build GRCh38
rs186894090 Chr.1: 1790584-1790584 on Build GRCh38
rs887902723 Chr.1: 1790302-1790302 on Build GRCh38
rs997795163 Chr.1: 1790320-1790320 on Build GRCh38
rs377062472 Chr.1: 1790619-1790619 on Build GRCh38
rs758356105 Chr.1: 1790419-1790419 on Build GRCh38
rs776174775 Chr.1: 1790285-1790285 on Build GRCh38
rs568867207 Chr.1: 1790642-1790642 on Build GRCh38
rs781369159 Chr.1: 1790709-1790709 on Build GRCh38
rs909047058 Chr.1: 1790558-1790558 on Build GRCh38
rs982034145 Chr.1: 1790607-1790607 on Build GRCh38
rs143955960 Chr.1: 1790627-1790627 on Build GRCh38
rs775873217 Chr.1: 1790349-1790349 on Build GRCh38
rs76013166 Chr.1: 1790609-1790609 on Build GRCh38
rs537432215 Chr.1: 1790663-1790663 on Build GRCh38
rs761712176 Chr.1: 1790631-1790631 on Build GRCh38
rs1047753391 Chr.1: 1790299-1790299 on Build GRCh38
rs749789231 Chr.1: 1790356-1790356 on Build GRCh38

View Full Product Details