Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1955516-1956031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP74
Entrez Gene ID
85452
Gene Name
cilia and flagella associated protein 74
Gene Aliases
C1orf222, KIAA1751
UniGene
Hs.232092
Gene Chromosome Location
Chr.1: 1921951-2003837 on Build GRCh38
Transcripts
NM_001304360.1 XM_011542333.2 XM_011542335.1 XM_011542336.2 XM_017002641.1 XM_017002642.1 XM_017002643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7322450 c.2126G>T p.R709L Chr.1: 1955741-1955741 on Build GRCh38
7306093 c.2210C>T p.A737V Chr.1: 1955657-1955657 on Build GRCh38
1255750 c.2187C>T p.G729G Chr.1: 1955680-1955680 on Build GRCh38
4998933 c.2069G>A p.S690N Chr.1: 1955798-1955798 on Build GRCh38
1192790 c.2214delC p.G739fs*>24 Chr.1: 1955653-1955653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746111410 Chr.1: 1955810-1955810 on Build GRCh38
rs758368682 Chr.1: 1955757-1955757 on Build GRCh38
rs778214674 Chr.1: 1955602-1955602 on Build GRCh38
rs745996385 Chr.1: 1955722-1955722 on Build GRCh38
rs536945147 Chr.1: 1955859-1955859 on Build GRCh38
rs139735565 Chr.1: 1955653-1955653 on Build GRCh38
rs773029054 Chr.1: 1955750-1955750 on Build GRCh38
rs114413755 Chr.1: 1955965-1955965 on Build GRCh38
rs778228109 Chr.1: 1955668-1955668 on Build GRCh38
rs757642446 Chr.1: 1955878-1955878 on Build GRCh38
rs762548045 Chr.1: 1955870-1955870 on Build GRCh38
rs753259412 Chr.1: 1955686-1955686 on Build GRCh38
rs376487653 Chr.1: 1956001-1956001 on Build GRCh38
rs552997945 Chr.1: 1955958-1955958 on Build GRCh38
rs1015084851 Chr.1: 1955587-1955587 on Build GRCh38
rs1013991577 Chr.1: 1955798-1955798 on Build GRCh38
rs750092290 Chr.1: 1955561-1955561 on Build GRCh38
rs375375631 Chr.1: 1955863-1955863 on Build GRCh38
rs913046526 Chr.1: 1955709-1955709 on Build GRCh38
rs201687693 Chr.1: 1955756-1955756 on Build GRCh38
rs759044054 Chr.1: 1955764-1955764 on Build GRCh38
rs913988305 Chr.1: 1955900-1955900 on Build GRCh38
rs1021520194 Chr.1: 1955554-1955554 on Build GRCh38
rs1055905600 Chr.1: 1955594-1955594 on Build GRCh38
rs1042926098 Chr.1: 1955742-1955742 on Build GRCh38
rs772062184 Chr.1: 1955828-1955828 on Build GRCh38
rs181015303 Chr.1: 1955698-1955698 on Build GRCh38
rs199780644 Chr.1: 1955660-1955660 on Build GRCh38
rs776358837 Chr.1: 1955759-1955759 on Build GRCh38
rs764667105 Chr.1: 1955781-1955781 on Build GRCh38
rs748580301 Chr.1: 1955598-1955598 on Build GRCh38
rs773116789 Chr.1: 1955829-1955829 on Build GRCh38
rs776247816 Chr.1: 1955833-1955833 on Build GRCh38
rs545149274 Chr.1: 1955989-1955989 on Build GRCh38
rs796266880 Chr.1: 1955674-1955674 on Build GRCh38
rs779467159 Chr.1: 1955585-1955585 on Build GRCh38
rs1014353136 Chr.1: 1955998-1955998 on Build GRCh38
rs201482356 Chr.1: 1955639-1955639 on Build GRCh38
rs562842260 Chr.1: 1955657-1955657 on Build GRCh38
rs748119135 Chr.1: 1955737-1955737 on Build GRCh38
rs374463126 Chr.1: 1955896-1955896 on Build GRCh38
rs771972359 Chr.1: 1955741-1955741 on Build GRCh38
rs115884258 Chr.1: 1955861-1955861 on Build GRCh38
rs367731550 Chr.1: 1955808-1955808 on Build GRCh38
rs571138811 Chr.1: 1955847-1955847 on Build GRCh38
rs764071575 Chr.1: 1955548-1955548 on Build GRCh38
rs913788418 Chr.1: 1955748-1955748 on Build GRCh38
rs3838976 Chr.1: 1955653-1955653 on Build GRCh38
rs774261814 Chr.1: 1955652-1955652 on Build GRCh38
rs987924391 Chr.1: 1955915-1955915 on Build GRCh38
rs776528035 Chr.1: 1955910-1955910 on Build GRCh38
rs747357991 Chr.1: 1955703-1955703 on Build GRCh38
rs760509301 Chr.1: 1955751-1955751 on Build GRCh38
rs895462335 Chr.1: 1955974-1955974 on Build GRCh38
rs752124090 Chr.1: 1955876-1955876 on Build GRCh38
rs761595925 Chr.1: 1955557-1955557 on Build GRCh38
rs199689924 Chr.1: 1955805-1955805 on Build GRCh38
rs534476358 Chr.1: 1955727-1955727 on Build GRCh38
rs767306505 Chr.1: 1955656-1955656 on Build GRCh38
rs1034479295 Chr.1: 1955559-1955559 on Build GRCh38
rs928758106 Chr.1: 1955885-1955885 on Build GRCh38
rs925827096 Chr.1: 1955937-1955937 on Build GRCh38
rs781473349 Chr.1: 1955719-1955719 on Build GRCh38
rs28575980 Chr.1: 1955806-1955806 on Build GRCh38
rs747264122 Chr.1: 1955607-1955607 on Build GRCh38
rs962271529 Chr.1: 1955649-1955649 on Build GRCh38
rs564430420 Chr.1: 1955567-1955567 on Build GRCh38
rs1056250926 Chr.1: 1955794-1955794 on Build GRCh38
rs753364651 Chr.1: 1955576-1955576 on Build GRCh38
rs959818049 Chr.1: 1955572-1955572 on Build GRCh38
rs200358578 Chr.1: 1955880-1955880 on Build GRCh38
rs751351968 Chr.1: 1955556-1955556 on Build GRCh38
rs3838975 Chr.1: 1955673-1955673 on Build GRCh38
rs973630703 Chr.1: 1955599-1955599 on Build GRCh38
rs937251709 Chr.1: 1955946-1955946 on Build GRCh38
rs188111949 Chr.1: 1955616-1955616 on Build GRCh38
rs764869467 Chr.1: 1955770-1955770 on Build GRCh38
rs937887162 Chr.1: 1955785-1955785 on Build GRCh38
rs755699694 Chr.1: 1955571-1955571 on Build GRCh38
rs752037325 Chr.1: 1955796-1955796 on Build GRCh38
rs1055461649 Chr.1: 1955767-1955767 on Build GRCh38
rs371520785 Chr.1: 1955832-1955832 on Build GRCh38
rs775484041 Chr.1: 1955732-1955732 on Build GRCh38
rs750707653 Chr.1: 1955892-1955892 on Build GRCh38
rs986272099 Chr.1: 1955673-1955673 on Build GRCh38
rs28548017 Chr.1: 1955580-1955580 on Build GRCh38
rs753351957 Chr.1: 1955661-1955661 on Build GRCh38
rs561538566 Chr.1: 1955931-1955931 on Build GRCh38
rs759630522 Chr.1: 1955627-1955627 on Build GRCh38
rs377168472 Chr.1: 1955711-1955711 on Build GRCh38
rs780132668 Chr.1: 1955822-1955822 on Build GRCh38
rs200281905 Chr.1: 1955610-1955610 on Build GRCh38
rs535891538 Chr.1: 1955924-1955924 on Build GRCh38
rs370914985 Chr.1: 1955755-1955755 on Build GRCh38
rs758932249 Chr.1: 1955693-1955693 on Build GRCh38
rs759024199 Chr.1: 1955779-1955779 on Build GRCh38
rs565474492 Chr.1: 1956000-1956000 on Build GRCh38
rs767218755 Chr.1: 1955558-1955558 on Build GRCh38
rs895790978 Chr.1: 1955601-1955601 on Build GRCh38
rs368272325 Chr.1: 1955771-1955771 on Build GRCh38
rs962230662 Chr.1: 1955591-1955591 on Build GRCh38
rs764756868 Chr.1: 1955873-1955873 on Build GRCh38
rs756253346 Chr.1: 1955818-1955818 on Build GRCh38
rs573532536 Chr.1: 1955916-1955916 on Build GRCh38
rs749332493 Chr.1: 1955826-1955826 on Build GRCh38

View Full Product Details