Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2025396-2025898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRD
Entrez Gene ID
2563
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Gene Aliases
EIG10, EJM7, GEFSP5
UniGene
Hs.113882
Gene Chromosome Location
Chr.1: 2019329-2030753 on Build GRCh38
Transcripts
NM_000815.4 XM_011541194.2 XM_017000936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2124508 c.434G>T p.R145L Chr.1: 2025702-2025702 on Build GRCh38
4026433 c.470G>A p.R157Q Chr.1: 2025738-2025738 on Build GRCh38
6668626 c.448G>A p.G150S Chr.1: 2025716-2025716 on Build GRCh38
4135894 c.333G>A p.L111L Chr.1: 2025601-2025601 on Build GRCh38
424897 c.330T>C p.G110G Chr.1: 2025598-2025598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045485424 Chr.1: 2025747-2025747 on Build GRCh38
rs368857204 Chr.1: 2025765-2025765 on Build GRCh38
rs758319768 Chr.1: 2025447-2025447 on Build GRCh38
rs760581006 Chr.1: 2025748-2025748 on Build GRCh38
rs775423651 Chr.1: 2025737-2025737 on Build GRCh38
rs369289695 Chr.1: 2025757-2025757 on Build GRCh38
rs772285691 Chr.1: 2025508-2025508 on Build GRCh38
rs769972222 Chr.1: 2025721-2025721 on Build GRCh38
rs535570779 Chr.1: 2025769-2025769 on Build GRCh38
rs576021048 Chr.1: 2025857-2025857 on Build GRCh38
rs752680558 Chr.1: 2025442-2025442 on Build GRCh38
rs539748394 Chr.1: 2025842-2025842 on Build GRCh38
rs759099296 Chr.1: 2025755-2025755 on Build GRCh38
rs971778213 Chr.1: 2025849-2025849 on Build GRCh38
rs748573641 Chr.1: 2025603-2025603 on Build GRCh38
rs746064703 Chr.1: 2025719-2025719 on Build GRCh38
rs533947004 Chr.1: 2025790-2025790 on Build GRCh38
rs533453337 Chr.1: 2025745-2025745 on Build GRCh38
rs773326244 Chr.1: 2025787-2025787 on Build GRCh38
rs774665033 Chr.1: 2025498-2025498 on Build GRCh38
rs1050775980 Chr.1: 2025864-2025864 on Build GRCh38
rs760866031 Chr.1: 2025520-2025520 on Build GRCh38
rs767409551 Chr.1: 2025652-2025652 on Build GRCh38
rs772538793 Chr.1: 2025604-2025604 on Build GRCh38
rs3831907 Chr.1: 2025781-2025781 on Build GRCh38
rs41306994 Chr.1: 2025866-2025866 on Build GRCh38
rs531305926 Chr.1: 2025867-2025867 on Build GRCh38
rs776785350 Chr.1: 2025632-2025632 on Build GRCh38
rs372469742 Chr.1: 2025853-2025853 on Build GRCh38
rs1045343810 Chr.1: 2025496-2025496 on Build GRCh38
rs935406386 Chr.1: 2025433-2025433 on Build GRCh38
rs201900264 Chr.1: 2025431-2025431 on Build GRCh38
rs550493196 Chr.1: 2025868-2025868 on Build GRCh38
rs201306411 Chr.1: 2025660-2025660 on Build GRCh38
rs897992927 Chr.1: 2025551-2025551 on Build GRCh38
rs561793868 Chr.1: 2025862-2025862 on Build GRCh38
rs978679181 Chr.1: 2025685-2025685 on Build GRCh38
rs758066086 Chr.1: 2025751-2025751 on Build GRCh38
rs376701444 Chr.1: 2025613-2025613 on Build GRCh38
rs149832567 Chr.1: 2025586-2025586 on Build GRCh38
rs376054712 Chr.1: 2025701-2025701 on Build GRCh38
rs549716183 Chr.1: 2025756-2025756 on Build GRCh38
rs1009966197 Chr.1: 2025870-2025870 on Build GRCh38
rs766154975 Chr.1: 2025664-2025664 on Build GRCh38
rs554024339 Chr.1: 2025825-2025825 on Build GRCh38
rs781520848 Chr.1: 2025772-2025772 on Build GRCh38
rs1021476356 Chr.1: 2025792-2025792 on Build GRCh38
rs916882204 Chr.1: 2025830-2025830 on Build GRCh38
rs766416878 Chr.1: 2025436-2025436 on Build GRCh38
rs372728395 Chr.1: 2025758-2025758 on Build GRCh38
rs899554874 Chr.1: 2025863-2025864 on Build GRCh38
rs371111781 Chr.1: 2025505-2025505 on Build GRCh38
rs763019253 Chr.1: 2025738-2025738 on Build GRCh38
rs201757318 Chr.1: 2025676-2025676 on Build GRCh38
rs373508468 Chr.1: 2025531-2025531 on Build GRCh38
rs754491872 Chr.1: 2025593-2025593 on Build GRCh38
rs562124721 Chr.1: 2025647-2025647 on Build GRCh38
rs766248295 Chr.1: 2025753-2025753 on Build GRCh38
rs922412708 Chr.1: 2025429-2025429 on Build GRCh38
rs775338230 Chr.1: 2025715-2025715 on Build GRCh38
rs2229110 Chr.1: 2025598-2025598 on Build GRCh38
rs113553794 Chr.1: 2025451-2025451 on Build GRCh38
rs377333949 Chr.1: 2025432-2025432 on Build GRCh38
rs916955416 Chr.1: 2025718-2025718 on Build GRCh38
rs779789979 Chr.1: 2025582-2025582 on Build GRCh38
rs747299306 Chr.1: 2025611-2025611 on Build GRCh38
rs753305525 Chr.1: 2025691-2025691 on Build GRCh38
rs549493166 Chr.1: 2025780-2025780 on Build GRCh38
rs989550711 Chr.1: 2025803-2025803 on Build GRCh38
rs1047338761 Chr.1: 2025841-2025841 on Build GRCh38
rs77892827 Chr.1: 2025682-2025682 on Build GRCh38
rs767191610 Chr.1: 2025564-2025564 on Build GRCh38
rs750144283 Chr.1: 2025574-2025574 on Build GRCh38
rs988225589 Chr.1: 2025668-2025668 on Build GRCh38
rs937027520 Chr.1: 2025485-2025485 on Build GRCh38
rs1032882308 Chr.1: 2025804-2025804 on Build GRCh38
rs978989704 Chr.1: 2025795-2025795 on Build GRCh38
rs373729382 Chr.1: 2025480-2025480 on Build GRCh38
rs773525730 Chr.1: 2025515-2025515 on Build GRCh38
rs376672448 Chr.1: 2025430-2025430 on Build GRCh38
rs777154372 Chr.1: 2025764-2025765 on Build GRCh38
rs1007853733 Chr.1: 2025767-2025767 on Build GRCh38
rs866724003 Chr.1: 2025793-2025793 on Build GRCh38
rs948282222 Chr.1: 2025488-2025488 on Build GRCh38
rs554139861 Chr.1: 2025478-2025478 on Build GRCh38
rs202225187 Chr.1: 2025744-2025744 on Build GRCh38
rs778284512 Chr.1: 2025764-2025764 on Build GRCh38
rs377536787 Chr.1: 2025535-2025535 on Build GRCh38
rs746153181 Chr.1: 2025773-2025773 on Build GRCh38
rs200593470 Chr.1: 2025532-2025532 on Build GRCh38
rs371136891 Chr.1: 2025466-2025466 on Build GRCh38
rs114015660 Chr.1: 2025448-2025448 on Build GRCh38
rs369516333 Chr.1: 2025865-2025865 on Build GRCh38
rs765199464 Chr.1: 2025646-2025646 on Build GRCh38
rs781431062 Chr.1: 2025716-2025716 on Build GRCh38
rs918985978 Chr.1: 2025856-2025856 on Build GRCh38
rs949738184 Chr.1: 2025840-2025840 on Build GRCh38
rs755729863 Chr.1: 2025479-2025479 on Build GRCh38
rs755920072 Chr.1: 2025661-2025661 on Build GRCh38
rs886818332 Chr.1: 2025554-2025554 on Build GRCh38
rs773435504 Chr.1: 2025609-2025609 on Build GRCh38
rs3820007 Chr.1: 2025860-2025860 on Build GRCh38
rs568142217 Chr.1: 2025781-2025781 on Build GRCh38
rs1007549925 Chr.1: 2025614-2025614 on Build GRCh38
rs767173604 Chr.1: 2025490-2025491 on Build GRCh38
rs577169800 Chr.1: 2025826-2025826 on Build GRCh38
rs746967044 Chr.1: 2025452-2025452 on Build GRCh38
rs895558456 Chr.1: 2025502-2025502 on Build GRCh38
rs755819827 Chr.1: 2025575-2025575 on Build GRCh38
rs376827990 Chr.1: 2025495-2025495 on Build GRCh38
rs769909295 Chr.1: 2025779-2025779 on Build GRCh38
rs949766842 Chr.1: 2025735-2025735 on Build GRCh38
rs746471597 Chr.1: 2025778-2025778 on Build GRCh38
rs559765850 Chr.1: 2025702-2025702 on Build GRCh38
rs370931748 Chr.1: 2025542-2025542 on Build GRCh38
rs555875907 Chr.1: 2025843-2025843 on Build GRCh38
rs997009304 Chr.1: 2025864-2025864 on Build GRCh38
rs61742636 Chr.1: 2025673-2025673 on Build GRCh38
rs751411266 Chr.1: 2025555-2025555 on Build GRCh38
rs764113441 Chr.1: 2025541-2025541 on Build GRCh38
rs373608463 Chr.1: 2025712-2025712 on Build GRCh38
rs545154696 Chr.1: 2025789-2025789 on Build GRCh38
rs1019524374 Chr.1: 2025634-2025634 on Build GRCh38
rs1016544646 Chr.1: 2025580-2025580 on Build GRCh38
rs780927959 Chr.1: 2025470-2025470 on Build GRCh38
rs368774023 Chr.1: 2025640-2025640 on Build GRCh38

View Full Product Details