Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134145857-134146368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAM3
Entrez Gene ID
83700
Gene Name
junctional adhesion molecule 3
Gene Aliases
JAM-2, JAM-3, JAM-C, JAMC
UniGene
Hs.150718
Gene Chromosome Location
Chr.11: 134068925-134151757 on Build GRCh38
Transcripts
NM_001205329.1 NM_032801.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5776465 c.847+5G>A p.? Chr.11: 134146050-134146050 on Build GRCh38
925143 c.807C>T p.D269D Chr.11: 134146005-134146005 on Build GRCh38
6277812 c.753C>T p.F251F Chr.11: 134145951-134145951 on Build GRCh38
5682740 c.825T>C p.C275C Chr.11: 134146023-134146023 on Build GRCh38
6262795 c.817G>T p.A273S Chr.11: 134146015-134146015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770801809 Chr.11: 134146013-134146013 on Build GRCh38
rs397515438 Chr.11: 134145989-134145989 on Build GRCh38
rs370380432 Chr.11: 134146111-134146113 on Build GRCh38
rs765351565 Chr.11: 134146308-134146308 on Build GRCh38
rs368054070 Chr.11: 134146091-134146091 on Build GRCh38
rs570809084 Chr.11: 134146336-134146336 on Build GRCh38
rs768547979 Chr.11: 134145923-134145926 on Build GRCh38
rs921248347 Chr.11: 134146338-134146338 on Build GRCh38
rs554553258 Chr.11: 134146225-134146225 on Build GRCh38
rs549758554 Chr.11: 134146286-134146286 on Build GRCh38
rs373764298 Chr.11: 134146030-134146030 on Build GRCh38
rs915393716 Chr.11: 134146310-134146310 on Build GRCh38
rs745601586 Chr.11: 134145899-134145899 on Build GRCh38
rs563938968 Chr.11: 134146076-134146076 on Build GRCh38
rs58925654 Chr.11: 134146278-134146278 on Build GRCh38
rs1012976809 Chr.11: 134146201-134146201 on Build GRCh38
rs1018838584 Chr.11: 134146223-134146223 on Build GRCh38
rs77908320 Chr.11: 134146141-134146141 on Build GRCh38
rs780634235 Chr.11: 134145910-134145910 on Build GRCh38
rs562940931 Chr.11: 134146140-134146140 on Build GRCh38
rs1027892151 Chr.11: 134146256-134146256 on Build GRCh38
rs374005512 Chr.11: 134146075-134146075 on Build GRCh38
rs763469878 Chr.11: 134146058-134146058 on Build GRCh38
rs552685102 Chr.11: 134146089-134146089 on Build GRCh38
rs773197647 Chr.11: 134146057-134146057 on Build GRCh38
rs762642857 Chr.11: 134145928-134145928 on Build GRCh38
rs776634938 Chr.11: 134146054-134146055 on Build GRCh38
rs758833938 Chr.11: 134145967-134145967 on Build GRCh38
rs199892991 Chr.11: 134146078-134146078 on Build GRCh38
rs568088101 Chr.11: 134146189-134146189 on Build GRCh38
rs776699779 Chr.11: 134146039-134146039 on Build GRCh38
rs888770916 Chr.11: 134146213-134146213 on Build GRCh38
rs745701102 Chr.11: 134146021-134146021 on Build GRCh38
rs753941001 Chr.11: 134145948-134145948 on Build GRCh38
rs893550867 Chr.11: 134145970-134145970 on Build GRCh38
rs747651494 Chr.11: 134145992-134145992 on Build GRCh38
rs951153870 Chr.11: 134145893-134145893 on Build GRCh38
rs773266699 Chr.11: 134145922-134145922 on Build GRCh38
rs770697848 Chr.11: 134145929-134145929 on Build GRCh38
rs1000117499 Chr.11: 134145943-134145943 on Build GRCh38
rs778785861 Chr.11: 134146083-134146083 on Build GRCh38
rs11223714 Chr.11: 134146116-134146116 on Build GRCh38
rs148544487 Chr.11: 134145946-134145946 on Build GRCh38
rs1023690023 Chr.11: 134146284-134146284 on Build GRCh38
rs564450384 Chr.11: 134146228-134146228 on Build GRCh38
rs536563329 Chr.11: 134146207-134146207 on Build GRCh38
rs769932418 Chr.11: 134146094-134146094 on Build GRCh38
rs984201852 Chr.11: 134146108-134146108 on Build GRCh38
rs200823092 Chr.11: 134146024-134146024 on Build GRCh38
rs768330788 Chr.11: 134145908-134145908 on Build GRCh38
rs1053042541 Chr.11: 134146165-134146165 on Build GRCh38
rs1025686354 Chr.11: 134146093-134146093 on Build GRCh38
rs184817716 Chr.11: 134146260-134146260 on Build GRCh38
rs998370619 Chr.11: 134146132-134146132 on Build GRCh38
rs935575201 Chr.11: 134146306-134146306 on Build GRCh38
rs952805096 Chr.11: 134146098-134146098 on Build GRCh38
rs148216858 Chr.11: 134145972-134145972 on Build GRCh38
rs1046656742 Chr.11: 134145892-134145892 on Build GRCh38
rs141227203 Chr.11: 134146035-134146035 on Build GRCh38
rs749178421 Chr.11: 134146007-134146007 on Build GRCh38
rs768080269 Chr.11: 134146174-134146174 on Build GRCh38
rs948086715 Chr.11: 134146119-134146119 on Build GRCh38
rs374704989 Chr.11: 134146088-134146088 on Build GRCh38
rs769687465 Chr.11: 134145921-134145921 on Build GRCh38
rs988258148 Chr.11: 134146282-134146282 on Build GRCh38
rs201299164 Chr.11: 134145990-134145990 on Build GRCh38
rs755401442 Chr.11: 134146072-134146072 on Build GRCh38
rs1050195585 Chr.11: 134146199-134146199 on Build GRCh38
rs765652742 Chr.11: 134145956-134145956 on Build GRCh38
rs751888935 Chr.11: 134146067-134146067 on Build GRCh38
rs201213184 Chr.11: 134146005-134146005 on Build GRCh38
rs751892763 Chr.11: 134145978-134145978 on Build GRCh38
rs755193592 Chr.11: 134146290-134146290 on Build GRCh38
rs762053520 Chr.11: 134145954-134145954 on Build GRCh38
rs755679060 Chr.11: 134145995-134145995 on Build GRCh38
rs558742872 Chr.11: 134146299-134146299 on Build GRCh38
rs915798823 Chr.11: 134145969-134145969 on Build GRCh38
rs371949998 Chr.11: 134146084-134146084 on Build GRCh38
rs1008580249 Chr.11: 134146219-134146219 on Build GRCh38
rs928205744 Chr.11: 134146304-134146304 on Build GRCh38
rs1039985108 Chr.11: 134146117-134146117 on Build GRCh38
rs376942013 Chr.11: 134146061-134146061 on Build GRCh38
rs369456826 Chr.11: 134146136-134146136 on Build GRCh38
rs1040129144 Chr.11: 134146303-134146303 on Build GRCh38
rs377681822 Chr.11: 134145916-134145916 on Build GRCh38
rs946897029 Chr.11: 134146318-134146318 on Build GRCh38
rs773861809 Chr.11: 134146193-134146193 on Build GRCh38
rs924441125 Chr.11: 134146277-134146277 on Build GRCh38
rs748341169 Chr.11: 134146092-134146092 on Build GRCh38
rs745648231 Chr.11: 134146087-134146087 on Build GRCh38
rs754605284 Chr.11: 134145984-134145984 on Build GRCh38
rs774144223 Chr.11: 134146015-134146015 on Build GRCh38
rs191505825 Chr.11: 134145934-134145934 on Build GRCh38
rs532183964 Chr.11: 134145887-134145887 on Build GRCh38
rs750888879 Chr.11: 134145957-134145957 on Build GRCh38
rs774017835 Chr.11: 134145930-134145930 on Build GRCh38
rs777054864 Chr.11: 134146027-134146027 on Build GRCh38
rs111246151 Chr.11: 134146118-134146118 on Build GRCh38
rs971314702 Chr.11: 134146257-134146257 on Build GRCh38
rs201197644 Chr.11: 134146074-134146074 on Build GRCh38

View Full Product Details