Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2858793-2859302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXM1
Entrez Gene ID
2305
Gene Name
forkhead box M1
Gene Aliases
FKHL16, FOXM1B, HFH-11, HFH11, HNF-3, INS-1, MPHOSPH2, MPP-2, MPP2, PIG29, TRIDENT
UniGene
Hs.239
Gene Chromosome Location
Chr.12: 2857681-2877155 on Build GRCh38
Transcripts
NM_001243088.1 NM_001243089.1 NM_021953.3 NM_202002.2 NM_202003.2 XM_005253676.3 XM_011520930.2 XM_011520931.2 XM_011520934.2 NR_038933.1

Gene Symbol
LOC100507424
Entrez Gene ID
100507424
Gene Name
uncharacterized LOC100507424
Gene Aliases
-
UniGene
Hs.239
Gene Chromosome Location
Chr.12: 2836816-2859795 on Build GRCh38
Transcripts
NM_001243088.1 NM_001243089.1 NM_021953.3 NM_202002.2 NM_202003.2 XM_005253676.3 XM_011520930.2 XM_011520931.2 XM_011520934.2 NR_038933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4685417 c.1822C>T p.R608C Chr.12: 2859222-2859222 on Build GRCh38
3688161 c.1747C>T p.R583W Chr.12: 2859297-2859297 on Build GRCh38
5925678 c.1886C>T p.S629F Chr.12: 2859158-2859158 on Build GRCh38
3811820 c.1976C>T p.P659L Chr.12: 2859068-2859068 on Build GRCh38
4846046 c.1878C>G p.L626L Chr.12: 2859166-2859166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759166699 Chr.12: 2859043-2859043 on Build GRCh38
rs138730141 Chr.12: 2858894-2858894 on Build GRCh38
rs778653164 Chr.12: 2859259-2859259 on Build GRCh38
rs28919869 Chr.12: 2858924-2858924 on Build GRCh38
rs114608193 Chr.12: 2859265-2859265 on Build GRCh38
rs555600796 Chr.12: 2859173-2859173 on Build GRCh38
rs757898539 Chr.12: 2858830-2858830 on Build GRCh38
rs760464820 Chr.12: 2859224-2859224 on Build GRCh38
rs999276165 Chr.12: 2859104-2859104 on Build GRCh38
rs997222743 Chr.12: 2859112-2859112 on Build GRCh38
rs3742076 Chr.12: 2859003-2859003 on Build GRCh38
rs141697908 Chr.12: 2858974-2858974 on Build GRCh38
rs926667225 Chr.12: 2858990-2858990 on Build GRCh38
rs775398816 Chr.12: 2858856-2858856 on Build GRCh38
rs535471991 Chr.12: 2859149-2859149 on Build GRCh38
rs201504517 Chr.12: 2859050-2859050 on Build GRCh38
rs548147648 Chr.12: 2858822-2858822 on Build GRCh38
rs568377566 Chr.12: 2858933-2858933 on Build GRCh38
rs903578380 Chr.12: 2859266-2859266 on Build GRCh38
rs745897759 Chr.12: 2858975-2858975 on Build GRCh38
rs759192397 Chr.12: 2859128-2859128 on Build GRCh38
rs746630510 Chr.12: 2859269-2859269 on Build GRCh38
rs749330783 Chr.12: 2858833-2858833 on Build GRCh38
rs374998139 Chr.12: 2859049-2859049 on Build GRCh38
rs765766328 Chr.12: 2859133-2859133 on Build GRCh38
rs751169233 Chr.12: 2858897-2858897 on Build GRCh38
rs749656919 Chr.12: 2858926-2858926 on Build GRCh38
rs781380281 Chr.12: 2858883-2858883 on Build GRCh38
rs202090024 Chr.12: 2859097-2859097 on Build GRCh38
rs747232406 Chr.12: 2859094-2859094 on Build GRCh38
rs780259609 Chr.12: 2858984-2858984 on Build GRCh38
rs1042306150 Chr.12: 2859019-2859019 on Build GRCh38
rs753788611 Chr.12: 2858871-2858871 on Build GRCh38
rs773778648 Chr.12: 2859127-2859127 on Build GRCh38
rs189481330 Chr.12: 2858928-2858928 on Build GRCh38
rs770458870 Chr.12: 2859126-2859126 on Build GRCh38
rs767233350 Chr.12: 2859129-2859129 on Build GRCh38
rs995881166 Chr.12: 2858905-2858905 on Build GRCh38
rs755898214 Chr.12: 2859187-2859187 on Build GRCh38
rs768487105 Chr.12: 2859002-2859002 on Build GRCh38
rs144897055 Chr.12: 2858836-2858836 on Build GRCh38
rs544627443 Chr.12: 2859202-2859202 on Build GRCh38
rs112974593 Chr.12: 2859261-2859261 on Build GRCh38
rs944297456 Chr.12: 2859004-2859004 on Build GRCh38
rs903678903 Chr.12: 2858918-2858918 on Build GRCh38
rs748643345 Chr.12: 2858983-2858983 on Build GRCh38
rs374013446 Chr.12: 2859222-2859222 on Build GRCh38
rs760924577 Chr.12: 2859145-2859145 on Build GRCh38
rs780209186 Chr.12: 2858970-2858970 on Build GRCh38
rs759543718 Chr.12: 2859248-2859248 on Build GRCh38
rs769677621 Chr.12: 2859013-2859013 on Build GRCh38
rs764617916 Chr.12: 2858957-2858957 on Build GRCh38
rs771889374 Chr.12: 2858947-2858947 on Build GRCh38
rs369897716 Chr.12: 2859131-2859131 on Build GRCh38
rs760422215 Chr.12: 2858857-2858857 on Build GRCh38
rs753533687 Chr.12: 2859256-2859256 on Build GRCh38
rs753064494 Chr.12: 2858888-2858888 on Build GRCh38
rs144159370 Chr.12: 2859180-2859180 on Build GRCh38
rs779453012 Chr.12: 2858934-2858934 on Build GRCh38
rs903630934 Chr.12: 2859100-2859100 on Build GRCh38
rs745319066 Chr.12: 2859214-2859214 on Build GRCh38
rs764795562 Chr.12: 2858875-2858875 on Build GRCh38
rs751554328 Chr.12: 2859159-2859159 on Build GRCh38
rs200806960 Chr.12: 2858816-2858816 on Build GRCh38
rs912730887 Chr.12: 2859221-2859221 on Build GRCh38
rs749814710 Chr.12: 2858911-2858911 on Build GRCh38
rs766746704 Chr.12: 2859245-2859245 on Build GRCh38
rs758503889 Chr.12: 2859069-2859069 on Build GRCh38
rs750901706 Chr.12: 2859068-2859068 on Build GRCh38
rs545895205 Chr.12: 2859275-2859275 on Build GRCh38
rs779761386 Chr.12: 2859268-2859268 on Build GRCh38
rs747889767 Chr.12: 2859111-2859111 on Build GRCh38
rs556861533 Chr.12: 2859277-2859277 on Build GRCh38
rs752120067 Chr.12: 2859246-2859246 on Build GRCh38
rs150443540 Chr.12: 2858923-2858923 on Build GRCh38
rs770809844 Chr.12: 2859203-2859203 on Build GRCh38
rs778937651 Chr.12: 2859209-2859209 on Build GRCh38
rs759900571 Chr.12: 2858839-2858839 on Build GRCh38
rs757884959 Chr.12: 2858972-2858972 on Build GRCh38
rs771746117 Chr.12: 2858849-2858849 on Build GRCh38
rs139471742 Chr.12: 2859015-2859015 on Build GRCh38
rs754379620 Chr.12: 2858960-2858960 on Build GRCh38
rs369158837 Chr.12: 2858835-2858835 on Build GRCh38
rs758119183 Chr.12: 2858877-2858877 on Build GRCh38
rs778481065 Chr.12: 2858814-2858814 on Build GRCh38
rs1017404133 Chr.12: 2859081-2859081 on Build GRCh38
rs751184218 Chr.12: 2858950-2858950 on Build GRCh38
rs757721801 Chr.12: 2859055-2859055 on Build GRCh38
rs376414243 Chr.12: 2858916-2858916 on Build GRCh38
rs778172822 Chr.12: 2858831-2858831 on Build GRCh38
rs375821639 Chr.12: 2859213-2859213 on Build GRCh38
rs199632659 Chr.12: 2859116-2859116 on Build GRCh38
rs941533989 Chr.12: 2858978-2858978 on Build GRCh38
rs748580011 Chr.12: 2858821-2858821 on Build GRCh38
rs769587730 Chr.12: 2859115-2859115 on Build GRCh38
rs374220211 Chr.12: 2858955-2858955 on Build GRCh38
rs28919870 Chr.12: 2858912-2858912 on Build GRCh38
rs147746152 Chr.12: 2859226-2859226 on Build GRCh38
rs201149076 Chr.12: 2859236-2859236 on Build GRCh38
rs762144757 Chr.12: 2858999-2858999 on Build GRCh38
rs200635586 Chr.12: 2859255-2859255 on Build GRCh38
rs138298327 Chr.12: 2859082-2859082 on Build GRCh38
rs779597736 Chr.12: 2858973-2858973 on Build GRCh38
rs778283602 Chr.12: 2858899-2858899 on Build GRCh38
rs757092453 Chr.12: 2859257-2859257 on Build GRCh38
rs113780841 Chr.12: 2858872-2858872 on Build GRCh38
rs201630079 Chr.12: 2858815-2858815 on Build GRCh38
rs563190488 Chr.12: 2858921-2858921 on Build GRCh38
rs758423768 Chr.12: 2858981-2858981 on Build GRCh38
rs765784897 Chr.12: 2859056-2859056 on Build GRCh38
rs767649634 Chr.12: 2859250-2859250 on Build GRCh38
rs766451213 Chr.12: 2859155-2859155 on Build GRCh38
rs749248133 Chr.12: 2859122-2859122 on Build GRCh38
rs745702333 Chr.12: 2858921-2858922 on Build GRCh38
rs770372738 Chr.12: 2858828-2858828 on Build GRCh38
rs765810099 Chr.12: 2859016-2859016 on Build GRCh38
rs764047099 Chr.12: 2858869-2858869 on Build GRCh38
rs764705130 Chr.12: 2858927-2858927 on Build GRCh38
rs763421291 Chr.12: 2859152-2859152 on Build GRCh38
rs919982920 Chr.12: 2859182-2859182 on Build GRCh38
rs983561029 Chr.12: 2859147-2859147 on Build GRCh38
rs774045890 Chr.12: 2859034-2859034 on Build GRCh38
rs781663757 Chr.12: 2858820-2858820 on Build GRCh38

View Full Product Details