Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2866307-2866814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXM1
Entrez Gene ID
2305
Gene Name
forkhead box M1
Gene Aliases
FKHL16, FOXM1B, HFH-11, HFH11, HNF-3, INS-1, MPHOSPH2, MPP-2, MPP2, PIG29, TRIDENT
UniGene
Hs.239
Gene Chromosome Location
Chr.12: 2857681-2877155 on Build GRCh38
Transcripts
NM_001243088.1 NM_001243089.1 NM_021953.3 NM_202002.2 NM_202003.2 XM_005253676.3 XM_011520930.2 XM_011520931.2 XM_011520932.1 XM_011520933.1 XM_011520934.2 XM_011520935.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6136360 c.965A>T p.Q322L Chr.12: 2866403-2866403 on Build GRCh38
5893663 c.851C>T p.S284F Chr.12: 2866517-2866517 on Build GRCh38
2204959 c.970T>C p.F324L Chr.12: 2866398-2866398 on Build GRCh38
3460049 c.933C>T p.H311H Chr.12: 2866435-2866435 on Build GRCh38
415895 c.954G>C p.L318F Chr.12: 2866414-2866414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142769604 Chr.12: 2866504-2866504 on Build GRCh38
rs1014775673 Chr.12: 2866671-2866671 on Build GRCh38
rs991066082 Chr.12: 2866441-2866441 on Build GRCh38
rs567889714 Chr.12: 2866636-2866636 on Build GRCh38
rs926129456 Chr.12: 2866576-2866576 on Build GRCh38
rs1054313519 Chr.12: 2866519-2866519 on Build GRCh38
rs114917364 Chr.12: 2866725-2866725 on Build GRCh38
rs28990707 Chr.12: 2866624-2866624 on Build GRCh38
rs758088474 Chr.12: 2866600-2866600 on Build GRCh38
rs759078343 Chr.12: 2866471-2866471 on Build GRCh38
rs774157004 Chr.12: 2866434-2866434 on Build GRCh38
rs369008241 Chr.12: 2866472-2866472 on Build GRCh38
rs190384526 Chr.12: 2866676-2866676 on Build GRCh38
rs116218980 Chr.12: 2866516-2866516 on Build GRCh38
rs924103974 Chr.12: 2866759-2866759 on Build GRCh38
rs935541155 Chr.12: 2866776-2866776 on Build GRCh38
rs766279339 Chr.12: 2866426-2866426 on Build GRCh38
rs768686024 Chr.12: 2866373-2866373 on Build GRCh38
rs916805601 Chr.12: 2866702-2866702 on Build GRCh38
rs781264755 Chr.12: 2866421-2866421 on Build GRCh38
rs527842541 Chr.12: 2866611-2866611 on Build GRCh38
rs747044464 Chr.12: 2866366-2866366 on Build GRCh38
rs202093648 Chr.12: 2866572-2866572 on Build GRCh38
rs973176435 Chr.12: 2866680-2866680 on Build GRCh38
rs769684541 Chr.12: 2866414-2866414 on Build GRCh38
rs991573634 Chr.12: 2866694-2866694 on Build GRCh38
rs141851364 Chr.12: 2866666-2866666 on Build GRCh38
rs147930497 Chr.12: 2866608-2866608 on Build GRCh38
rs16930046 Chr.12: 2866477-2866477 on Build GRCh38
rs958383307 Chr.12: 2866686-2866686 on Build GRCh38
rs371728843 Chr.12: 2866396-2866396 on Build GRCh38
rs758695261 Chr.12: 2866488-2866488 on Build GRCh38
rs773579764 Chr.12: 2866544-2866544 on Build GRCh38
rs28990706 Chr.12: 2866685-2866685 on Build GRCh38
rs748271869 Chr.12: 2866512-2866512 on Build GRCh38
rs920268372 Chr.12: 2866395-2866395 on Build GRCh38
rs749042600 Chr.12: 2866554-2866554 on Build GRCh38
rs776979377 Chr.12: 2866385-2866385 on Build GRCh38
rs778495642 Chr.12: 2866622-2866622 on Build GRCh38
rs961267856 Chr.12: 2866762-2866762 on Build GRCh38
rs780607249 Chr.12: 2866489-2866489 on Build GRCh38
rs893804836 Chr.12: 2866533-2866533 on Build GRCh38
rs535184735 Chr.12: 2866637-2866637 on Build GRCh38
rs568725395 Chr.12: 2866675-2866675 on Build GRCh38
rs906222947 Chr.12: 2866668-2866668 on Build GRCh38
rs1015429633 Chr.12: 2866743-2866743 on Build GRCh38
rs922585299 Chr.12: 2866781-2866781 on Build GRCh38
rs750730928 Chr.12: 2866487-2866487 on Build GRCh38
rs763764903 Chr.12: 2866480-2866480 on Build GRCh38
rs746967336 Chr.12: 2866487-2866488 on Build GRCh38
rs770695133 Chr.12: 2866565-2866565 on Build GRCh38
rs866784751 Chr.12: 2866730-2866730 on Build GRCh38
rs768358645 Chr.12: 2866614-2866614 on Build GRCh38
rs199517222 Chr.12: 2866360-2866360 on Build GRCh38
rs1008697987 Chr.12: 2866665-2866665 on Build GRCh38
rs773105134 Chr.12: 2866420-2866420 on Build GRCh38
rs557583484 Chr.12: 2866677-2866677 on Build GRCh38
rs373291558 Chr.12: 2866492-2866492 on Build GRCh38
rs572470946 Chr.12: 2866731-2866731 on Build GRCh38
rs755801968 Chr.12: 2866478-2866478 on Build GRCh38
rs747566493 Chr.12: 2866454-2866454 on Build GRCh38
rs28990705 Chr.12: 2866754-2866754 on Build GRCh38
rs374671659 Chr.12: 2866422-2866422 on Build GRCh38
rs1053340372 Chr.12: 2866620-2866620 on Build GRCh38
rs781464913 Chr.12: 2866511-2866511 on Build GRCh38

View Full Product Details