Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4592917-4593418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK4
Entrez Gene ID
8798
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 4
Gene Aliases
-
UniGene
Hs.439530
Gene Chromosome Location
Chr.12: 4590072-4613888 on Build GRCh38
Transcripts
NM_003845.2 NR_104115.1

SNP Detail

SNP ID Chromosome Location
rs370138021 Chr.12: 4592964-4592964 on Build GRCh38
rs751946437 Chr.12: 4592958-4592958 on Build GRCh38
rs774917137 Chr.12: 4593148-4593148 on Build GRCh38
rs71579238 Chr.12: 4593095-4593095 on Build GRCh38
rs776182813 Chr.12: 4593173-4593173 on Build GRCh38
rs962146615 Chr.12: 4593255-4593255 on Build GRCh38
rs539945708 Chr.12: 4593239-4593239 on Build GRCh38
rs903523440 Chr.12: 4593099-4593099 on Build GRCh38
rs1002099396 Chr.12: 4593149-4593149 on Build GRCh38
rs777689803 Chr.12: 4593060-4593060 on Build GRCh38
rs71579237 Chr.12: 4592989-4592989 on Build GRCh38
rs200597785 Chr.12: 4593022-4593022 on Build GRCh38
rs779643219 Chr.12: 4593341-4593341 on Build GRCh38
rs188558655 Chr.12: 4593209-4593209 on Build GRCh38
rs756169882 Chr.12: 4593214-4593214 on Build GRCh38
rs371271508 Chr.12: 4593151-4593151 on Build GRCh38
rs370590188 Chr.12: 4593010-4593010 on Build GRCh38
rs925056940 Chr.12: 4593322-4593322 on Build GRCh38
rs756336354 Chr.12: 4593109-4593109 on Build GRCh38
rs746129299 Chr.12: 4593055-4593055 on Build GRCh38
rs144925034 Chr.12: 4593395-4593395 on Build GRCh38
rs757254983 Chr.12: 4593240-4593240 on Build GRCh38
rs763341139 Chr.12: 4593180-4593180 on Build GRCh38
rs774187466 Chr.12: 4593065-4593065 on Build GRCh38
rs542189982 Chr.12: 4593274-4593274 on Build GRCh38
rs12306234 Chr.12: 4593198-4593198 on Build GRCh38
rs201436897 Chr.12: 4593023-4593023 on Build GRCh38
rs746539217 Chr.12: 4593160-4593160 on Build GRCh38
rs140089283 Chr.12: 4593108-4593108 on Build GRCh38
rs950077808 Chr.12: 4593313-4593313 on Build GRCh38
rs774530514 Chr.12: 4593002-4593003 on Build GRCh38
rs953251522 Chr.12: 4593250-4593250 on Build GRCh38
rs55825154 Chr.12: 4593045-4593045 on Build GRCh38
rs767760845 Chr.12: 4593107-4593107 on Build GRCh38
rs777871159 Chr.12: 4593110-4593110 on Build GRCh38
rs549254908 Chr.12: 4592946-4592946 on Build GRCh38
rs768309434 Chr.12: 4593170-4593170 on Build GRCh38
rs766865898 Chr.12: 4593103-4593103 on Build GRCh38
rs757042179 Chr.12: 4593134-4593134 on Build GRCh38
rs935077892 Chr.12: 4593364-4593364 on Build GRCh38
rs34403469 Chr.12: 4593068-4593069 on Build GRCh38
rs1052071898 Chr.12: 4593373-4593373 on Build GRCh38
rs752043678 Chr.12: 4593105-4593105 on Build GRCh38
rs775111536 Chr.12: 4593075-4593075 on Build GRCh38
rs763252711 Chr.12: 4593102-4593102 on Build GRCh38
rs752765034 Chr.12: 4593212-4593212 on Build GRCh38
rs750893511 Chr.12: 4593024-4593024 on Build GRCh38
rs2240763 Chr.12: 4593007-4593007 on Build GRCh38
rs567542352 Chr.12: 4593106-4593106 on Build GRCh38
rs761669124 Chr.12: 4593310-4593310 on Build GRCh38
rs879096470 Chr.12: 4593181-4593181 on Build GRCh38
rs193116386 Chr.12: 4593242-4593242 on Build GRCh38
rs1013770747 Chr.12: 4593182-4593182 on Build GRCh38
rs759059511 Chr.12: 4592982-4592982 on Build GRCh38
rs546679771 Chr.12: 4592987-4592987 on Build GRCh38
rs555679721 Chr.12: 4593131-4593131 on Build GRCh38
rs780443245 Chr.12: 4593026-4593026 on Build GRCh38
rs554288890 Chr.12: 4593083-4593083 on Build GRCh38
rs377694754 Chr.12: 4593086-4593086 on Build GRCh38
rs778888694 Chr.12: 4593137-4593137 on Build GRCh38
rs749041571 Chr.12: 4593063-4593063 on Build GRCh38
rs562162234 Chr.12: 4593276-4593276 on Build GRCh38
rs758217868 Chr.12: 4593283-4593283 on Build GRCh38
rs565335118 Chr.12: 4593324-4593324 on Build GRCh38
rs969520766 Chr.12: 4592941-4592941 on Build GRCh38
rs771567729 Chr.12: 4593143-4593143 on Build GRCh38
rs71582876 Chr.12: 4593268-4593268 on Build GRCh38
rs911033284 Chr.12: 4593047-4593047 on Build GRCh38
rs374489268 Chr.12: 4593027-4593027 on Build GRCh38
rs781529973 Chr.12: 4593036-4593036 on Build GRCh38
rs760184652 Chr.12: 4593205-4593205 on Build GRCh38
rs1014566510 Chr.12: 4593223-4593223 on Build GRCh38
rs185292390 Chr.12: 4593201-4593201 on Build GRCh38
rs184727610 Chr.12: 4593281-4593281 on Build GRCh38
rs755136864 Chr.12: 4593030-4593030 on Build GRCh38
rs760272336 Chr.12: 4593025-4593026 on Build GRCh38
rs917567676 Chr.12: 4593382-4593382 on Build GRCh38
rs764346695 Chr.12: 4593215-4593215 on Build GRCh38
rs551663306 Chr.12: 4593287-4593287 on Build GRCh38
rs993537187 Chr.12: 4593265-4593265 on Build GRCh38
rs927340957 Chr.12: 4593354-4593354 on Build GRCh38
rs759016274 Chr.12: 4593066-4593066 on Build GRCh38
rs754058062 Chr.12: 4593217-4593217 on Build GRCh38
rs150048576 Chr.12: 4593138-4593138 on Build GRCh38
rs12306130 Chr.12: 4593064-4593064 on Build GRCh38
rs979214362 Chr.12: 4593317-4593317 on Build GRCh38
rs905329466 Chr.12: 4593231-4593231 on Build GRCh38
rs771620956 Chr.12: 4593069-4593069 on Build GRCh38

View Full Product Details