Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4607237-4607741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK4
Entrez Gene ID
8798
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 4
Gene Aliases
-
UniGene
Hs.439530
Gene Chromosome Location
Chr.12: 4590072-4613888 on Build GRCh38
Transcripts
NM_001282285.1 NM_001282286.1 NM_003845.2

SNP Detail

SNP ID Chromosome Location
rs893250431 Chr.12: 4607604-4607604 on Build GRCh38
rs780410372 Chr.12: 4607282-4607282 on Build GRCh38
rs201010702 Chr.12: 4607353-4607353 on Build GRCh38
rs769045603 Chr.12: 4607304-4607304 on Build GRCh38
rs897296709 Chr.12: 4607495-4607495 on Build GRCh38
rs774138093 Chr.12: 4607418-4607418 on Build GRCh38
rs749595912 Chr.12: 4607402-4607402 on Build GRCh38
rs954081369 Chr.12: 4607288-4607288 on Build GRCh38
rs746582847 Chr.12: 4607517-4607517 on Build GRCh38
rs67382133 Chr.12: 4607501-4607501 on Build GRCh38
rs576622049 Chr.12: 4607430-4607430 on Build GRCh38
rs759452878 Chr.12: 4607343-4607343 on Build GRCh38
rs992611095 Chr.12: 4607370-4607370 on Build GRCh38
rs889814737 Chr.12: 4607549-4607549 on Build GRCh38
rs984448864 Chr.12: 4607649-4607649 on Build GRCh38
rs759273652 Chr.12: 4607422-4607422 on Build GRCh38
rs745351016 Chr.12: 4607691-4607691 on Build GRCh38
rs889849918 Chr.12: 4607540-4607540 on Build GRCh38
rs547124797 Chr.12: 4607399-4607399 on Build GRCh38
rs746186342 Chr.12: 4607278-4607278 on Build GRCh38
rs769960503 Chr.12: 4607330-4607330 on Build GRCh38
rs1006869217 Chr.12: 4607565-4607565 on Build GRCh38
rs1010321610 Chr.12: 4607610-4607610 on Build GRCh38
rs957541688 Chr.12: 4607283-4607283 on Build GRCh38
rs1010439827 Chr.12: 4607291-4607291 on Build GRCh38
rs766256962 Chr.12: 4607340-4607340 on Build GRCh38
rs780642431 Chr.12: 4607686-4607686 on Build GRCh38
rs114607952 Chr.12: 4607400-4607400 on Build GRCh38
rs139849675 Chr.12: 4607345-4607345 on Build GRCh38
rs923091246 Chr.12: 4607500-4607501 on Build GRCh38
rs958411034 Chr.12: 4607645-4607645 on Build GRCh38
rs531454820 Chr.12: 4607681-4607681 on Build GRCh38
rs561251317 Chr.12: 4607522-4607522 on Build GRCh38
rs752737393 Chr.12: 4607373-4607373 on Build GRCh38
rs748245896 Chr.12: 4607326-4607326 on Build GRCh38
rs1022267322 Chr.12: 4607630-4607630 on Build GRCh38
rs145000504 Chr.12: 4607617-4607617 on Build GRCh38
rs373848550 Chr.12: 4607316-4607316 on Build GRCh38
rs961805157 Chr.12: 4607654-4607654 on Build GRCh38
rs562611245 Chr.12: 4607259-4607259 on Build GRCh38
rs368524975 Chr.12: 4607350-4607350 on Build GRCh38
rs760247708 Chr.12: 4607425-4607425 on Build GRCh38
rs757298596 Chr.12: 4607389-4607390 on Build GRCh38
rs933125471 Chr.12: 4607664-4607664 on Build GRCh38
rs1054448336 Chr.12: 4607576-4607576 on Build GRCh38
rs182119678 Chr.12: 4607663-4607663 on Build GRCh38
rs998187452 Chr.12: 4607265-4607265 on Build GRCh38
rs971935974 Chr.12: 4607627-4607627 on Build GRCh38
rs774154805 Chr.12: 4607341-4607341 on Build GRCh38
rs764815013 Chr.12: 4607429-4607429 on Build GRCh38
rs186669253 Chr.12: 4607692-4607692 on Build GRCh38
rs564723354 Chr.12: 4607696-4607696 on Build GRCh38
rs371657903 Chr.12: 4607338-4607338 on Build GRCh38
rs1036194438 Chr.12: 4607480-4607480 on Build GRCh38
rs539344867 Chr.12: 4607432-4607432 on Build GRCh38
rs867539331 Chr.12: 4607637-4607637 on Build GRCh38
rs532208286 Chr.12: 4607660-4607660 on Build GRCh38
rs772096518 Chr.12: 4607423-4607423 on Build GRCh38
rs574895551 Chr.12: 4607539-4607539 on Build GRCh38
rs746906453 Chr.12: 4607439-4607439 on Build GRCh38
rs201862244 Chr.12: 4607337-4607337 on Build GRCh38
rs559194593 Chr.12: 4607481-4607481 on Build GRCh38
rs1020447453 Chr.12: 4607306-4607306 on Build GRCh38
rs112353066 Chr.12: 4607324-4607324 on Build GRCh38
rs200012895 Chr.12: 4607296-4607296 on Build GRCh38
rs781684958 Chr.12: 4607396-4607396 on Build GRCh38
rs541518631 Chr.12: 4607503-4607503 on Build GRCh38
rs758684611 Chr.12: 4607367-4607367 on Build GRCh38
rs1029744074 Chr.12: 4607504-4607504 on Build GRCh38
rs556140744 Chr.12: 4607407-4607407 on Build GRCh38
rs997373 Chr.12: 4607406-4607406 on Build GRCh38
rs369656940 Chr.12: 4607662-4607662 on Build GRCh38
rs761467727 Chr.12: 4607427-4607427 on Build GRCh38
rs760294743 Chr.12: 4607348-4607348 on Build GRCh38
rs758845880 Chr.12: 4607279-4607279 on Build GRCh38
rs2255588 Chr.12: 4607347-4607347 on Build GRCh38
rs778981458 Chr.12: 4607404-4607405 on Build GRCh38
rs1029628178 Chr.12: 4607275-4607275 on Build GRCh38
rs146803881 Chr.12: 4607388-4607388 on Build GRCh38
rs763763291 Chr.12: 4607426-4607426 on Build GRCh38
rs751880189 Chr.12: 4607384-4607384 on Build GRCh38
rs1034991009 Chr.12: 4607634-4607634 on Build GRCh38
rs749213179 Chr.12: 4607357-4607360 on Build GRCh38
rs780398613 Chr.12: 4607380-4607380 on Build GRCh38

View Full Product Details