Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 5046311-5046822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA5
Entrez Gene ID
3741
Gene Name
potassium voltage-gated channel subfamily A member 5
Gene Aliases
ATFB7, HCK1, HK2, HPCN1, KV1.5, PCN1
UniGene
Hs.150208
Gene Chromosome Location
Chr.12: 5043919-5046788 on Build GRCh38
Transcripts
NM_002234.3

SNP Detail

SNP ID Chromosome Location
rs911321770 Chr.12: 5046528-5046528 on Build GRCh38
rs886049577 Chr.12: 5046482-5046482 on Build GRCh38
rs775655212 Chr.12: 5046510-5046510 on Build GRCh38
rs904051089 Chr.12: 5046555-5046555 on Build GRCh38
rs1043815479 Chr.12: 5046547-5046547 on Build GRCh38
rs191708388 Chr.12: 5046534-5046534 on Build GRCh38
rs1022103073 Chr.12: 5046731-5046731 on Build GRCh38
rs917432901 Chr.12: 5046582-5046582 on Build GRCh38
rs66942317 Chr.12: 5046725-5046725 on Build GRCh38
rs1056468 Chr.12: 5046497-5046497 on Build GRCh38
rs183802942 Chr.12: 5046603-5046603 on Build GRCh38
rs971832035 Chr.12: 5046715-5046715 on Build GRCh38
rs961891869 Chr.12: 5046678-5046678 on Build GRCh38
rs934009637 Chr.12: 5046480-5046480 on Build GRCh38
rs1019232814 Chr.12: 5046372-5046372 on Build GRCh38
rs923198418 Chr.12: 5046720-5046720 on Build GRCh38
rs768706206 Chr.12: 5046549-5046549 on Build GRCh38
rs4625555 Chr.12: 5046789-5046789 on Build GRCh38
rs746050844 Chr.12: 5046467-5046467 on Build GRCh38
rs961837873 Chr.12: 5046449-5046449 on Build GRCh38
rs1023696882 Chr.12: 5046662-5046662 on Build GRCh38
rs1043867601 Chr.12: 5046783-5046783 on Build GRCh38
rs971987208 Chr.12: 5046463-5046463 on Build GRCh38
rs756734662 Chr.12: 5046778-5046778 on Build GRCh38
rs1040274443 Chr.12: 5046701-5046701 on Build GRCh38
rs140521061 Chr.12: 5046677-5046677 on Build GRCh38
rs187564684 Chr.12: 5046527-5046527 on Build GRCh38
rs1056466 Chr.12: 5046393-5046393 on Build GRCh38
rs900321599 Chr.12: 5046484-5046484 on Build GRCh38
rs920356222 Chr.12: 5046643-5046643 on Build GRCh38
rs545846214 Chr.12: 5046667-5046667 on Build GRCh38
rs948895540 Chr.12: 5046600-5046600 on Build GRCh38
rs909072134 Chr.12: 5046444-5046444 on Build GRCh38
rs757431595 Chr.12: 5046622-5046622 on Build GRCh38
rs996958727 Chr.12: 5046496-5046496 on Build GRCh38
rs978768131 Chr.12: 5046535-5046535 on Build GRCh38
rs1031640028 Chr.12: 5046552-5046552 on Build GRCh38
rs999686578 Chr.12: 5046601-5046601 on Build GRCh38
rs1013260931 Chr.12: 5046638-5046638 on Build GRCh38
rs3208989 Chr.12: 5046495-5046495 on Build GRCh38
rs993088811 Chr.12: 5046576-5046576 on Build GRCh38
rs930361646 Chr.12: 5046659-5046659 on Build GRCh38
rs548875295 Chr.12: 5046687-5046687 on Build GRCh38
rs189015926 Chr.12: 5046771-5046771 on Build GRCh38
rs986838587 Chr.12: 5046739-5046739 on Build GRCh38
rs71581018 Chr.12: 5046668-5046668 on Build GRCh38
rs144290959 Chr.12: 5046798-5046798 on Build GRCh38
rs531579517 Chr.12: 5046619-5046619 on Build GRCh38
rs80304472 Chr.12: 5046532-5046532 on Build GRCh38
rs896692627 Chr.12: 5046624-5046624 on Build GRCh38

View Full Product Details