Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6730580-6731071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS7A
Entrez Gene ID
50813
Gene Name
COP9 signalosome subunit 7A
Gene Aliases
CSN7, CSN7A, SGN7a
UniGene
Hs.530823
Gene Chromosome Location
Chr.12: 6723984-6731875 on Build GRCh38
Transcripts
NM_001164093.1 NM_001164094.1 NM_001164095.2 NM_016319.3 XM_005253694.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7250804 c.697C>G p.Q233E Chr.12: 6730729-6730729 on Build GRCh38
7250801 c.792C>G p.L264L Chr.12: 6731003-6731003 on Build GRCh38
7201094 c.658A>G p.I220V Chr.12: 6730690-6730690 on Build GRCh38
1363860 c.675A>G p.A225A Chr.12: 6730707-6730707 on Build GRCh38
4044407 c.665T>G p.V222G Chr.12: 6730697-6730697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754363837 Chr.12: 6730979-6730982 on Build GRCh38
rs748981640 Chr.12: 6730768-6730768 on Build GRCh38
rs2286730 Chr.12: 6730747-6730747 on Build GRCh38
rs367598603 Chr.12: 6730977-6730978 on Build GRCh38
rs368132382 Chr.12: 6730988-6730988 on Build GRCh38
rs769264964 Chr.12: 6730792-6730792 on Build GRCh38
rs777276786 Chr.12: 6730752-6730752 on Build GRCh38
rs1010286935 Chr.12: 6730707-6730707 on Build GRCh38
rs778758413 Chr.12: 6730967-6730967 on Build GRCh38
rs747470345 Chr.12: 6730787-6730787 on Build GRCh38
rs1007935764 Chr.12: 6730956-6730956 on Build GRCh38
rs370260459 Chr.12: 6731013-6731013 on Build GRCh38
rs757644452 Chr.12: 6730985-6730988 on Build GRCh38
rs56992815 Chr.12: 6730889-6730889 on Build GRCh38
rs745344099 Chr.12: 6730968-6730968 on Build GRCh38
rs759869466 Chr.12: 6730961-6730962 on Build GRCh38
rs779618375 Chr.12: 6731040-6731043 on Build GRCh38
rs777322286 Chr.12: 6730793-6730793 on Build GRCh38
rs764400753 Chr.12: 6730974-6730974 on Build GRCh38
rs770254306 Chr.12: 6730818-6730818 on Build GRCh38
rs766481814 Chr.12: 6730704-6730704 on Build GRCh38
rs762321124 Chr.12: 6730659-6730659 on Build GRCh38
rs764980021 Chr.12: 6731042-6731042 on Build GRCh38
rs77365023 Chr.12: 6730955-6730955 on Build GRCh38
rs374517867 Chr.12: 6730987-6730987 on Build GRCh38
rs371592739 Chr.12: 6730949-6730949 on Build GRCh38
rs866770862 Chr.12: 6730771-6730771 on Build GRCh38
rs770280665 Chr.12: 6730774-6730774 on Build GRCh38
rs767596556 Chr.12: 6731006-6731006 on Build GRCh38
rs750398835 Chr.12: 6731049-6731049 on Build GRCh38
rs942951678 Chr.12: 6730618-6730618 on Build GRCh38
rs758397165 Chr.12: 6730703-6730703 on Build GRCh38
rs1045775625 Chr.12: 6730978-6730978 on Build GRCh38
rs938724180 Chr.12: 6730881-6730882 on Build GRCh38
rs1039858688 Chr.12: 6730627-6730627 on Build GRCh38
rs750727471 Chr.12: 6730701-6730701 on Build GRCh38
rs373193931 Chr.12: 6730778-6730778 on Build GRCh38
rs759648205 Chr.12: 6730860-6730860 on Build GRCh38
rs148675442 Chr.12: 6730717-6730717 on Build GRCh38
rs898388055 Chr.12: 6730932-6730932 on Build GRCh38
rs757043808 Chr.12: 6731039-6731039 on Build GRCh38
rs140864400 Chr.12: 6730830-6730830 on Build GRCh38
rs766317321 Chr.12: 6730854-6730854 on Build GRCh38
rs549562987 Chr.12: 6730723-6730723 on Build GRCh38
rs34594558 Chr.12: 6730861-6730862 on Build GRCh38
rs770530289 Chr.12: 6730637-6730637 on Build GRCh38
rs1021820849 Chr.12: 6730971-6730978 on Build GRCh38
rs146849329 Chr.12: 6730665-6730665 on Build GRCh38
rs925546788 Chr.12: 6730909-6730909 on Build GRCh38
rs759598959 Chr.12: 6731003-6731003 on Build GRCh38
rs752688777 Chr.12: 6730951-6730951 on Build GRCh38
rs752562238 Chr.12: 6731009-6731009 on Build GRCh38
rs574585424 Chr.12: 6730911-6730911 on Build GRCh38
rs753589620 Chr.12: 6730926-6730926 on Build GRCh38
rs973901065 Chr.12: 6730799-6730799 on Build GRCh38
rs760313769 Chr.12: 6730858-6730858 on Build GRCh38
rs1054593291 Chr.12: 6730699-6730699 on Build GRCh38
rs200637358 Chr.12: 6730980-6730980 on Build GRCh38
rs143075130 Chr.12: 6730881-6730881 on Build GRCh38
rs34310050 Chr.12: 6730838-6730839 on Build GRCh38
rs199950835 Chr.12: 6730716-6730716 on Build GRCh38
rs931247182 Chr.12: 6730939-6730939 on Build GRCh38
rs377622273 Chr.12: 6730957-6730957 on Build GRCh38
rs148232906 Chr.12: 6730982-6730982 on Build GRCh38
rs1034263531 Chr.12: 6730780-6730780 on Build GRCh38
rs151177416 Chr.12: 6731004-6731004 on Build GRCh38
rs1001415163 Chr.12: 6730757-6730757 on Build GRCh38
rs753645445 Chr.12: 6731033-6731033 on Build GRCh38
rs375043464 Chr.12: 6730984-6730984 on Build GRCh38
rs565116686 Chr.12: 6730654-6730654 on Build GRCh38
rs757926052 Chr.12: 6731047-6731047 on Build GRCh38
rs763749330 Chr.12: 6731021-6731021 on Build GRCh38
rs756957079 Chr.12: 6730964-6730964 on Build GRCh38
rs771149812 Chr.12: 6730990-6730990 on Build GRCh38
rs762268025 Chr.12: 6730803-6730803 on Build GRCh38
rs748743191 Chr.12: 6730625-6730625 on Build GRCh38
rs376925970 Chr.12: 6730844-6730844 on Build GRCh38
rs774622741 Chr.12: 6731001-6731001 on Build GRCh38
rs748551302 Chr.12: 6731050-6731050 on Build GRCh38
rs369598803 Chr.12: 6730624-6730624 on Build GRCh38
rs374957064 Chr.12: 6730647-6730647 on Build GRCh38
rs1026137039 Chr.12: 6730800-6730800 on Build GRCh38
rs755804954 Chr.12: 6730954-6730954 on Build GRCh38
rs181174531 Chr.12: 6730888-6730888 on Build GRCh38
rs752457696 Chr.12: 6730746-6730746 on Build GRCh38
rs753109596 Chr.12: 6730961-6730964 on Build GRCh38
rs774230583 Chr.12: 6730698-6730699 on Build GRCh38
rs767622469 Chr.12: 6730960-6730961 on Build GRCh38
rs200625984 Chr.12: 6730642-6730642 on Build GRCh38
rs115573296 Chr.12: 6730972-6730972 on Build GRCh38
rs371056900 Chr.12: 6731045-6731046 on Build GRCh38
rs3168600 Chr.12: 6731043-6731043 on Build GRCh38
rs118080174 Chr.12: 6730891-6730891 on Build GRCh38

View Full Product Details