Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6826564-6827064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR162
Entrez Gene ID
27239
Gene Name
G protein-coupled receptor 162
Gene Aliases
A-2, GRCA
UniGene
Hs.631654
Gene Chromosome Location
Chr.12: 6821797-6827418 on Build GRCh38
Transcripts
NM_014449.1 NM_019858.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
468841 c.1415A>T p.E472V Chr.12: 6826852-6826852 on Build GRCh38
1722007 c.1512G>A p.E504E Chr.12: 6826949-6826949 on Build GRCh38
5631438 c.1453A>T p.S485C Chr.12: 6826890-6826890 on Build GRCh38
7235803 c.1266C>T p.T422T Chr.12: 6826703-6826703 on Build GRCh38
5607425 c.1450G>A p.E484K Chr.12: 6826887-6826887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781976248 Chr.12: 6826854-6826854 on Build GRCh38
rs200776021 Chr.12: 6826801-6826801 on Build GRCh38
rs782428915 Chr.12: 6827025-6827025 on Build GRCh38
rs782557583 Chr.12: 6827031-6827031 on Build GRCh38
rs782471102 Chr.12: 6826629-6826629 on Build GRCh38
rs1039240609 Chr.12: 6826750-6826750 on Build GRCh38
rs373524161 Chr.12: 6826988-6826988 on Build GRCh38
rs782569847 Chr.12: 6826837-6826837 on Build GRCh38
rs782641467 Chr.12: 6826958-6826958 on Build GRCh38
rs200837170 Chr.12: 6826867-6826867 on Build GRCh38
rs371891969 Chr.12: 6826882-6826882 on Build GRCh38
rs781812109 Chr.12: 6826928-6826928 on Build GRCh38
rs375081664 Chr.12: 6827014-6827014 on Build GRCh38
rs782764652 Chr.12: 6826863-6826863 on Build GRCh38
rs782514558 Chr.12: 6827039-6827039 on Build GRCh38
rs140695531 Chr.12: 6826639-6826639 on Build GRCh38
rs367712836 Chr.12: 6826876-6826876 on Build GRCh38
rs376023985 Chr.12: 6826707-6826707 on Build GRCh38
rs750884919 Chr.12: 6826889-6826889 on Build GRCh38
rs145213201 Chr.12: 6826858-6826858 on Build GRCh38
rs2071082 Chr.12: 6826622-6826622 on Build GRCh38
rs782258380 Chr.12: 6826855-6826855 on Build GRCh38
rs374204662 Chr.12: 6826827-6826827 on Build GRCh38
rs781938786 Chr.12: 6826739-6826739 on Build GRCh38
rs368008300 Chr.12: 6826902-6826902 on Build GRCh38
rs144834616 Chr.12: 6826913-6826913 on Build GRCh38
rs782300470 Chr.12: 6826617-6826617 on Build GRCh38
rs782808475 Chr.12: 6826870-6826870 on Build GRCh38
rs377367080 Chr.12: 6826828-6826828 on Build GRCh38
rs944659798 Chr.12: 6826726-6826726 on Build GRCh38
rs782480870 Chr.12: 6826831-6826831 on Build GRCh38
rs782013622 Chr.12: 6826997-6826997 on Build GRCh38
rs782275323 Chr.12: 6827037-6827037 on Build GRCh38
rs782038710 Chr.12: 6827016-6827016 on Build GRCh38
rs781950849 Chr.12: 6826861-6826861 on Build GRCh38
rs113662266 Chr.12: 6826787-6826787 on Build GRCh38
rs58069762 Chr.12: 6826835-6826843 on Build GRCh38
rs782562218 Chr.12: 6826784-6826784 on Build GRCh38
rs368449781 Chr.12: 6826920-6826920 on Build GRCh38
rs144704717 Chr.12: 6826829-6826829 on Build GRCh38
rs781874188 Chr.12: 6826864-6826864 on Build GRCh38
rs782023420 Chr.12: 6826791-6826791 on Build GRCh38
rs782352922 Chr.12: 6826628-6826628 on Build GRCh38
rs781913714 Chr.12: 6826856-6826857 on Build GRCh38
rs149420287 Chr.12: 6826875-6826875 on Build GRCh38
rs370685085 Chr.12: 6826815-6826815 on Build GRCh38
rs782542892 Chr.12: 6826679-6826679 on Build GRCh38
rs145851125 Chr.12: 6826703-6826703 on Build GRCh38
rs781988790 Chr.12: 6826755-6826755 on Build GRCh38
rs148572255 Chr.12: 6826919-6826919 on Build GRCh38
rs138514784 Chr.12: 6826823-6826831 on Build GRCh38
rs142902354 Chr.12: 6826884-6826884 on Build GRCh38
rs782309960 Chr.12: 6826708-6826708 on Build GRCh38
rs891834231 Chr.12: 6826593-6826593 on Build GRCh38
rs782662913 Chr.12: 6826832-6826832 on Build GRCh38
rs140779117 Chr.12: 6827044-6827044 on Build GRCh38
rs200791974 Chr.12: 6827004-6827004 on Build GRCh38
rs781834006 Chr.12: 6826769-6826769 on Build GRCh38
rs782339192 Chr.12: 6826965-6826965 on Build GRCh38
rs148895065 Chr.12: 6826785-6826785 on Build GRCh38
rs782247317 Chr.12: 6826789-6826789 on Build GRCh38
rs782036082 Chr.12: 6826655-6826655 on Build GRCh38
rs202234706 Chr.12: 6826756-6826756 on Build GRCh38
rs782702848 Chr.12: 6826749-6826749 on Build GRCh38
rs781883498 Chr.12: 6826778-6826778 on Build GRCh38
rs782258888 Chr.12: 6826904-6826904 on Build GRCh38
rs781917240 Chr.12: 6826964-6826964 on Build GRCh38
rs782762782 Chr.12: 6826696-6826696 on Build GRCh38
rs782051240 Chr.12: 6826662-6826662 on Build GRCh38
rs781968979 Chr.12: 6826911-6826911 on Build GRCh38
rs141548833 Chr.12: 6826838-6826838 on Build GRCh38
rs368878139 Chr.12: 6826877-6826877 on Build GRCh38
rs782203552 Chr.12: 6826627-6826627 on Build GRCh38
rs200462810 Chr.12: 6826764-6826764 on Build GRCh38
rs782235597 Chr.12: 6826923-6826923 on Build GRCh38
rs782295663 Chr.12: 6826713-6826713 on Build GRCh38
rs552567389 Chr.12: 6826905-6826905 on Build GRCh38
rs782712077 Chr.12: 6826826-6826826 on Build GRCh38
rs782782013 Chr.12: 6827020-6827020 on Build GRCh38
rs782315085 Chr.12: 6826631-6826631 on Build GRCh38
rs782490981 Chr.12: 6826776-6826776 on Build GRCh38
rs782440772 Chr.12: 6826932-6826932 on Build GRCh38
rs782279652 Chr.12: 6826833-6826833 on Build GRCh38
rs782617380 Chr.12: 6826603-6826603 on Build GRCh38
rs376022819 Chr.12: 6826908-6826908 on Build GRCh38
rs921798324 Chr.12: 6826757-6826757 on Build GRCh38
rs782728164 Chr.12: 6826860-6826860 on Build GRCh38
rs782312229 Chr.12: 6826720-6826720 on Build GRCh38
rs782298473 Chr.12: 6826799-6826799 on Build GRCh38
rs201141145 Chr.12: 6826987-6826987 on Build GRCh38
rs782392363 Chr.12: 6826668-6826668 on Build GRCh38
rs782141264 Chr.12: 6826818-6826818 on Build GRCh38
rs782624984 Chr.12: 6827042-6827042 on Build GRCh38
rs1004644248 Chr.12: 6827019-6827019 on Build GRCh38
rs199760751 Chr.12: 6826786-6826786 on Build GRCh38
rs975050150 Chr.12: 6827040-6827040 on Build GRCh38
rs141933631 Chr.12: 6827010-6827010 on Build GRCh38
rs782687439 Chr.12: 6826941-6826941 on Build GRCh38
rs181209093 Chr.12: 6826862-6826862 on Build GRCh38
rs781852174 Chr.12: 6826880-6826880 on Build GRCh38
rs149007947 Chr.12: 6826839-6826839 on Build GRCh38
rs936078338 Chr.12: 6826768-6826768 on Build GRCh38
rs782692117 Chr.12: 6826693-6826693 on Build GRCh38
rs913292743 Chr.12: 6826895-6826895 on Build GRCh38
rs782105211 Chr.12: 6826924-6826924 on Build GRCh38
rs35717080 Chr.12: 6826790-6826790 on Build GRCh38
rs782724815 Chr.12: 6826762-6826762 on Build GRCh38
rs782223891 Chr.12: 6826589-6826589 on Build GRCh38
rs781892948 Chr.12: 6826942-6826942 on Build GRCh38
rs782325207 Chr.12: 6826917-6826917 on Build GRCh38
rs782082516 Chr.12: 6826701-6826701 on Build GRCh38
rs782069608 Chr.12: 6827013-6827013 on Build GRCh38
rs781960887 Chr.12: 6827012-6827012 on Build GRCh38
rs782436180 Chr.12: 6826901-6826901 on Build GRCh38
rs781913769 Chr.12: 6826758-6826758 on Build GRCh38
rs11612427 Chr.12: 6826812-6826812 on Build GRCh38
rs201319224 Chr.12: 6826669-6826669 on Build GRCh38
rs782542925 Chr.12: 6826942-6826943 on Build GRCh38
rs147854198 Chr.12: 6826777-6826777 on Build GRCh38
rs782663904 Chr.12: 6826712-6826712 on Build GRCh38
rs781798047 Chr.12: 6826694-6826694 on Build GRCh38
rs144632808 Chr.12: 6826666-6826666 on Build GRCh38
rs138946653 Chr.12: 6826859-6826859 on Build GRCh38
rs782353327 Chr.12: 6826849-6826849 on Build GRCh38
rs782401586 Chr.12: 6826857-6826857 on Build GRCh38
rs782678241 Chr.12: 6827032-6827032 on Build GRCh38
rs782060990 Chr.12: 6826759-6826759 on Build GRCh38
rs199722367 Chr.12: 6826949-6826949 on Build GRCh38
rs782179188 Chr.12: 6826986-6826986 on Build GRCh38
rs151087736 Chr.12: 6827017-6827017 on Build GRCh38
rs372753671 Chr.12: 6826803-6826803 on Build GRCh38
rs782186100 Chr.12: 6826892-6826892 on Build GRCh38
rs782708126 Chr.12: 6826665-6826665 on Build GRCh38
rs782583140 Chr.12: 6826927-6826927 on Build GRCh38
rs782081889 Chr.12: 6826747-6826747 on Build GRCh38

View Full Product Details