Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12789447-12789993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOLD1
Entrez Gene ID
81575
Gene Name
apolipoprotein L domain containing 1
Gene Aliases
VERGE
UniGene
Hs.23388
Gene Chromosome Location
Chr.12: 12725917-12791466 on Build GRCh38
Transcripts
NM_001130415.1 NM_030817.2

SNP Detail

SNP ID Chromosome Location
rs189070008 Chr.12: 12789491-12789491 on Build GRCh38
rs192218731 Chr.12: 12789735-12789735 on Build GRCh38
rs1016565323 Chr.12: 12789865-12789865 on Build GRCh38
rs963615874 Chr.12: 12789850-12789850 on Build GRCh38
rs751763541 Chr.12: 12789582-12789582 on Build GRCh38
rs1053002266 Chr.12: 12789740-12789740 on Build GRCh38
rs1015035451 Chr.12: 12789838-12789838 on Build GRCh38
rs770596113 Chr.12: 12789753-12789753 on Build GRCh38
rs771836224 Chr.12: 12789827-12789827 on Build GRCh38
rs925774058 Chr.12: 12789957-12789957 on Build GRCh38
rs987440377 Chr.12: 12789917-12789917 on Build GRCh38
rs963521653 Chr.12: 12789872-12789872 on Build GRCh38
rs950920363 Chr.12: 12789563-12789563 on Build GRCh38
rs541175956 Chr.12: 12789653-12789653 on Build GRCh38
rs922113372 Chr.12: 12789904-12789904 on Build GRCh38
rs184541967 Chr.12: 12789736-12789736 on Build GRCh38
rs974227342 Chr.12: 12789639-12789639 on Build GRCh38
rs745518463 Chr.12: 12789806-12789806 on Build GRCh38
rs905916548 Chr.12: 12789770-12789770 on Build GRCh38
rs562154974 Chr.12: 12789946-12789946 on Build GRCh38
rs577749103 Chr.12: 12789731-12789731 on Build GRCh38
rs552522471 Chr.12: 12789910-12789910 on Build GRCh38
rs781425488 Chr.12: 12789690-12789690 on Build GRCh38
rs922167671 Chr.12: 12789906-12789906 on Build GRCh38
rs1052200620 Chr.12: 12789529-12789529 on Build GRCh38
rs983695655 Chr.12: 12789825-12789825 on Build GRCh38
rs1050761022 Chr.12: 12789708-12789708 on Build GRCh38
rs942209541 Chr.12: 12789634-12789634 on Build GRCh38
rs950701907 Chr.12: 12789823-12789823 on Build GRCh38
rs892971885 Chr.12: 12789743-12789743 on Build GRCh38
rs878950993 Chr.12: 12789866-12789866 on Build GRCh38
rs1041548660 Chr.12: 12789612-12789612 on Build GRCh38
rs748847946 Chr.12: 12789752-12789752 on Build GRCh38
rs951113144 Chr.12: 12789805-12789805 on Build GRCh38
rs911912756 Chr.12: 12789924-12789924 on Build GRCh38
rs921052015 Chr.12: 12789692-12789692 on Build GRCh38
rs766631981 Chr.12: 12789539-12789539 on Build GRCh38
rs576832654 Chr.12: 12789615-12789615 on Build GRCh38
rs932038474 Chr.12: 12789528-12789528 on Build GRCh38
rs76714436 Chr.12: 12789702-12789702 on Build GRCh38
rs7301360 Chr.12: 12789599-12789599 on Build GRCh38
rs894386371 Chr.12: 12789762-12789762 on Build GRCh38
rs747118755 Chr.12: 12789879-12789879 on Build GRCh38
rs146148285 Chr.12: 12789766-12789766 on Build GRCh38
rs756739098 Chr.12: 12789721-12789721 on Build GRCh38
rs1023939943 Chr.12: 12789779-12789779 on Build GRCh38
rs7301803 Chr.12: 12789956-12789956 on Build GRCh38

View Full Product Details