Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 16036551-16037080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERA
Entrez Gene ID
51071
Gene Name
deoxyribose-phosphate aldolase
Gene Aliases
CGI-26, DEOC
UniGene
Hs.39429
Gene Chromosome Location
Chr.12: 15911172-16037381 on Build GRCh38
Transcripts
NM_001300779.1 NM_015954.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6553060 c.940G>T p.D314Y Chr.12: 16036729-16036729 on Build GRCh38
7198713 c.949A>G p.M317V Chr.12: 16036738-16036738 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774876170 Chr.12: 16036839-16036839 on Build GRCh38
rs570764959 Chr.12: 16036962-16036962 on Build GRCh38
rs2290979 Chr.12: 16036760-16036760 on Build GRCh38
rs767531414 Chr.12: 16036902-16036902 on Build GRCh38
rs545261253 Chr.12: 16036805-16036805 on Build GRCh38
rs182777797 Chr.12: 16036804-16036804 on Build GRCh38
rs772134034 Chr.12: 16036660-16036662 on Build GRCh38
rs764119755 Chr.12: 16036674-16036674 on Build GRCh38
rs2290978 Chr.12: 16036608-16036608 on Build GRCh38
rs772492264 Chr.12: 16036738-16036738 on Build GRCh38
rs1018607017 Chr.12: 16036950-16036950 on Build GRCh38
rs139343788 Chr.12: 16036706-16036706 on Build GRCh38
rs1020017041 Chr.12: 16036909-16036909 on Build GRCh38
rs781202176 Chr.12: 16036705-16036705 on Build GRCh38
rs763878829 Chr.12: 16036772-16036772 on Build GRCh38
rs549975988 Chr.12: 16036638-16036638 on Build GRCh38
rs759052798 Chr.12: 16036646-16036646 on Build GRCh38
rs769004885 Chr.12: 16036724-16036724 on Build GRCh38
rs759603275 Chr.12: 16036878-16036878 on Build GRCh38
rs751262228 Chr.12: 16036675-16036675 on Build GRCh38
rs776421336 Chr.12: 16036740-16036740 on Build GRCh38
rs986518569 Chr.12: 16036949-16036949 on Build GRCh38
rs551627538 Chr.12: 16036604-16036607 on Build GRCh38
rs757088378 Chr.12: 16036697-16036697 on Build GRCh38
rs780158960 Chr.12: 16036715-16036715 on Build GRCh38
rs767403269 Chr.12: 16036778-16036778 on Build GRCh38
rs1031332951 Chr.12: 16036666-16036666 on Build GRCh38
rs1053227408 Chr.12: 16036906-16036906 on Build GRCh38
rs766528210 Chr.12: 16036871-16036872 on Build GRCh38
rs756174296 Chr.12: 16036709-16036709 on Build GRCh38
rs1011759429 Chr.12: 16036947-16036947 on Build GRCh38
rs750623353 Chr.12: 16036782-16036782 on Build GRCh38
rs901518217 Chr.12: 16036639-16036639 on Build GRCh38
rs537283390 Chr.12: 16036939-16036939 on Build GRCh38
rs905475855 Chr.12: 16036837-16036837 on Build GRCh38
rs758445698 Chr.12: 16036664-16036664 on Build GRCh38
rs748407972 Chr.12: 16036736-16036736 on Build GRCh38
rs530251273 Chr.12: 16036659-16036659 on Build GRCh38
rs943151156 Chr.12: 16036859-16036859 on Build GRCh38
rs745985130 Chr.12: 16036657-16036660 on Build GRCh38
rs774948387 Chr.12: 16036732-16036732 on Build GRCh38
rs563339873 Chr.12: 16036860-16036860 on Build GRCh38
rs892774141 Chr.12: 16036918-16036918 on Build GRCh38
rs1032480722 Chr.12: 16036877-16036877 on Build GRCh38
rs762544203 Chr.12: 16036767-16036767 on Build GRCh38
rs764806248 Chr.12: 16036756-16036756 on Build GRCh38
rs752656657 Chr.12: 16036661-16036661 on Build GRCh38
rs751519138 Chr.12: 16036773-16036773 on Build GRCh38
rs1007502348 Chr.12: 16036592-16036592 on Build GRCh38
rs957372465 Chr.12: 16036685-16036685 on Build GRCh38
rs749763487 Chr.12: 16036718-16036718 on Build GRCh38
rs756996851 Chr.12: 16036777-16036777 on Build GRCh38
rs571182058 Chr.12: 16036780-16036780 on Build GRCh38
rs910391876 Chr.12: 16036814-16036814 on Build GRCh38
rs1013614436 Chr.12: 16036779-16036779 on Build GRCh38
rs113331171 Chr.12: 16036915-16036915 on Build GRCh38
rs199536426 Chr.12: 16036651-16036651 on Build GRCh38
rs759309030 Chr.12: 16036755-16036755 on Build GRCh38
rs776439912 Chr.12: 16036979-16036979 on Build GRCh38

View Full Product Details