Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 25499765-25500266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNTD1
Entrez Gene ID
160492
Gene Name
lamin tail domain containing 1
Gene Aliases
IFLTD1, PAS1C1
UniGene
Hs.44647
Gene Chromosome Location
Chr.12: 25476082-25648604 on Build GRCh38
Transcripts
NM_001145727.2 NM_001145728.2 NM_001145729.1 NM_001256266.1 NM_152590.3 XM_011520575.1 XM_011520576.2 XM_011520585.2 XM_017018892.1 XM_017018893.1

SNP Detail

SNP ID Chromosome Location
rs779438340 Chr.12: 25499820-25499820 on Build GRCh38
rs989615993 Chr.12: 25500141-25500141 on Build GRCh38
rs747617234 Chr.12: 25499869-25499869 on Build GRCh38
rs911442039 Chr.12: 25499891-25499891 on Build GRCh38
rs942706069 Chr.12: 25499917-25499917 on Build GRCh38
rs967150303 Chr.12: 25500077-25500077 on Build GRCh38
rs535637100 Chr.12: 25500164-25500164 on Build GRCh38
rs1007158656 Chr.12: 25500006-25500006 on Build GRCh38
rs534346653 Chr.12: 25499984-25499984 on Build GRCh38
rs969572588 Chr.12: 25500121-25500121 on Build GRCh38
rs374787754 Chr.12: 25499981-25499981 on Build GRCh38
rs188532910 Chr.12: 25499867-25499867 on Build GRCh38
rs562400111 Chr.12: 25499803-25499803 on Build GRCh38
rs568057023 Chr.12: 25500156-25500156 on Build GRCh38
rs1034683291 Chr.12: 25500020-25500020 on Build GRCh38
rs909650011 Chr.12: 25500106-25500106 on Build GRCh38
rs185629495 Chr.12: 25499799-25499799 on Build GRCh38
rs910014090 Chr.12: 25499808-25499808 on Build GRCh38
rs374820588 Chr.12: 25500033-25500033 on Build GRCh38
rs977183603 Chr.12: 25500113-25500113 on Build GRCh38
rs939560988 Chr.12: 25500191-25500191 on Build GRCh38
rs11048081 Chr.12: 25499896-25499896 on Build GRCh38
rs888291360 Chr.12: 25499983-25499983 on Build GRCh38
rs913898090 Chr.12: 25500206-25500206 on Build GRCh38
rs928227575 Chr.12: 25499866-25499866 on Build GRCh38
rs925670882 Chr.12: 25500120-25500120 on Build GRCh38
rs759324216 Chr.12: 25500163-25500163 on Build GRCh38
rs76662229 Chr.12: 25500181-25500181 on Build GRCh38
rs994192979 Chr.12: 25499964-25499964 on Build GRCh38
rs889227549 Chr.12: 25500233-25500233 on Build GRCh38
rs552114813 Chr.12: 25499940-25499940 on Build GRCh38
rs1052645407 Chr.12: 25500204-25500204 on Build GRCh38
rs1014377614 Chr.12: 25500045-25500045 on Build GRCh38
rs770675800 Chr.12: 25500097-25500097 on Build GRCh38
rs1041463886 Chr.12: 25500212-25500212 on Build GRCh38
rs946758577 Chr.12: 25500208-25500208 on Build GRCh38
rs942960355 Chr.12: 25500205-25500205 on Build GRCh38
rs904353420 Chr.12: 25499999-25499999 on Build GRCh38
rs114939066 Chr.12: 25499868-25499868 on Build GRCh38
rs951032760 Chr.12: 25500068-25500068 on Build GRCh38
rs1046907484 Chr.12: 25499980-25499980 on Build GRCh38
rs556882099 Chr.12: 25500165-25500165 on Build GRCh38
rs138505361 Chr.12: 25500139-25500139 on Build GRCh38
rs530527306 Chr.12: 25499873-25499873 on Build GRCh38
rs78952431 Chr.12: 25499862-25499862 on Build GRCh38
rs746724545 Chr.12: 25500059-25500059 on Build GRCh38
rs772835058 Chr.12: 25500012-25500012 on Build GRCh38
rs754085312 Chr.12: 25500192-25500192 on Build GRCh38
rs1043770993 Chr.12: 25500210-25500210 on Build GRCh38
rs983861168 Chr.12: 25500068-25500068 on Build GRCh38
rs776615407 Chr.12: 25500143-25500143 on Build GRCh38
rs901290163 Chr.12: 25499925-25499925 on Build GRCh38
rs1041041801 Chr.12: 25499923-25499923 on Build GRCh38
rs1025272857 Chr.12: 25500053-25500053 on Build GRCh38

View Full Product Details