Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 25526092-25526586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNTD1
Entrez Gene ID
160492
Gene Name
lamin tail domain containing 1
Gene Aliases
IFLTD1, PAS1C1
UniGene
Hs.44647
Gene Chromosome Location
Chr.12: 25476082-25648604 on Build GRCh38
Transcripts
NM_001145727.2 NM_001145728.2 NM_001145729.1 NM_001256266.1 NM_152590.3 XM_011520575.1 XM_011520576.2 XM_011520577.2 XM_011520578.2 XM_011520579.2 XM_011520580.1 XM_011520582.1 XM_011520583.1 XM_011520584.1 XM_011520585.2 XM_011520587.1 XM_011520588.1 XM_017018891.1 XM_017018892.1 XM_017018893.1 XM_017018894.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5913390 c.793G>A p.G265S Chr.12: 25526104-25526104 on Build GRCh38
5036634 c.735A>C p.Q245H Chr.12: 25526099-25526099 on Build GRCh38
938173 c.664C>A p.L222I Chr.12: 25526170-25526170 on Build GRCh38
3459517 c.788C>G p.P263R Chr.12: 25526109-25526109 on Build GRCh38
7295395 c.722A>C p.D241A Chr.12: 25526175-25526175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766030891 Chr.12: 25526145-25526145 on Build GRCh38
rs1006282833 Chr.12: 25526289-25526289 on Build GRCh38
rs372209204 Chr.12: 25526270-25526270 on Build GRCh38
rs780509582 Chr.12: 25526254-25526254 on Build GRCh38
rs567871230 Chr.12: 25526544-25526544 on Build GRCh38
rs774518513 Chr.12: 25526124-25526124 on Build GRCh38
rs140304237 Chr.12: 25526507-25526507 on Build GRCh38
rs765210431 Chr.12: 25526167-25526167 on Build GRCh38
rs886430766 Chr.12: 25526274-25526274 on Build GRCh38
rs923522971 Chr.12: 25526238-25526238 on Build GRCh38
rs751203404 Chr.12: 25526205-25526205 on Build GRCh38
rs970649582 Chr.12: 25526499-25526499 on Build GRCh38
rs776493848 Chr.12: 25526146-25526146 on Build GRCh38
rs768765045 Chr.12: 25526119-25526119 on Build GRCh38
rs533366640 Chr.12: 25526339-25526339 on Build GRCh38
rs1024769321 Chr.12: 25526488-25526488 on Build GRCh38
rs188696389 Chr.12: 25526184-25526184 on Build GRCh38
rs759320315 Chr.12: 25526165-25526165 on Build GRCh38
rs147130460 Chr.12: 25526203-25526203 on Build GRCh38
rs762119076 Chr.12: 25526128-25526128 on Build GRCh38
rs11832810 Chr.12: 25526400-25526400 on Build GRCh38
rs749827766 Chr.12: 25526262-25526262 on Build GRCh38
rs1047736851 Chr.12: 25526273-25526273 on Build GRCh38
rs1039170280 Chr.12: 25526226-25526226 on Build GRCh38
rs1054584146 Chr.12: 25526539-25526539 on Build GRCh38
rs926742927 Chr.12: 25526369-25526369 on Build GRCh38
rs551657748 Chr.12: 25526350-25526350 on Build GRCh38
rs368015363 Chr.12: 25526223-25526223 on Build GRCh38
rs1000318841 Chr.12: 25526215-25526215 on Build GRCh38
rs1019948223 Chr.12: 25526115-25526115 on Build GRCh38
rs1029873948 Chr.12: 25526551-25526551 on Build GRCh38
rs562700905 Chr.12: 25526303-25526303 on Build GRCh38
rs191744286 Chr.12: 25526561-25526561 on Build GRCh38
rs781343664 Chr.12: 25526222-25526222 on Build GRCh38
rs1030700950 Chr.12: 25526513-25526513 on Build GRCh38
rs1479499 Chr.12: 25526436-25526436 on Build GRCh38
rs1006541444 Chr.12: 25526307-25526339 on Build GRCh38
rs374111917 Chr.12: 25526197-25526197 on Build GRCh38
rs371407437 Chr.12: 25526243-25526243 on Build GRCh38
rs371877407 Chr.12: 25526178-25526178 on Build GRCh38
rs756947017 Chr.12: 25526211-25526211 on Build GRCh38
rs914785755 Chr.12: 25526166-25526166 on Build GRCh38
rs201115757 Chr.12: 25526258-25526258 on Build GRCh38
rs962747585 Chr.12: 25526348-25526348 on Build GRCh38
rs767974800 Chr.12: 25526555-25526555 on Build GRCh38
rs997036204 Chr.12: 25526260-25526260 on Build GRCh38
rs997403882 Chr.12: 25526547-25526547 on Build GRCh38
rs767279190 Chr.12: 25526269-25526269 on Build GRCh38
rs950005488 Chr.12: 25526455-25526455 on Build GRCh38
rs974541294 Chr.12: 25526351-25526351 on Build GRCh38
rs567377630 Chr.12: 25526402-25526405 on Build GRCh38

View Full Product Details