Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26398785-26399291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPR2
Entrez Gene ID
3709
Gene Name
inositol 1,4,5-trisphosphate receptor type 2
Gene Aliases
ANHD, CFAP48, INSP3R2, IP3R2
UniGene
Hs.512235
Gene Chromosome Location
Chr.12: 26335337-26833198 on Build GRCh38
Transcripts
NM_002223.3 XM_017019266.1 XM_017019267.1 XM_017019268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
108558 c.7656G>A p.Q2552Q Chr.12: 26398916-26398916 on Build GRCh38
7275699 c.7540T>C p.F2514L Chr.12: 26399032-26399032 on Build GRCh38
1135367 c.7696+2T>C p.? Chr.12: 26398874-26398874 on Build GRCh38
3811720 c.7647C>T p.S2549S Chr.12: 26398925-26398925 on Build GRCh38
3671037 c.7549C>A p.R2517R Chr.12: 26399023-26399023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748602264 Chr.12: 26398854-26398854 on Build GRCh38
rs555893173 Chr.12: 26398830-26398833 on Build GRCh38
rs556256417 Chr.12: 26398821-26398821 on Build GRCh38
rs916533346 Chr.12: 26398876-26398876 on Build GRCh38
rs373724797 Chr.12: 26399026-26399026 on Build GRCh38
rs375931279 Chr.12: 26398877-26398877 on Build GRCh38
rs370237159 Chr.12: 26398973-26398973 on Build GRCh38
rs921719372 Chr.12: 26399149-26399149 on Build GRCh38
rs868832126 Chr.12: 26399251-26399251 on Build GRCh38
rs990921850 Chr.12: 26398846-26398846 on Build GRCh38
rs750647184 Chr.12: 26399101-26399101 on Build GRCh38
rs763790368 Chr.12: 26398898-26398898 on Build GRCh38
rs780932029 Chr.12: 26398836-26398836 on Build GRCh38
rs575839834 Chr.12: 26399058-26399059 on Build GRCh38
rs778443554 Chr.12: 26399068-26399068 on Build GRCh38
rs776728857 Chr.12: 26398869-26398869 on Build GRCh38
rs748599300 Chr.12: 26399056-26399056 on Build GRCh38
rs772767712 Chr.12: 26398859-26398859 on Build GRCh38
rs935597820 Chr.12: 26398985-26398985 on Build GRCh38
rs193145513 Chr.12: 26398949-26398949 on Build GRCh38
rs199972885 Chr.12: 26398844-26398844 on Build GRCh38
rs376530867 Chr.12: 26399038-26399038 on Build GRCh38
rs754729754 Chr.12: 26399033-26399033 on Build GRCh38
rs773390113 Chr.12: 26398861-26398861 on Build GRCh38
rs1036979136 Chr.12: 26399242-26399242 on Build GRCh38
rs767482355 Chr.12: 26398970-26398970 on Build GRCh38
rs1029984978 Chr.12: 26399249-26399249 on Build GRCh38
rs545817072 Chr.12: 26399230-26399230 on Build GRCh38
rs983236999 Chr.12: 26398819-26398819 on Build GRCh38
rs146419029 Chr.12: 26398916-26398916 on Build GRCh38
rs1004209834 Chr.12: 26398823-26398823 on Build GRCh38
rs907781436 Chr.12: 26399228-26399228 on Build GRCh38
rs371747205 Chr.12: 26399059-26399059 on Build GRCh38
rs370914649 Chr.12: 26399069-26399069 on Build GRCh38
rs185661173 Chr.12: 26399257-26399257 on Build GRCh38
rs950661716 Chr.12: 26398818-26398818 on Build GRCh38
rs747728928 Chr.12: 26398998-26398999 on Build GRCh38
rs755644595 Chr.12: 26398993-26398993 on Build GRCh38
rs751427230 Chr.12: 26398925-26398925 on Build GRCh38
rs1053935118 Chr.12: 26399022-26399022 on Build GRCh38
rs749617306 Chr.12: 26398825-26398825 on Build GRCh38
rs779700879 Chr.12: 26399017-26399017 on Build GRCh38
rs974591249 Chr.12: 26399148-26399148 on Build GRCh38
rs936375987 Chr.12: 26399140-26399140 on Build GRCh38
rs776835750 Chr.12: 26399091-26399091 on Build GRCh38
rs760865222 Chr.12: 26398865-26398865 on Build GRCh38
rs771225790 Chr.12: 26398866-26398866 on Build GRCh38
rs375180640 Chr.12: 26399089-26399089 on Build GRCh38
rs372773826 Chr.12: 26398841-26398841 on Build GRCh38
rs997122020 Chr.12: 26399197-26399197 on Build GRCh38
rs753439959 Chr.12: 26399023-26399023 on Build GRCh38
rs933258165 Chr.12: 26399193-26399193 on Build GRCh38

View Full Product Details