Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48916657-48917157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC65
Entrez Gene ID
85478
Gene Name
coiled-coil domain containing 65
Gene Aliases
CFAP250, CILD27, DRC2, FAP250, NYD-SP28
UniGene
Hs.512805
Gene Chromosome Location
Chr.12: 48904110-48921576 on Build GRCh38
Transcripts
NM_001286957.1 NM_033124.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3753281 c.505C>A p.H169N Chr.12: 48917004-48917004 on Build GRCh38
7222213 c.517G>C p.D173H Chr.12: 48917016-48917016 on Build GRCh38
4999115 c.478A>T p.I160F Chr.12: 48916977-48916977 on Build GRCh38
1200027 c.498A>C p.K166N Chr.12: 48916997-48916997 on Build GRCh38
4435373 c.598C>G p.L200V Chr.12: 48917097-48917097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948359562 Chr.12: 48917100-48917100 on Build GRCh38
rs964780090 Chr.12: 48916756-48916756 on Build GRCh38
rs776606625 Chr.12: 48916959-48916959 on Build GRCh38
rs940537637 Chr.12: 48916734-48916734 on Build GRCh38
rs1037994097 Chr.12: 48916875-48916875 on Build GRCh38
rs975341889 Chr.12: 48916758-48916758 on Build GRCh38
rs775477618 Chr.12: 48917069-48917069 on Build GRCh38
rs10875891 Chr.12: 48916941-48916941 on Build GRCh38
rs1036149426 Chr.12: 48916841-48916841 on Build GRCh38
rs140228766 Chr.12: 48917048-48917048 on Build GRCh38
rs764026673 Chr.12: 48916940-48916941 on Build GRCh38
rs923328763 Chr.12: 48917039-48917039 on Build GRCh38
rs370764448 Chr.12: 48917112-48917112 on Build GRCh38
rs200901661 Chr.12: 48916931-48916931 on Build GRCh38
rs747595035 Chr.12: 48917017-48917017 on Build GRCh38
rs753859313 Chr.12: 48916942-48916943 on Build GRCh38
rs879313076 Chr.12: 48916941-48916942 on Build GRCh38
rs572814773 Chr.12: 48916783-48916783 on Build GRCh38
rs867712662 Chr.12: 48916747-48916747 on Build GRCh38
rs367593202 Chr.12: 48917096-48917096 on Build GRCh38
rs574543245 Chr.12: 48916820-48916820 on Build GRCh38
rs907048204 Chr.12: 48916896-48916896 on Build GRCh38
rs770687072 Chr.12: 48916949-48916949 on Build GRCh38
rs908096076 Chr.12: 48916859-48916859 on Build GRCh38
rs760766502 Chr.12: 48917101-48917101 on Build GRCh38
rs939697978 Chr.12: 48916864-48916864 on Build GRCh38
rs148688058 Chr.12: 48916981-48916981 on Build GRCh38
rs940612750 Chr.12: 48917056-48917056 on Build GRCh38
rs202100898 Chr.12: 48917122-48917122 on Build GRCh38
rs374408625 Chr.12: 48916889-48916889 on Build GRCh38
rs894490768 Chr.12: 48916899-48916899 on Build GRCh38
rs369336416 Chr.12: 48917008-48917008 on Build GRCh38
rs776456662 Chr.12: 48917035-48917035 on Build GRCh38
rs145361359 Chr.12: 48917078-48917078 on Build GRCh38
rs191064674 Chr.12: 48916814-48916814 on Build GRCh38
rs942284709 Chr.12: 48917070-48917070 on Build GRCh38
rs767635649 Chr.12: 48917094-48917094 on Build GRCh38
rs370107530 Chr.12: 48917113-48917113 on Build GRCh38
rs750702659 Chr.12: 48916989-48916989 on Build GRCh38
rs974870839 Chr.12: 48917020-48917020 on Build GRCh38
rs544629601 Chr.12: 48916784-48916784 on Build GRCh38
rs760117476 Chr.12: 48916940-48916940 on Build GRCh38
rs774359975 Chr.12: 48916972-48916972 on Build GRCh38
rs754089090 Chr.12: 48917118-48917118 on Build GRCh38
rs1056227879 Chr.12: 48916895-48916895 on Build GRCh38
rs757958583 Chr.12: 48917127-48917127 on Build GRCh38
rs923451487 Chr.12: 48916766-48916766 on Build GRCh38
rs151189508 Chr.12: 48917047-48917047 on Build GRCh38
rs933457677 Chr.12: 48916812-48916812 on Build GRCh38
rs754258267 Chr.12: 48917006-48917006 on Build GRCh38
rs747140463 Chr.12: 48916879-48916879 on Build GRCh38
rs909085884 Chr.12: 48916712-48916712 on Build GRCh38
rs376799979 Chr.12: 48916998-48916998 on Build GRCh38
rs779575242 Chr.12: 48917013-48917013 on Build GRCh38
rs1011968882 Chr.12: 48916900-48916900 on Build GRCh38
rs956041573 Chr.12: 48916727-48916727 on Build GRCh38
rs777553497 Chr.12: 48917032-48917032 on Build GRCh38
rs146444713 Chr.12: 48917092-48917092 on Build GRCh38
rs201665288 Chr.12: 48917120-48917120 on Build GRCh38
rs780069177 Chr.12: 48916709-48916709 on Build GRCh38
rs386762874 Chr.12: 48916941-48916942 on Build GRCh38
rs552756013 Chr.12: 48916782-48916782 on Build GRCh38
rs746553246 Chr.12: 48917131-48917131 on Build GRCh38
rs898288513 Chr.12: 48916880-48916880 on Build GRCh38
rs990108193 Chr.12: 48916738-48916738 on Build GRCh38
rs771792077 Chr.12: 48916935-48916935 on Build GRCh38
rs907338752 Chr.12: 48916846-48916846 on Build GRCh38
rs201278735 Chr.12: 48916941-48916941 on Build GRCh38
rs1031592691 Chr.12: 48916937-48916937 on Build GRCh38
rs1047996314 Chr.12: 48916829-48916829 on Build GRCh38
rs765245229 Chr.12: 48916969-48916969 on Build GRCh38
rs769551409 Chr.12: 48917066-48917066 on Build GRCh38
rs758048490 Chr.12: 48917029-48917029 on Build GRCh38
rs201180905 Chr.12: 48917129-48917129 on Build GRCh38
rs759345369 Chr.12: 48917044-48917044 on Build GRCh38
rs761890305 Chr.12: 48916973-48916973 on Build GRCh38
rs371448136 Chr.12: 48916934-48916934 on Build GRCh38
rs879200198 Chr.12: 48916862-48916863 on Build GRCh38
rs1038023506 Chr.12: 48917083-48917083 on Build GRCh38
rs759355081 Chr.12: 48916961-48916961 on Build GRCh38
rs775056619 Chr.12: 48916939-48916940 on Build GRCh38
rs79716342 Chr.12: 48917004-48917004 on Build GRCh38
rs10875892 Chr.12: 48916942-48916942 on Build GRCh38
rs914665687 Chr.12: 48916748-48916749 on Build GRCh38
rs772000330 Chr.12: 48916916-48916916 on Build GRCh38
rs938750055 Chr.12: 48916892-48916892 on Build GRCh38

View Full Product Details