Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48921011-48921556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC65
Entrez Gene ID
85478
Gene Name
coiled-coil domain containing 65
Gene Aliases
CFAP250, CILD27, DRC2, FAP250, NYD-SP28
UniGene
Hs.512805
Gene Chromosome Location
Chr.12: 48904110-48921576 on Build GRCh38
Transcripts
NM_001286957.1 NM_033124.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6072544 c.1423_1424CA>AT p.H475I Chr.12: 48921411-48921412 on Build GRCh38
6221374 c.1268C>G p.A423G Chr.12: 48921256-48921256 on Build GRCh38
70049 c.1444C>G p.P482A Chr.12: 48921432-48921432 on Build GRCh38
4669990 c.1112T>C p.L371S Chr.12: 48921015-48921015 on Build GRCh38
1361933 c.1415G>A p.R472H Chr.12: 48921403-48921403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923783746 Chr.12: 48921251-48921251 on Build GRCh38
rs780035020 Chr.12: 48921355-48921355 on Build GRCh38
rs116473422 Chr.12: 48921037-48921037 on Build GRCh38
rs760068206 Chr.12: 48921290-48921290 on Build GRCh38
rs117927481 Chr.12: 48921417-48921417 on Build GRCh38
rs754441180 Chr.12: 48921216-48921216 on Build GRCh38
rs767797913 Chr.12: 48921159-48921159 on Build GRCh38
rs752274392 Chr.12: 48921231-48921231 on Build GRCh38
rs746833335 Chr.12: 48921116-48921116 on Build GRCh38
rs143053157 Chr.12: 48921057-48921057 on Build GRCh38
rs754502330 Chr.12: 48921223-48921223 on Build GRCh38
rs745677371 Chr.12: 48921329-48921329 on Build GRCh38
rs774359865 Chr.12: 48921264-48921264 on Build GRCh38
rs760906822 Chr.12: 48921377-48921377 on Build GRCh38
rs757928586 Chr.12: 48921235-48921235 on Build GRCh38
rs368453285 Chr.12: 48921108-48921108 on Build GRCh38
rs117755186 Chr.12: 48921312-48921312 on Build GRCh38
rs201567284 Chr.12: 48921104-48921104 on Build GRCh38
rs951225338 Chr.12: 48921091-48921091 on Build GRCh38
rs763620920 Chr.12: 48921414-48921414 on Build GRCh38
rs958977565 Chr.12: 48921415-48921415 on Build GRCh38
rs770810172 Chr.12: 48921248-48921248 on Build GRCh38
rs765833580 Chr.12: 48921402-48921402 on Build GRCh38
rs1026893169 Chr.12: 48921252-48921252 on Build GRCh38
rs756603557 Chr.12: 48921424-48921426 on Build GRCh38
rs200077078 Chr.12: 48921094-48921094 on Build GRCh38
rs773544356 Chr.12: 48921284-48921284 on Build GRCh38
rs776696901 Chr.12: 48921133-48921133 on Build GRCh38
rs192209412 Chr.12: 48921160-48921160 on Build GRCh38
rs143901776 Chr.12: 48921426-48921426 on Build GRCh38
rs12146813 Chr.12: 48921043-48921043 on Build GRCh38
rs767165177 Chr.12: 48921436-48921436 on Build GRCh38
rs772247089 Chr.12: 48921360-48921360 on Build GRCh38
rs750659386 Chr.12: 48921305-48921305 on Build GRCh38
rs745767342 Chr.12: 48921256-48921256 on Build GRCh38
rs750825575 Chr.12: 48921083-48921083 on Build GRCh38
rs749604328 Chr.12: 48921492-48921493 on Build GRCh38
rs557816572 Chr.12: 48921359-48921359 on Build GRCh38
rs755274383 Chr.12: 48921353-48921353 on Build GRCh38
rs187332866 Chr.12: 48921117-48921117 on Build GRCh38
rs866658813 Chr.12: 48921268-48921268 on Build GRCh38
rs752844049 Chr.12: 48921422-48921423 on Build GRCh38
rs762661201 Chr.12: 48921092-48921092 on Build GRCh38
rs748970407 Chr.12: 48921214-48921214 on Build GRCh38
rs963652208 Chr.12: 48921046-48921046 on Build GRCh38
rs754898426 Chr.12: 48921490-48921490 on Build GRCh38
rs202224195 Chr.12: 48921126-48921126 on Build GRCh38
rs768586223 Chr.12: 48921321-48921321 on Build GRCh38
rs370371605 Chr.12: 48921479-48921479 on Build GRCh38
rs138566110 Chr.12: 48921517-48921518 on Build GRCh38
rs766802543 Chr.12: 48921286-48921286 on Build GRCh38
rs150275571 Chr.12: 48921262-48921262 on Build GRCh38
rs79263537 Chr.12: 48921134-48921134 on Build GRCh38
rs780617540 Chr.12: 48921095-48921095 on Build GRCh38
rs973979644 Chr.12: 48921050-48921050 on Build GRCh38
rs771077761 Chr.12: 48921378-48921378 on Build GRCh38
rs761888390 Chr.12: 48921271-48921271 on Build GRCh38
rs575969790 Chr.12: 48921482-48921482 on Build GRCh38
rs747838048 Chr.12: 48921101-48921101 on Build GRCh38
rs570315013 Chr.12: 48921141-48921141 on Build GRCh38
rs963685141 Chr.12: 48921215-48921215 on Build GRCh38
rs781077704 Chr.12: 48921253-48921253 on Build GRCh38
rs890906368 Chr.12: 48921184-48921184 on Build GRCh38
rs945597736 Chr.12: 48921388-48921388 on Build GRCh38
rs1045103648 Chr.12: 48921110-48921110 on Build GRCh38
rs756877540 Chr.12: 48921422-48921422 on Build GRCh38
rs751749066 Chr.12: 48921247-48921248 on Build GRCh38
rs746775030 Chr.12: 48921247-48921247 on Build GRCh38
rs778270366 Chr.12: 48921467-48921467 on Build GRCh38
rs375713691 Chr.12: 48921100-48921100 on Build GRCh38
rs767555870 Chr.12: 48921357-48921358 on Build GRCh38
rs777483666 Chr.12: 48921239-48921239 on Build GRCh38
rs777075126 Chr.12: 48921391-48921391 on Build GRCh38
rs766579746 Chr.12: 48921142-48921142 on Build GRCh38
rs772183527 Chr.12: 48921274-48921274 on Build GRCh38
rs878928990 Chr.12: 48921518-48921518 on Build GRCh38
rs143009264 Chr.12: 48921451-48921451 on Build GRCh38
rs138929942 Chr.12: 48921285-48921285 on Build GRCh38
rs10875894 Chr.12: 48921079-48921079 on Build GRCh38
rs1008405856 Chr.12: 48921192-48921192 on Build GRCh38
rs981264324 Chr.12: 48921298-48921298 on Build GRCh38
rs990451120 Chr.12: 48921206-48921206 on Build GRCh38
rs4760600 Chr.12: 48921211-48921211 on Build GRCh38
rs759982254 Chr.12: 48921392-48921392 on Build GRCh38
rs762339147 Chr.12: 48921406-48921406 on Build GRCh38
rs765852026 Chr.12: 48921296-48921296 on Build GRCh38
rs777277007 Chr.12: 48921323-48921323 on Build GRCh38
rs201731115 Chr.12: 48921092-48921092 on Build GRCh38
rs773271813 Chr.12: 48921365-48921365 on Build GRCh38
rs535147502 Chr.12: 48921330-48921330 on Build GRCh38
rs767734975 Chr.12: 48921080-48921080 on Build GRCh38
rs765323004 Chr.12: 48921042-48921044 on Build GRCh38
rs374717179 Chr.12: 48921487-48921487 on Build GRCh38
rs773082894 Chr.12: 48921089-48921089 on Build GRCh38
rs574437005 Chr.12: 48921518-48921519 on Build GRCh38
rs372396268 Chr.12: 48921207-48921207 on Build GRCh38
rs756890716 Chr.12: 48921325-48921325 on Build GRCh38
rs977948670 Chr.12: 48921249-48921249 on Build GRCh38
rs888628227 Chr.12: 48921398-48921398 on Build GRCh38
rs141754315 Chr.12: 48921324-48921324 on Build GRCh38
rs752186333 Chr.12: 48921297-48921297 on Build GRCh38
rs73107887 Chr.12: 48921097-48921097 on Build GRCh38
rs757468183 Chr.12: 48921168-48921168 on Build GRCh38
rs780918338 Chr.12: 48921326-48921326 on Build GRCh38
rs778523498 Chr.12: 48921229-48921229 on Build GRCh38
rs80052257 Chr.12: 48921403-48921403 on Build GRCh38
rs774445773 Chr.12: 48921140-48921140 on Build GRCh38
rs372852763 Chr.12: 48921035-48921035 on Build GRCh38
rs892712838 Chr.12: 48921147-48921147 on Build GRCh38
rs761171312 Chr.12: 48921182-48921182 on Build GRCh38

View Full Product Details