Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3815134-3815628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP104
Entrez Gene ID
9731
Gene Name
centrosomal protein 104
Gene Aliases
CFAP256, GlyBP, JBTS25, KIAA0562, ROC22
UniGene
Hs.133089
Gene Chromosome Location
Chr.1: 3812081-3857387 on Build GRCh38
Transcripts
NM_014704.3 XM_005244815.4 XM_011542473.2 XM_011542474.2 XM_011542475.2 XM_011542476.2 XM_011542477.2 XM_017002917.1 XM_017002918.1 XM_017002919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5905381 c.2698G>A p.G900R Chr.1: 3815482-3815482 on Build GRCh38
4007539 c.2709C>T p.A903A Chr.1: 3815471-3815471 on Build GRCh38
908952 c.2762C>T p.T921M Chr.1: 3815418-3815418 on Build GRCh38
3741262 c.2767G>T p.A923S Chr.1: 3815413-3815413 on Build GRCh38
908953 c.2704A>G p.K902E Chr.1: 3815476-3815476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990842947 Chr.1: 3815204-3815204 on Build GRCh38
rs888262256 Chr.1: 3815272-3815272 on Build GRCh38
rs1030992136 Chr.1: 3815178-3815178 on Build GRCh38
rs753093605 Chr.1: 3815182-3815182 on Build GRCh38
rs757805931 Chr.1: 3815498-3815498 on Build GRCh38
rs754145994 Chr.1: 3815496-3815496 on Build GRCh38
rs1006646913 Chr.1: 3815278-3815278 on Build GRCh38
rs900872514 Chr.1: 3815287-3815287 on Build GRCh38
rs200182606 Chr.1: 3815451-3815451 on Build GRCh38
rs184182963 Chr.1: 3815406-3815406 on Build GRCh38
rs775255288 Chr.1: 3815548-3815548 on Build GRCh38
rs767762091 Chr.1: 3815471-3815471 on Build GRCh38
rs770990029 Chr.1: 3815565-3815565 on Build GRCh38
rs746218491 Chr.1: 3815385-3815385 on Build GRCh38
rs981059088 Chr.1: 3815337-3815337 on Build GRCh38
rs766405628 Chr.1: 3815355-3815355 on Build GRCh38
rs4648409 Chr.1: 3815539-3815539 on Build GRCh38
rs769779921 Chr.1: 3815542-3815542 on Build GRCh38
rs371857904 Chr.1: 3815446-3815446 on Build GRCh38
rs756967631 Chr.1: 3815515-3815515 on Build GRCh38
rs764252937 Chr.1: 3815364-3815364 on Build GRCh38
rs754342571 Chr.1: 3815312-3815312 on Build GRCh38
rs936769479 Chr.1: 3815203-3815203 on Build GRCh38
rs916658169 Chr.1: 3815390-3815390 on Build GRCh38
rs777590113 Chr.1: 3815503-3815503 on Build GRCh38
rs200173861 Chr.1: 3815361-3815361 on Build GRCh38
rs12729542 Chr.1: 3815395-3815395 on Build GRCh38
rs760951895 Chr.1: 3815561-3815561 on Build GRCh38
rs778322536 Chr.1: 3815290-3815290 on Build GRCh38
rs759178611 Chr.1: 3815196-3815196 on Build GRCh38
rs978337067 Chr.1: 3815328-3815328 on Build GRCh38
rs767581799 Chr.1: 3815408-3815408 on Build GRCh38
rs779443196 Chr.1: 3815362-3815362 on Build GRCh38
rs759751517 Chr.1: 3815566-3815566 on Build GRCh38
rs929896052 Chr.1: 3815400-3815400 on Build GRCh38
rs781191434 Chr.1: 3815518-3815518 on Build GRCh38
rs776537838 Chr.1: 3815447-3815447 on Build GRCh38
rs904992516 Chr.1: 3815241-3815241 on Build GRCh38
rs772191034 Chr.1: 3815553-3815553 on Build GRCh38
rs372708011 Chr.1: 3815497-3815497 on Build GRCh38
rs1018002551 Chr.1: 3815280-3815280 on Build GRCh38
rs1053604661 Chr.1: 3815374-3815374 on Build GRCh38
rs1046332909 Chr.1: 3815256-3815256 on Build GRCh38
rs538187962 Chr.1: 3815429-3815429 on Build GRCh38
rs199654344 Chr.1: 3815413-3815413 on Build GRCh38
rs773195110 Chr.1: 3815396-3815396 on Build GRCh38
rs992946703 Chr.1: 3815323-3815323 on Build GRCh38
rs746706047 Chr.1: 3815506-3815506 on Build GRCh38
rs949305915 Chr.1: 3815230-3815230 on Build GRCh38
rs771769043 Chr.1: 3815431-3815431 on Build GRCh38
rs748830711 Chr.1: 3815389-3815389 on Build GRCh38
rs1032564266 Chr.1: 3815580-3815580 on Build GRCh38
rs925416967 Chr.1: 3815184-3815184 on Build GRCh38
rs963525521 Chr.1: 3815180-3815180 on Build GRCh38
rs12728401 Chr.1: 3815401-3815401 on Build GRCh38
rs754388554 Chr.1: 3815418-3815418 on Build GRCh38
rs770996627 Chr.1: 3815399-3815399 on Build GRCh38
rs773241837 Chr.1: 3815557-3815557 on Build GRCh38
rs148360595 Chr.1: 3815444-3815444 on Build GRCh38
rs367595724 Chr.1: 3815375-3815375 on Build GRCh38
rs929850398 Chr.1: 3815262-3815262 on Build GRCh38
rs995023146 Chr.1: 3815609-3815609 on Build GRCh38
rs757390694 Chr.1: 3815371-3815371 on Build GRCh38
rs1045004741 Chr.1: 3815233-3815233 on Build GRCh38
rs4233019 Chr.1: 3815608-3815608 on Build GRCh38
rs751858907 Chr.1: 3815369-3815369 on Build GRCh38
rs892213543 Chr.1: 3815254-3815255 on Build GRCh38
rs750598679 Chr.1: 3815474-3815474 on Build GRCh38
rs748284478 Chr.1: 3815552-3815552 on Build GRCh38
rs958235304 Chr.1: 3815584-3815584 on Build GRCh38
rs371225799 Chr.1: 3815417-3815417 on Build GRCh38
rs1053660015 Chr.1: 3815247-3815247 on Build GRCh38
rs1030605065 Chr.1: 3815317-3815317 on Build GRCh38
rs376014386 Chr.1: 3815392-3815392 on Build GRCh38
rs761736219 Chr.1: 3815470-3815470 on Build GRCh38
rs142026108 Chr.1: 3815234-3815234 on Build GRCh38
rs917282922 Chr.1: 3815222-3815222 on Build GRCh38
rs766767006 Chr.1: 3815495-3815495 on Build GRCh38
rs978646060 Chr.1: 3815183-3815183 on Build GRCh38
rs890146163 Chr.1: 3815263-3815263 on Build GRCh38
rs916584744 Chr.1: 3815207-3815207 on Build GRCh38
rs1048690860 Chr.1: 3815271-3815271 on Build GRCh38
rs374365915 Chr.1: 3815398-3815398 on Build GRCh38
rs989416332 Chr.1: 3815490-3815490 on Build GRCh38
rs774502989 Chr.1: 3815466-3815466 on Build GRCh38
rs967349015 Chr.1: 3815327-3815327 on Build GRCh38
rs768166299 Chr.1: 3815391-3815391 on Build GRCh38
rs377336810 Chr.1: 3815407-3815407 on Build GRCh38
rs780476689 Chr.1: 3815388-3815388 on Build GRCh38
rs377546546 Chr.1: 3815563-3815563 on Build GRCh38
rs770798606 Chr.1: 3815445-3815445 on Build GRCh38
rs530172111 Chr.1: 3815239-3815239 on Build GRCh38
rs997878393 Chr.1: 3815491-3815491 on Build GRCh38
rs755406055 Chr.1: 3815425-3815425 on Build GRCh38
rs750794560 Chr.1: 3815414-3815414 on Build GRCh38
rs936493949 Chr.1: 3815242-3815242 on Build GRCh38
rs369737290 Chr.1: 3815477-3815477 on Build GRCh38
rs943145678 Chr.1: 3815551-3815551 on Build GRCh38
rs371734853 Chr.1: 3815386-3815386 on Build GRCh38
rs867619101 Chr.1: 3815281-3815281 on Build GRCh38
rs201733773 Chr.1: 3815443-3815443 on Build GRCh38
rs897062531 Chr.1: 3815260-3815260 on Build GRCh38
rs200742076 Chr.1: 3815450-3815450 on Build GRCh38

View Full Product Details