Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3829112-3829655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP104
Entrez Gene ID
9731
Gene Name
centrosomal protein 104
Gene Aliases
CFAP256, GlyBP, JBTS25, KIAA0562, ROC22
UniGene
Hs.133089
Gene Chromosome Location
Chr.1: 3812081-3857387 on Build GRCh38
Transcripts
NM_014704.3 XM_005244815.4 XM_011542473.2 XM_011542474.2 XM_011542475.2 XM_011542476.2 XM_011542477.2 XM_011542478.1 XM_017002917.1 XM_017002918.1 XM_017002919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
459249 c.2131G>C p.E711Q Chr.1: 3829286-3829286 on Build GRCh38
6406488 c.2111G>T p.G704V Chr.1: 3829306-3829306 on Build GRCh38
7132817 c.2089G>C p.E697Q Chr.1: 3829328-3829328 on Build GRCh38
6352623 c.2057C>T p.A686V Chr.1: 3829360-3829360 on Build GRCh38
1342368 c.2119G>A p.A707T Chr.1: 3829298-3829298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557247562 Chr.1: 3829365-3829365 on Build GRCh38
rs779930589 Chr.1: 3829321-3829321 on Build GRCh38
rs149317299 Chr.1: 3829537-3829537 on Build GRCh38
rs763765041 Chr.1: 3829377-3829377 on Build GRCh38
rs753810711 Chr.1: 3829410-3829410 on Build GRCh38
rs530874961 Chr.1: 3829152-3829152 on Build GRCh38
rs748254073 Chr.1: 3829336-3829336 on Build GRCh38
rs777854593 Chr.1: 3829508-3829508 on Build GRCh38
rs922659497 Chr.1: 3829456-3829456 on Build GRCh38
rs956218390 Chr.1: 3829420-3829420 on Build GRCh38
rs773370802 Chr.1: 3829251-3829251 on Build GRCh38
rs2275831 Chr.1: 3829360-3829360 on Build GRCh38
rs561062414 Chr.1: 3829630-3829630 on Build GRCh38
rs772184811 Chr.1: 3829338-3829338 on Build GRCh38
rs780909069 Chr.1: 3829314-3829314 on Build GRCh38
rs139817583 Chr.1: 3829293-3829293 on Build GRCh38
rs773944219 Chr.1: 3829235-3829236 on Build GRCh38
rs756686128 Chr.1: 3829530-3829531 on Build GRCh38
rs777771164 Chr.1: 3829414-3829414 on Build GRCh38
rs760138419 Chr.1: 3829399-3829399 on Build GRCh38
rs143442406 Chr.1: 3829287-3829288 on Build GRCh38
rs2275832 Chr.1: 3829223-3829223 on Build GRCh38
rs765755329 Chr.1: 3829412-3829412 on Build GRCh38
rs563680254 Chr.1: 3829571-3829571 on Build GRCh38
rs763522630 Chr.1: 3829373-3829373 on Build GRCh38
rs754902038 Chr.1: 3829413-3829413 on Build GRCh38
rs751210989 Chr.1: 3829308-3829308 on Build GRCh38
rs776090603 Chr.1: 3829370-3829370 on Build GRCh38
rs752259859 Chr.1: 3829305-3829305 on Build GRCh38
rs1041125696 Chr.1: 3829163-3829163 on Build GRCh38
rs771237087 Chr.1: 3829263-3829263 on Build GRCh38
rs543889769 Chr.1: 3829516-3829516 on Build GRCh38
rs572650414 Chr.1: 3829517-3829517 on Build GRCh38
rs11586111 Chr.1: 3829155-3829155 on Build GRCh38
rs994482620 Chr.1: 3829168-3829168 on Build GRCh38
rs937349224 Chr.1: 3829565-3829565 on Build GRCh38
rs76442708 Chr.1: 3829497-3829497 on Build GRCh38
rs749023447 Chr.1: 3829555-3829556 on Build GRCh38
rs1056906111 Chr.1: 3829601-3829601 on Build GRCh38
rs35089650 Chr.1: 3829286-3829287 on Build GRCh38
rs965568323 Chr.1: 3829271-3829271 on Build GRCh38
rs965976157 Chr.1: 3829522-3829522 on Build GRCh38
rs375242891 Chr.1: 3829396-3829396 on Build GRCh38
rs1138630 Chr.1: 3829438-3829438 on Build GRCh38
rs544509506 Chr.1: 3829620-3829620 on Build GRCh38
rs1138631 Chr.1: 3829437-3829437 on Build GRCh38
rs752848486 Chr.1: 3829624-3829624 on Build GRCh38
rs1007590179 Chr.1: 3829504-3829504 on Build GRCh38
rs750177184 Chr.1: 3829316-3829316 on Build GRCh38
rs2275833 Chr.1: 3829157-3829157 on Build GRCh38
rs575419906 Chr.1: 3829564-3829564 on Build GRCh38
rs760924936 Chr.1: 3829252-3829252 on Build GRCh38
rs772747609 Chr.1: 3829377-3829378 on Build GRCh38
rs917193863 Chr.1: 3829632-3829632 on Build GRCh38
rs932738365 Chr.1: 3829493-3829495 on Build GRCh38
rs771531523 Chr.1: 3829333-3829336 on Build GRCh38
rs937279172 Chr.1: 3829346-3829346 on Build GRCh38
rs991864138 Chr.1: 3829580-3829580 on Build GRCh38
rs571804006 Chr.1: 3829320-3829320 on Build GRCh38
rs186117937 Chr.1: 3829171-3829171 on Build GRCh38
rs756932409 Chr.1: 3829311-3829311 on Build GRCh38
rs555483380 Chr.1: 3829538-3829538 on Build GRCh38
rs779798001 Chr.1: 3829409-3829409 on Build GRCh38
rs745861832 Chr.1: 3829221-3829221 on Build GRCh38
rs761514784 Chr.1: 3829326-3829329 on Build GRCh38
rs370817774 Chr.1: 3829224-3829224 on Build GRCh38
rs140499832 Chr.1: 3829369-3829369 on Build GRCh38
rs755857555 Chr.1: 3829404-3829404 on Build GRCh38
rs932622397 Chr.1: 3829182-3829182 on Build GRCh38
rs1027300504 Chr.1: 3829443-3829443 on Build GRCh38
rs765780308 Chr.1: 3829295-3829295 on Build GRCh38
rs747259195 Chr.1: 3829348-3829348 on Build GRCh38
rs771168215 Chr.1: 3829359-3829359 on Build GRCh38
rs772315672 Chr.1: 3829250-3829250 on Build GRCh38
rs777795350 Chr.1: 3829340-3829340 on Build GRCh38
rs139181137 Chr.1: 3829306-3829306 on Build GRCh38
rs1055712248 Chr.1: 3829356-3829356 on Build GRCh38
rs142507755 Chr.1: 3829289-3829289 on Build GRCh38
rs1836 Chr.1: 3829552-3829552 on Build GRCh38
rs751069180 Chr.1: 3829384-3829384 on Build GRCh38
rs897593384 Chr.1: 3829397-3829397 on Build GRCh38
rs35169848 Chr.1: 3829361-3829362 on Build GRCh38
rs895535502 Chr.1: 3829616-3829616 on Build GRCh38
rs1012622987 Chr.1: 3829631-3829631 on Build GRCh38
rs976963611 Chr.1: 3829536-3829536 on Build GRCh38
rs761386725 Chr.1: 3829389-3829389 on Build GRCh38

View Full Product Details