Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49868605-49869116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAIM2
Entrez Gene ID
23017
Gene Name
Fas apoptotic inhibitory molecule 2
Gene Aliases
LFG, LFG2, NGP35, NMP35, TMBIM2
UniGene
Hs.567424
Gene Chromosome Location
Chr.12: 49866896-49904275 on Build GRCh38
Transcripts
NM_012306.3 XM_005268730.3 XM_017019040.1

SNP Detail

SNP ID Chromosome Location
rs936077661 Chr.12: 49869009-49869009 on Build GRCh38
rs374219929 Chr.12: 49868699-49868699 on Build GRCh38
rs890073604 Chr.12: 49868749-49868749 on Build GRCh38
rs1017034394 Chr.12: 49868809-49868809 on Build GRCh38
rs902146340 Chr.12: 49868945-49868945 on Build GRCh38
rs902193943 Chr.12: 49868690-49868690 on Build GRCh38
rs891978895 Chr.12: 49868724-49868724 on Build GRCh38
rs992003734 Chr.12: 49868869-49868869 on Build GRCh38
rs554002763 Chr.12: 49869081-49869081 on Build GRCh38
rs925961540 Chr.12: 49868995-49868995 on Build GRCh38
rs186740048 Chr.12: 49868661-49868661 on Build GRCh38
rs777751458 Chr.12: 49868909-49868909 on Build GRCh38
rs867367215 Chr.12: 49868652-49868652 on Build GRCh38
rs1025678779 Chr.12: 49869059-49869059 on Build GRCh38
rs972853494 Chr.12: 49868774-49868774 on Build GRCh38
rs948715528 Chr.12: 49868670-49868670 on Build GRCh38
rs916496072 Chr.12: 49868931-49868931 on Build GRCh38
rs914203456 Chr.12: 49868650-49868650 on Build GRCh38
rs1006556823 Chr.12: 49868808-49868808 on Build GRCh38
rs923529422 Chr.12: 49868843-49868843 on Build GRCh38
rs974996805 Chr.12: 49868840-49868840 on Build GRCh38
rs937955266 Chr.12: 49868841-49868841 on Build GRCh38
rs998401045 Chr.12: 49868734-49868734 on Build GRCh38
rs879767818 Chr.12: 49868825-49868825 on Build GRCh38
rs868357140 Chr.12: 49868697-49868697 on Build GRCh38
rs778388900 Chr.12: 49868632-49868632 on Build GRCh38
rs1042422010 Chr.12: 49868668-49868668 on Build GRCh38
rs1026039901 Chr.12: 49868787-49868787 on Build GRCh38
rs934142151 Chr.12: 49869036-49869036 on Build GRCh38
rs752449603 Chr.12: 49868820-49868820 on Build GRCh38
rs533846057 Chr.12: 49869043-49869043 on Build GRCh38
rs1029930633 Chr.12: 49868741-49868741 on Build GRCh38
rs539598192 Chr.12: 49868998-49868999 on Build GRCh38
rs751094645 Chr.12: 49869039-49869039 on Build GRCh38
rs142094313 Chr.12: 49868846-49868846 on Build GRCh38
rs952839607 Chr.12: 49869089-49869089 on Build GRCh38
rs1007057343 Chr.12: 49868733-49868733 on Build GRCh38
rs913371411 Chr.12: 49869014-49869014 on Build GRCh38
rs1013360003 Chr.12: 49868667-49868667 on Build GRCh38
rs374964416 Chr.12: 49869038-49869038 on Build GRCh38
rs980115195 Chr.12: 49868979-49868979 on Build GRCh38
rs994334652 Chr.12: 49868836-49868836 on Build GRCh38
rs909132278 Chr.12: 49868835-49868835 on Build GRCh38
rs949133280 Chr.12: 49868925-49868925 on Build GRCh38
rs960530396 Chr.12: 49868847-49868847 on Build GRCh38
rs371477855 Chr.12: 49868673-49868673 on Build GRCh38
rs969301257 Chr.12: 49868946-49868946 on Build GRCh38
rs765041481 Chr.12: 49868815-49868815 on Build GRCh38
rs952725446 Chr.12: 49868791-49868791 on Build GRCh38
rs954317363 Chr.12: 49868715-49868715 on Build GRCh38
rs1032598201 Chr.12: 49868709-49868709 on Build GRCh38
rs962477261 Chr.12: 49868812-49868812 on Build GRCh38
rs936114242 Chr.12: 49868675-49868675 on Build GRCh38
rs145193912 Chr.12: 49868657-49868658 on Build GRCh38
rs1001084194 Chr.12: 49868693-49868693 on Build GRCh38
rs904589350 Chr.12: 49868674-49868674 on Build GRCh38
rs189471300 Chr.12: 49868933-49868933 on Build GRCh38
rs1053616502 Chr.12: 49869013-49869013 on Build GRCh38
rs1007169621 Chr.12: 49869014-49869015 on Build GRCh38

View Full Product Details