Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49887300-49887804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAIM2
Entrez Gene ID
23017
Gene Name
Fas apoptotic inhibitory molecule 2
Gene Aliases
LFG, LFG2, NGP35, NMP35, TMBIM2
UniGene
Hs.567424
Gene Chromosome Location
Chr.12: 49866896-49904275 on Build GRCh38
Transcripts
NM_012306.3 XM_005268730.3 XM_017019040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5375326 c.781G>A p.G261R Chr.12: 49887406-49887406 on Build GRCh38
4911725 c.801+2T>A p.? Chr.12: 49887384-49887384 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1041488259 Chr.12: 49887623-49887623 on Build GRCh38
rs1878234 Chr.12: 49887631-49887631 on Build GRCh38
rs371175967 Chr.12: 49887401-49887401 on Build GRCh38
rs753255026 Chr.12: 49887348-49887348 on Build GRCh38
rs768053978 Chr.12: 49887729-49887729 on Build GRCh38
rs916293655 Chr.12: 49887499-49887499 on Build GRCh38
rs981800628 Chr.12: 49887677-49887677 on Build GRCh38
rs1053105562 Chr.12: 49887561-49887561 on Build GRCh38
rs562250845 Chr.12: 49887728-49887728 on Build GRCh38
rs901276265 Chr.12: 49887624-49887624 on Build GRCh38
rs761491728 Chr.12: 49887395-49887395 on Build GRCh38
rs745376313 Chr.12: 49887444-49887444 on Build GRCh38
rs961296635 Chr.12: 49887667-49887667 on Build GRCh38
rs963514389 Chr.12: 49887467-49887469 on Build GRCh38
rs541844300 Chr.12: 49887676-49887676 on Build GRCh38
rs775097226 Chr.12: 49887504-49887504 on Build GRCh38
rs867651761 Chr.12: 49887709-49887709 on Build GRCh38
rs1006213484 Chr.12: 49887660-49887660 on Build GRCh38
rs1042330784 Chr.12: 49887533-49887533 on Build GRCh38
rs868226691 Chr.12: 49887518-49887518 on Build GRCh38
rs777206135 Chr.12: 49887344-49887344 on Build GRCh38
rs111630200 Chr.12: 49887383-49887383 on Build GRCh38
rs763331101 Chr.12: 49887380-49887380 on Build GRCh38
rs990556844 Chr.12: 49887717-49887717 on Build GRCh38
rs899920094 Chr.12: 49887337-49887337 on Build GRCh38
rs776022293 Chr.12: 49887376-49887376 on Build GRCh38
rs965502765 Chr.12: 49887387-49887387 on Build GRCh38
rs535180127 Chr.12: 49887332-49887332 on Build GRCh38
rs747084335 Chr.12: 49887439-49887439 on Build GRCh38
rs771342070 Chr.12: 49887443-49887443 on Build GRCh38
rs557523076 Chr.12: 49887460-49887460 on Build GRCh38
rs764472564 Chr.12: 49887564-49887564 on Build GRCh38
rs776070850 Chr.12: 49887377-49887377 on Build GRCh38
rs927723013 Chr.12: 49887451-49887451 on Build GRCh38
rs767151158 Chr.12: 49887397-49887397 on Build GRCh38
rs113953525 Chr.12: 49887385-49887385 on Build GRCh38
rs184762843 Chr.12: 49887362-49887362 on Build GRCh38
rs984420178 Chr.12: 49887418-49887418 on Build GRCh38
rs751044469 Chr.12: 49887390-49887390 on Build GRCh38
rs778042998 Chr.12: 49887425-49887425 on Build GRCh38
rs769474078 Chr.12: 49887500-49887500 on Build GRCh38
rs746012289 Chr.12: 49887459-49887459 on Build GRCh38
rs770227686 Chr.12: 49887469-49887469 on Build GRCh38
rs534936700 Chr.12: 49887416-49887416 on Build GRCh38
rs376005780 Chr.12: 49887335-49887335 on Build GRCh38
rs137990093 Chr.12: 49887525-49887525 on Build GRCh38
rs537284282 Chr.12: 49887751-49887751 on Build GRCh38
rs541807847 Chr.12: 49887420-49887420 on Build GRCh38
rs138641504 Chr.12: 49887407-49887407 on Build GRCh38
rs576416197 Chr.12: 49887634-49887634 on Build GRCh38
rs775670847 Chr.12: 49887470-49887470 on Build GRCh38
rs999343669 Chr.12: 49887365-49887365 on Build GRCh38
rs753842063 Chr.12: 49887410-49887410 on Build GRCh38
rs959127701 Chr.12: 49887694-49887694 on Build GRCh38
rs577517095 Chr.12: 49887600-49887600 on Build GRCh38
rs927631291 Chr.12: 49887531-49887531 on Build GRCh38
rs750536882 Chr.12: 49887778-49887778 on Build GRCh38
rs759970003 Chr.12: 49887355-49887355 on Build GRCh38
rs978381434 Chr.12: 49887768-49887768 on Build GRCh38
rs1034708150 Chr.12: 49887692-49887692 on Build GRCh38
rs556686473 Chr.12: 49887527-49887527 on Build GRCh38
rs923882973 Chr.12: 49887777-49887777 on Build GRCh38
rs946622244 Chr.12: 49887532-49887532 on Build GRCh38
rs536918202 Chr.12: 49887615-49887615 on Build GRCh38
rs937804759 Chr.12: 49887496-49887496 on Build GRCh38
rs558707463 Chr.12: 49887770-49887770 on Build GRCh38
rs982259492 Chr.12: 49887450-49887450 on Build GRCh38
rs996991372 Chr.12: 49887626-49887626 on Build GRCh38
rs903175893 Chr.12: 49887608-49887608 on Build GRCh38

View Full Product Details