Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50118355-50118898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX14
Entrez Gene ID
84987
Gene Name
COX14, cytochrome c oxidase assembly factor
Gene Aliases
C12orf62
UniGene
Hs.388645
Gene Chromosome Location
Chr.12: 50111981-50120457 on Build GRCh38
Transcripts
NM_001257133.1 NM_001257134.1 NM_032901.3

SNP Detail

SNP ID Chromosome Location
rs12815315 Chr.12: 50118595-50118595 on Build GRCh38
rs1019610667 Chr.12: 50118728-50118728 on Build GRCh38
rs752846519 Chr.12: 50118563-50118564 on Build GRCh38
rs55639096 Chr.12: 50118496-50118496 on Build GRCh38
rs893853511 Chr.12: 50118682-50118682 on Build GRCh38
rs56151073 Chr.12: 50118417-50118417 on Build GRCh38
rs12815274 Chr.12: 50118534-50118534 on Build GRCh38
rs1009135584 Chr.12: 50118726-50118726 on Build GRCh38
rs558708814 Chr.12: 50118732-50118732 on Build GRCh38
rs7307230 Chr.12: 50118849-50118849 on Build GRCh38
rs181740058 Chr.12: 50118414-50118414 on Build GRCh38
rs12229758 Chr.12: 50118510-50118510 on Build GRCh38
rs560088824 Chr.12: 50118684-50118684 on Build GRCh38
rs71441321 Chr.12: 50118773-50118773 on Build GRCh38
rs531806804 Chr.12: 50118761-50118761 on Build GRCh38
rs987664099 Chr.12: 50118478-50118478 on Build GRCh38
rs372981475 Chr.12: 50118557-50118557 on Build GRCh38
rs1036509471 Chr.12: 50118446-50118446 on Build GRCh38
rs951754367 Chr.12: 50118762-50118762 on Build GRCh38
rs921843324 Chr.12: 50118406-50118406 on Build GRCh38
rs1055223335 Chr.12: 50118677-50118677 on Build GRCh38
rs931813467 Chr.12: 50118411-50118411 on Build GRCh38
rs1022330899 Chr.12: 50118535-50118535 on Build GRCh38
rs1021174937 Chr.12: 50118426-50118426 on Build GRCh38
rs974783083 Chr.12: 50118745-50118745 on Build GRCh38
rs898856014 Chr.12: 50118463-50118463 on Build GRCh38
rs1046046786 Chr.12: 50118639-50118639 on Build GRCh38
rs12815644 Chr.12: 50118659-50118659 on Build GRCh38
rs576140687 Chr.12: 50118509-50118509 on Build GRCh38
rs920371642 Chr.12: 50118760-50118760 on Build GRCh38
rs868223114 Chr.12: 50118432-50118432 on Build GRCh38
rs540248928 Chr.12: 50118653-50118653 on Build GRCh38
rs112929561 Chr.12: 50118583-50118583 on Build GRCh38
rs949972907 Chr.12: 50118637-50118637 on Build GRCh38
rs1047568733 Chr.12: 50118586-50118586 on Build GRCh38
rs907739297 Chr.12: 50118766-50118766 on Build GRCh38
rs978685288 Chr.12: 50118403-50118403 on Build GRCh38
rs577152591 Chr.12: 50118776-50118776 on Build GRCh38
rs189329745 Chr.12: 50118402-50118402 on Build GRCh38
rs1002575491 Chr.12: 50118648-50118648 on Build GRCh38
rs12815933 Chr.12: 50118647-50118647 on Build GRCh38
rs563140671 Chr.12: 50118721-50118721 on Build GRCh38
rs921853446 Chr.12: 50118646-50118646 on Build GRCh38
rs886375761 Chr.12: 50118631-50118631 on Build GRCh38
rs369964487 Chr.12: 50118542-50118542 on Build GRCh38
rs115439264 Chr.12: 50118834-50118834 on Build GRCh38
rs983181646 Chr.12: 50118763-50118763 on Build GRCh38
rs905412990 Chr.12: 50118506-50118506 on Build GRCh38
rs112442737 Chr.12: 50118715-50118715 on Build GRCh38
rs12815927 Chr.12: 50118640-50118640 on Build GRCh38
rs552239161 Chr.12: 50118716-50118716 on Build GRCh38
rs940423457 Chr.12: 50118438-50118438 on Build GRCh38
rs966835824 Chr.12: 50118722-50118722 on Build GRCh38
rs953296576 Chr.12: 50118665-50118665 on Build GRCh38
rs949856532 Chr.12: 50118774-50118774 on Build GRCh38
rs74365295 Chr.12: 50118846-50118846 on Build GRCh38

View Full Product Details