Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51906993-51907499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVRL1
Entrez Gene ID
94
Gene Name
activin A receptor like type 1
Gene Aliases
ACVRLK1, ALK-1, ALK1, HHT, HHT2, ORW2, SKR3, TSR-I
UniGene
Hs.591026
Gene Chromosome Location
Chr.12: 51906913-51923361 on Build GRCh38
Transcripts
NM_000020.2 XM_005269235.2

SNP Detail

SNP ID Chromosome Location
rs1002184603 Chr.12: 51907469-51907469 on Build GRCh38
rs907552900 Chr.12: 51907211-51907211 on Build GRCh38
rs974805458 Chr.12: 51907309-51907309 on Build GRCh38
rs1053167705 Chr.12: 51907366-51907366 on Build GRCh38
rs1022158658 Chr.12: 51907219-51907219 on Build GRCh38
rs1046281153 Chr.12: 51907390-51907390 on Build GRCh38
rs866193526 Chr.12: 51907319-51907319 on Build GRCh38
rs920174872 Chr.12: 51907274-51907274 on Build GRCh38
rs907262231 Chr.12: 51907098-51907098 on Build GRCh38
rs1010655704 Chr.12: 51907209-51907209 on Build GRCh38
rs1039421673 Chr.12: 51907106-51907106 on Build GRCh38
rs974573319 Chr.12: 51907169-51907169 on Build GRCh38
rs911830063 Chr.12: 51907242-51907242 on Build GRCh38
rs1005122807 Chr.12: 51907148-51907148 on Build GRCh38
rs559417637 Chr.12: 51907051-51907051 on Build GRCh38
rs1021592881 Chr.12: 51907407-51907407 on Build GRCh38
rs867160753 Chr.12: 51907132-51907132 on Build GRCh38
rs968153463 Chr.12: 51907024-51907024 on Build GRCh38
rs1003385221 Chr.12: 51907363-51907363 on Build GRCh38
rs537514943 Chr.12: 51907448-51907448 on Build GRCh38
rs915067878 Chr.12: 51907058-51907058 on Build GRCh38
rs369090024 Chr.12: 51907061-51907061 on Build GRCh38
rs907338036 Chr.12: 51907327-51907327 on Build GRCh38
rs964606570 Chr.12: 51907465-51907465 on Build GRCh38
rs930721823 Chr.12: 51907115-51907115 on Build GRCh38
rs74094823 Chr.12: 51907111-51907111 on Build GRCh38
rs966520177 Chr.12: 51907220-51907220 on Build GRCh38
rs535774871 Chr.12: 51907184-51907184 on Build GRCh38
rs982157929 Chr.12: 51907018-51907018 on Build GRCh38
rs369003431 Chr.12: 51907223-51907223 on Build GRCh38
rs2272028 Chr.12: 51907291-51907291 on Build GRCh38
rs549261187 Chr.12: 51907199-51907199 on Build GRCh38
rs1022454310 Chr.12: 51907020-51907020 on Build GRCh38
rs983193222 Chr.12: 51907205-51907205 on Build GRCh38
rs534717776 Chr.12: 51907271-51907271 on Build GRCh38
rs55640904 Chr.12: 51907415-51907415 on Build GRCh38
rs1013029717 Chr.12: 51907067-51907067 on Build GRCh38
rs551317561 Chr.12: 51907072-51907072 on Build GRCh38
rs180906879 Chr.12: 51907129-51907129 on Build GRCh38
rs907458444 Chr.12: 51907415-51907415 on Build GRCh38
rs948911318 Chr.12: 51907302-51907302 on Build GRCh38
rs558085960 Chr.12: 51907326-51907326 on Build GRCh38
rs907938490 Chr.12: 51907335-51907335 on Build GRCh38
rs891811924 Chr.12: 51907375-51907375 on Build GRCh38
rs921925286 Chr.12: 51907316-51907316 on Build GRCh38
rs1033728241 Chr.12: 51907196-51907196 on Build GRCh38
rs571668567 Chr.12: 51907350-51907350 on Build GRCh38
rs1033454878 Chr.12: 51907026-51907026 on Build GRCh38
rs140213364 Chr.12: 51907177-51907177 on Build GRCh38
rs940291429 Chr.12: 51907060-51907060 on Build GRCh38
rs944736755 Chr.12: 51907099-51907099 on Build GRCh38
rs959341105 Chr.12: 51907029-51907029 on Build GRCh38
rs1047831624 Chr.12: 51907022-51907022 on Build GRCh38
rs1011578184 Chr.12: 51907387-51907387 on Build GRCh38
rs898675708 Chr.12: 51907066-51907066 on Build GRCh38
rs930752810 Chr.12: 51907325-51907325 on Build GRCh38
rs945312116 Chr.12: 51907059-51907059 on Build GRCh38
rs550122766 Chr.12: 51907153-51907153 on Build GRCh38

View Full Product Details