Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53268663-53269157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPL1
Entrez Gene ID
9700
Gene Name
extra spindle pole bodies like 1, separase
Gene Aliases
ESP1, SEPA
UniGene
Hs.153479
Gene Chromosome Location
Chr.12: 53268257-53293643 on Build GRCh38
Transcripts
NM_012291.4 XM_006719705.3 XM_011539024.2 XM_011539025.2 XM_017020253.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3398846 c.9C>A p.S3R Chr.12: 53268775-53268775 on Build GRCh38
3871937 c.12C>T p.F4F Chr.12: 53268778-53268778 on Build GRCh38
468570 c.176A>G p.Q59R Chr.12: 53269118-53269118 on Build GRCh38
5789924 c.24C>G p.N8K Chr.12: 53268790-53268790 on Build GRCh38
1606313 c.157A>G p.I53V Chr.12: 53269099-53269099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759933987 Chr.12: 53269109-53269109 on Build GRCh38
rs1028498208 Chr.12: 53268700-53268700 on Build GRCh38
rs753884441 Chr.12: 53268787-53268787 on Build GRCh38
rs752741826 Chr.12: 53268996-53268996 on Build GRCh38
rs866558693 Chr.12: 53269047-53269047 on Build GRCh38
rs748301352 Chr.12: 53268796-53268796 on Build GRCh38
rs760936390 Chr.12: 53268949-53268949 on Build GRCh38
rs747770966 Chr.12: 53269082-53269082 on Build GRCh38
rs755890836 Chr.12: 53268876-53268876 on Build GRCh38
rs777010588 Chr.12: 53268952-53268952 on Build GRCh38
rs756533275 Chr.12: 53268748-53268748 on Build GRCh38
rs768270715 Chr.12: 53269059-53269059 on Build GRCh38
rs763834441 Chr.12: 53268869-53268869 on Build GRCh38
rs1047224309 Chr.12: 53268937-53268937 on Build GRCh38
rs568840171 Chr.12: 53268693-53268693 on Build GRCh38
rs764204340 Chr.12: 53269019-53269019 on Build GRCh38
rs139777651 Chr.12: 53269029-53269029 on Build GRCh38
rs765258526 Chr.12: 53268902-53268902 on Build GRCh38
rs770277802 Chr.12: 53269118-53269118 on Build GRCh38
rs778301697 Chr.12: 53269069-53269069 on Build GRCh38
rs6580942 Chr.12: 53268840-53268840 on Build GRCh38
rs768163030 Chr.12: 53268941-53268941 on Build GRCh38
rs749524259 Chr.12: 53268957-53268962 on Build GRCh38
rs984426497 Chr.12: 53268795-53268795 on Build GRCh38
rs142528308 Chr.12: 53268793-53268793 on Build GRCh38
rs776118214 Chr.12: 53269130-53269130 on Build GRCh38
rs763155697 Chr.12: 53269012-53269012 on Build GRCh38
rs756103099 Chr.12: 53269053-53269053 on Build GRCh38
rs770738469 Chr.12: 53268835-53268835 on Build GRCh38
rs939106041 Chr.12: 53268879-53268879 on Build GRCh38
rs757131290 Chr.12: 53268927-53268927 on Build GRCh38
rs753063106 Chr.12: 53268854-53268854 on Build GRCh38
rs759985301 Chr.12: 53268978-53268978 on Build GRCh38
rs776772153 Chr.12: 53268838-53268838 on Build GRCh38
rs750268547 Chr.12: 53269050-53269050 on Build GRCh38
rs776957781 Chr.12: 53269107-53269107 on Build GRCh38
rs771733095 Chr.12: 53269089-53269089 on Build GRCh38
rs760040291 Chr.12: 53268891-53268891 on Build GRCh38
rs777383104 Chr.12: 53268814-53268814 on Build GRCh38
rs748881484 Chr.12: 53268940-53268940 on Build GRCh38
rs757421350 Chr.12: 53269037-53269037 on Build GRCh38
rs781096929 Chr.12: 53269049-53269049 on Build GRCh38
rs769886071 Chr.12: 53268939-53268939 on Build GRCh38
rs575434980 Chr.12: 53268925-53268925 on Build GRCh38
rs777920507 Chr.12: 53268901-53268902 on Build GRCh38
rs141171220 Chr.12: 53269058-53269058 on Build GRCh38
rs752796906 Chr.12: 53268915-53268915 on Build GRCh38
rs781145616 Chr.12: 53268933-53268933 on Build GRCh38
rs773431035 Chr.12: 53268871-53268871 on Build GRCh38
rs758525990 Chr.12: 53268921-53268921 on Build GRCh38
rs766960012 Chr.12: 53268760-53268760 on Build GRCh38
rs1043672620 Chr.12: 53268965-53268965 on Build GRCh38
rs746869871 Chr.12: 53268827-53268827 on Build GRCh38
rs754402636 Chr.12: 53268889-53268889 on Build GRCh38
rs778093522 Chr.12: 53268924-53268924 on Build GRCh38
rs866891946 Chr.12: 53268910-53268910 on Build GRCh38
rs189340711 Chr.12: 53268861-53268861 on Build GRCh38
rs976884887 Chr.12: 53268685-53268685 on Build GRCh38
rs957586490 Chr.12: 53268820-53268820 on Build GRCh38
rs4758965 Chr.12: 53268951-53268951 on Build GRCh38
rs902446506 Chr.12: 53268967-53268967 on Build GRCh38
rs977883078 Chr.12: 53268792-53268792 on Build GRCh38
rs998837399 Chr.12: 53268989-53268989 on Build GRCh38
rs998311078 Chr.12: 53269133-53269133 on Build GRCh38
rs752573806 Chr.12: 53268743-53268743 on Build GRCh38
rs1024444937 Chr.12: 53268703-53268703 on Build GRCh38
rs971591767 Chr.12: 53268755-53268755 on Build GRCh38
rs761142631 Chr.12: 53268877-53268877 on Build GRCh38
rs774119792 Chr.12: 53268946-53268946 on Build GRCh38
rs372965192 Chr.12: 53268878-53268878 on Build GRCh38
rs765782026 Chr.12: 53268986-53268986 on Build GRCh38
rs145977146 Chr.12: 53268800-53268800 on Build GRCh38
rs546034970 Chr.12: 53268934-53268934 on Build GRCh38

View Full Product Details