Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53274946-53275488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPL1
Entrez Gene ID
9700
Gene Name
extra spindle pole bodies like 1, separase
Gene Aliases
ESP1, SEPA
UniGene
Hs.153479
Gene Chromosome Location
Chr.12: 53268257-53293643 on Build GRCh38
Transcripts
NM_012291.4 XM_006719705.3 XM_011539024.2 XM_011539025.2 XM_017020253.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1932722 c.1651C>T p.R551W Chr.12: 53274961-53274961 on Build GRCh38
292310 c.1692A>G p.L564L Chr.12: 53275002-53275002 on Build GRCh38
6072919 c.1656C>G p.V552V Chr.12: 53274966-53274966 on Build GRCh38
239745 c.1667C>T p.A556V Chr.12: 53274977-53274977 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs535430306 Chr.12: 53275410-53275410 on Build GRCh38
rs12833691 Chr.12: 53275231-53275231 on Build GRCh38
rs897854387 Chr.12: 53275353-53275353 on Build GRCh38
rs977700148 Chr.12: 53275448-53275448 on Build GRCh38
rs546107819 Chr.12: 53274986-53274986 on Build GRCh38
rs1004125243 Chr.12: 53275322-53275322 on Build GRCh38
rs764765809 Chr.12: 53275069-53275069 on Build GRCh38
rs1036300976 Chr.12: 53275329-53275329 on Build GRCh38
rs527439130 Chr.12: 53275186-53275186 on Build GRCh38
rs1035753155 Chr.12: 53275392-53275392 on Build GRCh38
rs191843684 Chr.12: 53275415-53275415 on Build GRCh38
rs1003296717 Chr.12: 53275375-53275375 on Build GRCh38
rs528831111 Chr.12: 53275031-53275031 on Build GRCh38
rs1048223815 Chr.12: 53275316-53275316 on Build GRCh38
rs559401338 Chr.12: 53275082-53275082 on Build GRCh38
rs924298957 Chr.12: 53275239-53275239 on Build GRCh38
rs1025725112 Chr.12: 53275118-53275118 on Build GRCh38
rs921033541 Chr.12: 53275279-53275279 on Build GRCh38
rs1031211087 Chr.12: 53275091-53275091 on Build GRCh38
rs118113293 Chr.12: 53275179-53275179 on Build GRCh38
rs962435602 Chr.12: 53275087-53275087 on Build GRCh38
rs756967197 Chr.12: 53275021-53275021 on Build GRCh38
rs965051879 Chr.12: 53275462-53275462 on Build GRCh38
rs932462912 Chr.12: 53275290-53275290 on Build GRCh38
rs981572948 Chr.12: 53275128-53275128 on Build GRCh38
rs968943127 Chr.12: 53275447-53275447 on Build GRCh38
rs183453232 Chr.12: 53275129-53275129 on Build GRCh38
rs199711618 Chr.12: 53275029-53275029 on Build GRCh38
rs1018556870 Chr.12: 53275035-53275035 on Build GRCh38
rs751104677 Chr.12: 53275020-53275020 on Build GRCh38
rs187071253 Chr.12: 53275168-53275168 on Build GRCh38
rs909851075 Chr.12: 53275202-53275202 on Build GRCh38
rs1006143879 Chr.12: 53275043-53275043 on Build GRCh38
rs534613951 Chr.12: 53275233-53275233 on Build GRCh38
rs377322721 Chr.12: 53275038-53275038 on Build GRCh38
rs924872195 Chr.12: 53275459-53275459 on Build GRCh38
rs779634274 Chr.12: 53275048-53275048 on Build GRCh38
rs568128800 Chr.12: 53275247-53275247 on Build GRCh38
rs569798538 Chr.12: 53275157-53275157 on Build GRCh38
rs1014912541 Chr.12: 53275086-53275086 on Build GRCh38
rs959042941 Chr.12: 53275393-53275393 on Build GRCh38
rs902355979 Chr.12: 53275429-53275429 on Build GRCh38
rs976956708 Chr.12: 53275094-53275094 on Build GRCh38
rs552126006 Chr.12: 53275174-53275174 on Build GRCh38
rs867042483 Chr.12: 53275333-53275333 on Build GRCh38
rs758496811 Chr.12: 53275059-53275059 on Build GRCh38
rs977085412 Chr.12: 53275229-53275229 on Build GRCh38
rs888356436 Chr.12: 53275321-53275321 on Build GRCh38
rs758258936 Chr.12: 53275234-53275234 on Build GRCh38
rs537136245 Chr.12: 53275167-53275167 on Build GRCh38
rs529682457 Chr.12: 53275103-53275103 on Build GRCh38
rs552805888 Chr.12: 53275243-53275243 on Build GRCh38
rs762507993 Chr.12: 53274984-53274984 on Build GRCh38
rs547227185 Chr.12: 53275053-53275053 on Build GRCh38
rs376306182 Chr.12: 53275054-53275054 on Build GRCh38
rs999289983 Chr.12: 53275077-53275077 on Build GRCh38
rs958872225 Chr.12: 53275156-53275156 on Build GRCh38
rs758066818 Chr.12: 53275015-53275015 on Build GRCh38
rs949221064 Chr.12: 53275367-53275367 on Build GRCh38
rs373279969 Chr.12: 53275028-53275028 on Build GRCh38

View Full Product Details