Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53669783-53670284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5G2
Entrez Gene ID
517
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9)
Gene Aliases
ATP5A
UniGene
Hs.524464
Gene Chromosome Location
Chr.12: 53665160-53677546 on Build GRCh38
Transcripts
NM_001002031.2 NM_005176.5 XM_017019460.1 XM_017019461.1 XM_017019462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7422576 c.265C>T p.R89* Chr.12: 53669894-53669894 on Build GRCh38
4907553 c.230T>C p.L77P Chr.12: 53669929-53669929 on Build GRCh38
1492985 c.109C>T p.L37L Chr.12: 53669927-53669927 on Build GRCh38
7422577 c.142C>T p.R48* Chr.12: 53669894-53669894 on Build GRCh38
4899338 c.286G>A p.E96K Chr.12: 53669873-53669873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775752384 Chr.12: 53669823-53669823 on Build GRCh38
rs566986912 Chr.12: 53670131-53670131 on Build GRCh38
rs368833150 Chr.12: 53670199-53670199 on Build GRCh38
rs372557722 Chr.12: 53669836-53669836 on Build GRCh38
rs749900837 Chr.12: 53669851-53669851 on Build GRCh38
rs1008251571 Chr.12: 53670049-53670049 on Build GRCh38
rs937088596 Chr.12: 53670039-53670039 on Build GRCh38
rs368109750 Chr.12: 53670200-53670200 on Build GRCh38
rs772188326 Chr.12: 53669937-53669937 on Build GRCh38
rs773254587 Chr.12: 53669951-53669951 on Build GRCh38
rs79533383 Chr.12: 53670085-53670085 on Build GRCh38
rs140613495 Chr.12: 53669893-53669893 on Build GRCh38
rs753526098 Chr.12: 53669867-53669867 on Build GRCh38
rs747999624 Chr.12: 53670083-53670083 on Build GRCh38
rs768077338 Chr.12: 53670025-53670025 on Build GRCh38
rs201383280 Chr.12: 53669978-53669978 on Build GRCh38
rs374657376 Chr.12: 53669917-53669917 on Build GRCh38
rs768487576 Chr.12: 53669835-53669835 on Build GRCh38
rs371287380 Chr.12: 53669857-53669857 on Build GRCh38
rs929669269 Chr.12: 53669980-53669980 on Build GRCh38
rs560731277 Chr.12: 53669895-53669895 on Build GRCh38
rs762053300 Chr.12: 53669894-53669894 on Build GRCh38
rs900119975 Chr.12: 53670066-53670066 on Build GRCh38
rs765470874 Chr.12: 53669969-53669977 on Build GRCh38
rs914616966 Chr.12: 53669948-53669948 on Build GRCh38
rs765985656 Chr.12: 53669863-53669863 on Build GRCh38
rs749465320 Chr.12: 53669913-53669913 on Build GRCh38
rs1014173219 Chr.12: 53669843-53669843 on Build GRCh38
rs768344833 Chr.12: 53669916-53669916 on Build GRCh38
rs772677456 Chr.12: 53670028-53670028 on Build GRCh38
rs776916019 Chr.12: 53669967-53669967 on Build GRCh38
rs918240185 Chr.12: 53669979-53669979 on Build GRCh38
rs756157505 Chr.12: 53670030-53670030 on Build GRCh38
rs1053585588 Chr.12: 53669921-53669921 on Build GRCh38
rs909662704 Chr.12: 53670017-53670017 on Build GRCh38
rs539259937 Chr.12: 53670037-53670037 on Build GRCh38
rs755182398 Chr.12: 53670055-53670055 on Build GRCh38
rs764461297 Chr.12: 53669889-53669889 on Build GRCh38
rs748122288 Chr.12: 53669930-53669930 on Build GRCh38
rs1018826680 Chr.12: 53670239-53670239 on Build GRCh38
rs761078549 Chr.12: 53670119-53670119 on Build GRCh38
rs981833209 Chr.12: 53670189-53670189 on Build GRCh38
rs568994833 Chr.12: 53670018-53670018 on Build GRCh38
rs761740190 Chr.12: 53669967-53669982 on Build GRCh38
rs13328931 Chr.12: 53669822-53669822 on Build GRCh38
rs756211944 Chr.12: 53669909-53669909 on Build GRCh38
rs958186001 Chr.12: 53669910-53669910 on Build GRCh38
rs1032316640 Chr.12: 53670171-53670171 on Build GRCh38
rs534323908 Chr.12: 53670256-53670256 on Build GRCh38
rs959283628 Chr.12: 53670226-53670226 on Build GRCh38
rs762726781 Chr.12: 53669830-53669830 on Build GRCh38
rs751968676 Chr.12: 53670009-53670009 on Build GRCh38
rs181068632 Chr.12: 53669999-53669999 on Build GRCh38
rs779019321 Chr.12: 53669972-53669973 on Build GRCh38
rs746906986 Chr.12: 53670120-53670120 on Build GRCh38
rs779259027 Chr.12: 53670074-53670074 on Build GRCh38
rs370229773 Chr.12: 53669848-53669848 on Build GRCh38
rs764387964 Chr.12: 53669992-53669992 on Build GRCh38
rs776161633 Chr.12: 53669986-53669986 on Build GRCh38
rs566625928 Chr.12: 53669920-53669920 on Build GRCh38
rs749506992 Chr.12: 53670045-53670045 on Build GRCh38
rs375797649 Chr.12: 53669837-53669837 on Build GRCh38
rs970784956 Chr.12: 53670172-53670172 on Build GRCh38
rs778725444 Chr.12: 53670240-53670242 on Build GRCh38
rs773342703 Chr.12: 53670110-53670110 on Build GRCh38
rs1027158694 Chr.12: 53670079-53670079 on Build GRCh38
rs776157118 Chr.12: 53670133-53670133 on Build GRCh38
rs1006933390 Chr.12: 53670233-53670233 on Build GRCh38
rs758748203 Chr.12: 53669882-53669882 on Build GRCh38
rs759188547 Chr.12: 53669989-53669989 on Build GRCh38
rs760329775 Chr.12: 53669964-53669964 on Build GRCh38
rs1015238551 Chr.12: 53670158-53670158 on Build GRCh38
rs141952979 Chr.12: 53670075-53670076 on Build GRCh38
rs780195754 Chr.12: 53669911-53669911 on Build GRCh38
rs977039397 Chr.12: 53669974-53669974 on Build GRCh38
rs1055708428 Chr.12: 53669828-53669828 on Build GRCh38
rs752091763 Chr.12: 53669890-53669890 on Build GRCh38
rs745985019 Chr.12: 53669899-53669899 on Build GRCh38

View Full Product Details