Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53676361-53676867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5G2
Entrez Gene ID
517
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9)
Gene Aliases
ATP5A
UniGene
Hs.524464
Gene Chromosome Location
Chr.12: 53665160-53677546 on Build GRCh38
Transcripts
NM_001002031.2 XM_017019460.1 XM_017019461.1

SNP Detail

SNP ID Chromosome Location
rs569080682 Chr.12: 53676567-53676567 on Build GRCh38
rs1005592144 Chr.12: 53676702-53676702 on Build GRCh38
rs894945667 Chr.12: 53676419-53676419 on Build GRCh38
rs977333971 Chr.12: 53676456-53676456 on Build GRCh38
rs965388390 Chr.12: 53676544-53676544 on Build GRCh38
rs142467861 Chr.12: 53676764-53676764 on Build GRCh38
rs923963980 Chr.12: 53676506-53676506 on Build GRCh38
rs1036687049 Chr.12: 53676399-53676399 on Build GRCh38
rs769912706 Chr.12: 53676548-53676548 on Build GRCh38
rs193256885 Chr.12: 53676462-53676462 on Build GRCh38
rs984732549 Chr.12: 53676658-53676658 on Build GRCh38
rs555991004 Chr.12: 53676675-53676675 on Build GRCh38
rs971244609 Chr.12: 53676795-53676795 on Build GRCh38
rs1022578421 Chr.12: 53676734-53676734 on Build GRCh38
rs746464898 Chr.12: 53676385-53676385 on Build GRCh38
rs1024105945 Chr.12: 53676429-53676429 on Build GRCh38
rs1021709760 Chr.12: 53676480-53676480 on Build GRCh38
rs527466220 Chr.12: 53676677-53676677 on Build GRCh38
rs571640108 Chr.12: 53676666-53676666 on Build GRCh38
rs977406232 Chr.12: 53676514-53676514 on Build GRCh38
rs760570469 Chr.12: 53676440-53676440 on Build GRCh38
rs762851319 Chr.12: 53676597-53676597 on Build GRCh38
rs775147173 Chr.12: 53676583-53676583 on Build GRCh38
rs1011159478 Chr.12: 53676437-53676437 on Build GRCh38
rs1036761999 Chr.12: 53676673-53676673 on Build GRCh38
rs35259777 Chr.12: 53676565-53676566 on Build GRCh38
rs866846835 Chr.12: 53676595-53676595 on Build GRCh38
rs1024074639 Chr.12: 53676781-53676781 on Build GRCh38
rs770592388 Chr.12: 53676418-53676418 on Build GRCh38
rs1010732318 Chr.12: 53676729-53676729 on Build GRCh38
rs562354580 Chr.12: 53676592-53676615 on Build GRCh38
rs1039864228 Chr.12: 53676570-53676570 on Build GRCh38
rs1003393537 Chr.12: 53676407-53676407 on Build GRCh38
rs895095396 Chr.12: 53676712-53676712 on Build GRCh38
rs906444908 Chr.12: 53676403-53676403 on Build GRCh38
rs539931434 Chr.12: 53676584-53676584 on Build GRCh38
rs934210929 Chr.12: 53676647-53676647 on Build GRCh38
rs968824565 Chr.12: 53676513-53676513 on Build GRCh38
rs904588743 Chr.12: 53676744-53676744 on Build GRCh38
rs915272269 Chr.12: 53676551-53676551 on Build GRCh38
rs932794956 Chr.12: 53676626-53676626 on Build GRCh38
rs977001208 Chr.12: 53676607-53676607 on Build GRCh38
rs894202411 Chr.12: 53676752-53676752 on Build GRCh38
rs3892761 Chr.12: 53676661-53676661 on Build GRCh38
rs774642410 Chr.12: 53676559-53676562 on Build GRCh38
rs1055012357 Chr.12: 53676710-53676710 on Build GRCh38
rs148760549 Chr.12: 53676390-53676390 on Build GRCh38
rs745815869 Chr.12: 53676541-53676541 on Build GRCh38
rs939030823 Chr.12: 53676708-53676708 on Build GRCh38
rs776641706 Chr.12: 53676532-53676532 on Build GRCh38
rs759789375 Chr.12: 53676610-53676610 on Build GRCh38
rs956955407 Chr.12: 53676516-53676516 on Build GRCh38
rs989503789 Chr.12: 53676543-53676543 on Build GRCh38
rs551915357 Chr.12: 53676631-53676631 on Build GRCh38

View Full Product Details