Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54028723-54029234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXC4
Entrez Gene ID
3221
Gene Name
homeobox C4
Gene Aliases
HOX3, HOX3E, cp19
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54056030 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Gene Symbol
HOXC5
Entrez Gene ID
3222
Gene Name
homeobox C5
Gene Aliases
CP11, HOX3, HOX3D
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54035361 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Gene Symbol
HOXC6
Entrez Gene ID
3223
Gene Name
homeobox C6
Gene Aliases
CP25, HHO.C8, HOX3, HOX3C
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54030823 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1299597 c.348G>C p.Q116H Chr.12: 54028869-54028869 on Build GRCh38
4152877 c.277G>A p.G93R Chr.12: 54028798-54028798 on Build GRCh38
7241464 c.29T>C p.L10S Chr.12: 54028796-54028796 on Build GRCh38
6503046 c.374G>T p.W125L Chr.12: 54028895-54028895 on Build GRCh38
7211346 c.52A>G p.I18V Chr.12: 54028819-54028819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924424381 Chr.12: 54029168-54029168 on Build GRCh38
rs560916042 Chr.12: 54029172-54029172 on Build GRCh38
rs947512898 Chr.12: 54028995-54028995 on Build GRCh38
rs767298048 Chr.12: 54028771-54028771 on Build GRCh38
rs762991519 Chr.12: 54028884-54028884 on Build GRCh38
rs550099853 Chr.12: 54028755-54028755 on Build GRCh38
rs372116179 Chr.12: 54029047-54029047 on Build GRCh38
rs185812647 Chr.12: 54029154-54029154 on Build GRCh38
rs200362917 Chr.12: 54028945-54028945 on Build GRCh38
rs1013650194 Chr.12: 54029186-54029187 on Build GRCh38
rs565847774 Chr.12: 54028819-54028819 on Build GRCh38
rs993648202 Chr.12: 54028940-54028940 on Build GRCh38
rs764735870 Chr.12: 54028960-54028960 on Build GRCh38
rs771424142 Chr.12: 54028860-54028860 on Build GRCh38
rs752407433 Chr.12: 54028964-54028964 on Build GRCh38
rs532429929 Chr.12: 54028780-54028780 on Build GRCh38
rs772945356 Chr.12: 54028938-54028938 on Build GRCh38
rs991870481 Chr.12: 54028976-54028976 on Build GRCh38
rs959018834 Chr.12: 54029120-54029120 on Build GRCh38
rs764263179 Chr.12: 54028764-54028764 on Build GRCh38
rs754482979 Chr.12: 54028804-54028804 on Build GRCh38
rs768769275 Chr.12: 54028892-54028892 on Build GRCh38
rs779048299 Chr.12: 54029093-54029093 on Build GRCh38
rs982131346 Chr.12: 54028939-54028939 on Build GRCh38
rs553311752 Chr.12: 54029127-54029127 on Build GRCh38
rs761655147 Chr.12: 54028768-54028768 on Build GRCh38
rs367798571 Chr.12: 54028944-54028944 on Build GRCh38
rs574940018 Chr.12: 54029129-54029129 on Build GRCh38
rs1006428728 Chr.12: 54029158-54029158 on Build GRCh38
rs758073273 Chr.12: 54028968-54028968 on Build GRCh38
rs977993580 Chr.12: 54029017-54029017 on Build GRCh38
rs879224445 Chr.12: 54028856-54028856 on Build GRCh38
rs984951104 Chr.12: 54028801-54028801 on Build GRCh38
rs201594434 Chr.12: 54028765-54028765 on Build GRCh38
rs111518456 Chr.12: 54028800-54028800 on Build GRCh38
rs982555335 Chr.12: 54029086-54029086 on Build GRCh38
rs775380956 Chr.12: 54028751-54028751 on Build GRCh38
rs80157375 Chr.12: 54028891-54028891 on Build GRCh38
rs1015089034 Chr.12: 54029092-54029092 on Build GRCh38
rs376534127 Chr.12: 54028867-54028867 on Build GRCh38
rs936514158 Chr.12: 54029034-54029034 on Build GRCh38
rs770158000 Chr.12: 54028873-54028873 on Build GRCh38
rs576498700 Chr.12: 54028897-54028897 on Build GRCh38
rs766272049 Chr.12: 54028942-54028942 on Build GRCh38
rs200862396 Chr.12: 54028932-54028932 on Build GRCh38
rs1025297090 Chr.12: 54028979-54028979 on Build GRCh38
rs981676089 Chr.12: 54029110-54029110 on Build GRCh38
rs899741088 Chr.12: 54029204-54029204 on Build GRCh38
rs1024571922 Chr.12: 54029203-54029204 on Build GRCh38
rs148984909 Chr.12: 54028941-54028941 on Build GRCh38
rs796996772 Chr.12: 54029058-54029058 on Build GRCh38
rs908083356 Chr.12: 54028803-54028803 on Build GRCh38
rs541688247 Chr.12: 54029109-54029109 on Build GRCh38
rs899459501 Chr.12: 54029048-54029048 on Build GRCh38
rs910672367 Chr.12: 54029070-54029070 on Build GRCh38
rs952440673 Chr.12: 54029058-54029058 on Build GRCh38
rs1026685919 Chr.12: 54029057-54029057 on Build GRCh38
rs929515341 Chr.12: 54029145-54029145 on Build GRCh38
rs571612814 Chr.12: 54028773-54028773 on Build GRCh38
rs761958008 Chr.12: 54028919-54028919 on Build GRCh38
rs996925503 Chr.12: 54029054-54029054 on Build GRCh38
rs747794723 Chr.12: 54028858-54028858 on Build GRCh38
rs776708657 Chr.12: 54028844-54028844 on Build GRCh38
rs914597343 Chr.12: 54029148-54029148 on Build GRCh38
rs1040935142 Chr.12: 54029179-54029179 on Build GRCh38
rs1041513274 Chr.12: 54029006-54029006 on Build GRCh38
rs779640921 Chr.12: 54028795-54028795 on Build GRCh38
rs558447354 Chr.12: 54028802-54028802 on Build GRCh38
rs755990418 Chr.12: 54028788-54028788 on Build GRCh38
rs903052134 Chr.12: 54029018-54029018 on Build GRCh38
rs1008115998 Chr.12: 54029037-54029037 on Build GRCh38
rs1041796765 Chr.12: 54029156-54029156 on Build GRCh38
rs371058976 Chr.12: 54028876-54028876 on Build GRCh38
rs774503554 Chr.12: 54028917-54028917 on Build GRCh38
rs928888084 Chr.12: 54029111-54029111 on Build GRCh38
rs962219583 Chr.12: 54029108-54029108 on Build GRCh38
rs781639621 Chr.12: 54028866-54028866 on Build GRCh38

View Full Product Details