Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56237504-56238001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD52
Entrez Gene ID
283373
Gene Name
ankyrin repeat domain 52
Gene Aliases
ANKRD33
UniGene
Hs.524506
Gene Chromosome Location
Chr.12: 56237807-56258391 on Build GRCh38
Transcripts
NM_173595.3 NM_001135195.1 NM_173596.2 XM_005268803.1 XM_011538198.1 XM_011538199.1 XM_011538200.1 XM_011538201.1 XM_017019185.1 XM_017019186.1 XM_017019187.1

Gene Symbol
SLC39A5
Entrez Gene ID
283375
Gene Name
solute carrier family 39 member 5
Gene Aliases
LZT-Hs7, MYP24, ZIP5
UniGene
Hs.591018
Gene Chromosome Location
Chr.12: 56230036-56237846 on Build GRCh38
Transcripts
NM_173595.3 NM_001135195.1 NM_173596.2 XM_005268803.1 XM_011538198.1 XM_011538199.1 XM_011538200.1 XM_011538201.1 XM_017019185.1 XM_017019186.1 XM_017019187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4186780 c.1492C>T p.L498F Chr.12: 56237600-56237600 on Build GRCh38
5034606 c.1552C>A p.L518M Chr.12: 56237660-56237660 on Build GRCh38
4727344 c.1533G>A p.L511L Chr.12: 56237641-56237641 on Build GRCh38
263724 c.1605C>T p.P535P Chr.12: 56237713-56237713 on Build GRCh38
6782168 c.1495C>T p.R499C Chr.12: 56237603-56237603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866578114 Chr.12: 56237901-56237901 on Build GRCh38
rs371218776 Chr.12: 56237712-56237712 on Build GRCh38
rs969773696 Chr.12: 56237958-56237958 on Build GRCh38
rs768295346 Chr.12: 56237552-56237552 on Build GRCh38
rs764347098 Chr.12: 56237684-56237684 on Build GRCh38
rs771608343 Chr.12: 56237654-56237654 on Build GRCh38
rs576250198 Chr.12: 56237874-56237874 on Build GRCh38
rs1046716283 Chr.12: 56237875-56237875 on Build GRCh38
rs374918493 Chr.12: 56237611-56237611 on Build GRCh38
rs371179560 Chr.12: 56237604-56237604 on Build GRCh38
rs997432366 Chr.12: 56237914-56237914 on Build GRCh38
rs889834890 Chr.12: 56237792-56237792 on Build GRCh38
rs767783104 Chr.12: 56237775-56237775 on Build GRCh38
rs760089709 Chr.12: 56237663-56237663 on Build GRCh38
rs981472748 Chr.12: 56237887-56237887 on Build GRCh38
rs551214351 Chr.12: 56237816-56237816 on Build GRCh38
rs144141700 Chr.12: 56237703-56237703 on Build GRCh38
rs2293511 Chr.12: 56237625-56237625 on Build GRCh38
rs78460555 Chr.12: 56237580-56237580 on Build GRCh38
rs542763166 Chr.12: 56237830-56237830 on Build GRCh38
rs766893342 Chr.12: 56237569-56237569 on Build GRCh38
rs776367522 Chr.12: 56237661-56237662 on Build GRCh38
rs772719415 Chr.12: 56237751-56237751 on Build GRCh38
rs764342375 Chr.12: 56237595-56237595 on Build GRCh38
rs760003558 Chr.12: 56237574-56237574 on Build GRCh38
rs553923289 Chr.12: 56237761-56237761 on Build GRCh38
rs147102571 Chr.12: 56237610-56237610 on Build GRCh38
rs368587109 Chr.12: 56237754-56237754 on Build GRCh38
rs767764540 Chr.12: 56237702-56237702 on Build GRCh38
rs1051278495 Chr.12: 56237753-56237753 on Build GRCh38
rs202198891 Chr.12: 56237857-56237857 on Build GRCh38
rs201955617 Chr.12: 56237549-56237549 on Build GRCh38
rs781391170 Chr.12: 56237606-56237606 on Build GRCh38
rs764587690 Chr.12: 56237768-56237768 on Build GRCh38
rs547030232 Chr.12: 56237544-56237544 on Build GRCh38
rs375990457 Chr.12: 56237729-56237729 on Build GRCh38
rs764748493 Chr.12: 56237882-56237882 on Build GRCh38
rs751714685 Chr.12: 56237603-56237603 on Build GRCh38
rs768227562 Chr.12: 56237581-56237581 on Build GRCh38
rs751871759 Chr.12: 56237688-56237688 on Build GRCh38
rs186869310 Chr.12: 56237779-56237779 on Build GRCh38
rs1006024560 Chr.12: 56237925-56237925 on Build GRCh38
rs770227481 Chr.12: 56237755-56237755 on Build GRCh38
rs774004961 Chr.12: 56237645-56237645 on Build GRCh38
rs758556800 Chr.12: 56237594-56237594 on Build GRCh38
rs771696569 Chr.12: 56237740-56237740 on Build GRCh38
rs529756871 Chr.12: 56237557-56237557 on Build GRCh38
rs950614978 Chr.12: 56237921-56237921 on Build GRCh38
rs543644121 Chr.12: 56237919-56237919 on Build GRCh38
rs377671437 Chr.12: 56237669-56237669 on Build GRCh38
rs774843204 Chr.12: 56237770-56237770 on Build GRCh38
rs769753149 Chr.12: 56237539-56237539 on Build GRCh38
rs756009566 Chr.12: 56237708-56237708 on Build GRCh38
rs527639814 Chr.12: 56237573-56237574 on Build GRCh38
rs376019241 Chr.12: 56237668-56237668 on Build GRCh38
rs909600979 Chr.12: 56237688-56237688 on Build GRCh38
rs1016116461 Chr.12: 56237949-56237949 on Build GRCh38
rs760757440 Chr.12: 56237782-56237782 on Build GRCh38
rs374112801 Chr.12: 56237591-56237591 on Build GRCh38
rs201848896 Chr.12: 56237632-56237632 on Build GRCh38
rs762141282 Chr.12: 56237773-56237773 on Build GRCh38
rs754850184 Chr.12: 56237714-56237714 on Build GRCh38
rs754388503 Chr.12: 56237572-56237572 on Build GRCh38
rs776118073 Chr.12: 56237756-56237756 on Build GRCh38
rs375366401 Chr.12: 56237772-56237772 on Build GRCh38
rs558867316 Chr.12: 56237662-56237662 on Build GRCh38
rs982022692 Chr.12: 56237667-56237667 on Build GRCh38
rs757550067 Chr.12: 56237692-56237692 on Build GRCh38
rs765582947 Chr.12: 56237585-56237585 on Build GRCh38
rs773737896 Chr.12: 56237556-56237556 on Build GRCh38
rs976425903 Chr.12: 56237588-56237588 on Build GRCh38
rs778644607 Chr.12: 56237728-56237728 on Build GRCh38
rs150715610 Chr.12: 56237713-56237713 on Build GRCh38
rs747773429 Chr.12: 56237649-56237649 on Build GRCh38
rs775351499 Chr.12: 56237540-56237540 on Build GRCh38
rs781780136 Chr.12: 56237626-56237626 on Build GRCh38
rs980283520 Chr.12: 56237973-56237973 on Build GRCh38
rs901196688 Chr.12: 56237885-56237885 on Build GRCh38
rs565229600 Chr.12: 56237920-56237920 on Build GRCh38

View Full Product Details