Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56242460-56242965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD52
Entrez Gene ID
283373
Gene Name
ankyrin repeat domain 52
Gene Aliases
ANKRD33
UniGene
Hs.524506
Gene Chromosome Location
Chr.12: 56237807-56258391 on Build GRCh38
Transcripts
NM_173595.3 XM_011538197.2 XM_017019183.1 XM_017019184.1

SNP Detail

SNP ID Chromosome Location
rs762757869 Chr.12: 56242587-56242587 on Build GRCh38
rs757092807 Chr.12: 56242866-56242866 on Build GRCh38
rs192439110 Chr.12: 56242627-56242627 on Build GRCh38
rs931345983 Chr.12: 56242691-56242693 on Build GRCh38
rs935125473 Chr.12: 56242817-56242817 on Build GRCh38
rs765916389 Chr.12: 56242605-56242606 on Build GRCh38
rs927827740 Chr.12: 56242546-56242546 on Build GRCh38
rs747254904 Chr.12: 56242639-56242639 on Build GRCh38
rs765460113 Chr.12: 56242828-56242828 on Build GRCh38
rs1052173492 Chr.12: 56242603-56242603 on Build GRCh38
rs916469357 Chr.12: 56242489-56242489 on Build GRCh38
rs1023408817 Chr.12: 56242770-56242770 on Build GRCh38
rs573697630 Chr.12: 56242697-56242697 on Build GRCh38
rs991903577 Chr.12: 56242484-56242484 on Build GRCh38
rs887719630 Chr.12: 56242713-56242713 on Build GRCh38
rs140847739 Chr.12: 56242634-56242634 on Build GRCh38
rs555940394 Chr.12: 56242727-56242727 on Build GRCh38
rs947981452 Chr.12: 56242496-56242496 on Build GRCh38
rs369860846 Chr.12: 56242893-56242893 on Build GRCh38
rs988328616 Chr.12: 56242612-56242612 on Build GRCh38
rs981254563 Chr.12: 56242945-56242948 on Build GRCh38
rs952237886 Chr.12: 56242737-56242737 on Build GRCh38
rs927796680 Chr.12: 56242807-56242807 on Build GRCh38
rs187113422 Chr.12: 56242611-56242611 on Build GRCh38
rs767217473 Chr.12: 56242634-56242634 on Build GRCh38
rs982027051 Chr.12: 56242798-56242798 on Build GRCh38
rs915026541 Chr.12: 56242864-56242864 on Build GRCh38
rs898031750 Chr.12: 56242712-56242712 on Build GRCh38
rs1019110760 Chr.12: 56242637-56242637 on Build GRCh38
rs759946915 Chr.12: 56242715-56242715 on Build GRCh38
rs966205346 Chr.12: 56242490-56242490 on Build GRCh38
rs772695652 Chr.12: 56242635-56242635 on Build GRCh38
rs537650116 Chr.12: 56242716-56242716 on Build GRCh38
rs893549109 Chr.12: 56242610-56242610 on Build GRCh38
rs981889671 Chr.12: 56242513-56242513 on Build GRCh38
rs901554624 Chr.12: 56242739-56242739 on Build GRCh38
rs935072945 Chr.12: 56242596-56242596 on Build GRCh38
rs975739989 Chr.12: 56242495-56242495 on Build GRCh38
rs953151453 Chr.12: 56242816-56242816 on Build GRCh38
rs969262178 Chr.12: 56242793-56242793 on Build GRCh38
rs566485589 Chr.12: 56242558-56242558 on Build GRCh38
rs991479751 Chr.12: 56242760-56242760 on Build GRCh38
rs987941274 Chr.12: 56242820-56242820 on Build GRCh38
rs888750729 Chr.12: 56242943-56242943 on Build GRCh38
rs918079605 Chr.12: 56242917-56242917 on Build GRCh38
rs1050114627 Chr.12: 56242938-56242938 on Build GRCh38

View Full Product Details