Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56282340-56282839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CS
Entrez Gene ID
1431
Gene Name
citrate synthase
Gene Aliases
-
UniGene
Hs.743252
Gene Chromosome Location
Chr.12: 56271699-56300391 on Build GRCh38
Transcripts
NM_004077.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
224433 c.548G>A p.R183Q Chr.12: 56282460-56282460 on Build GRCh38
6635925 c.417A>G p.K139K Chr.12: 56282591-56282591 on Build GRCh38
1179481 c.560A>G p.Q187R Chr.12: 56282448-56282448 on Build GRCh38
4150252 c.462G>C p.M154I Chr.12: 56282546-56282546 on Build GRCh38
4802239 c.443C>T p.P148L Chr.12: 56282565-56282565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200506886 Chr.12: 56282815-56282815 on Build GRCh38
rs748206006 Chr.12: 56282478-56282478 on Build GRCh38
rs772112453 Chr.12: 56282479-56282479 on Build GRCh38
rs947119608 Chr.12: 56282364-56282364 on Build GRCh38
rs987731750 Chr.12: 56282732-56282732 on Build GRCh38
rs969763586 Chr.12: 56282719-56282719 on Build GRCh38
rs1035975617 Chr.12: 56282760-56282760 on Build GRCh38
rs757844675 Chr.12: 56282526-56282526 on Build GRCh38
rs546393279 Chr.12: 56282567-56282567 on Build GRCh38
rs199775872 Chr.12: 56282629-56282629 on Build GRCh38
rs754488514 Chr.12: 56282402-56282402 on Build GRCh38
rs1041670338 Chr.12: 56282451-56282451 on Build GRCh38
rs1005574214 Chr.12: 56282704-56282704 on Build GRCh38
rs988683683 Chr.12: 56282809-56282809 on Build GRCh38
rs998748560 Chr.12: 56282587-56282587 on Build GRCh38
rs1047209721 Chr.12: 56282639-56282639 on Build GRCh38
rs766111390 Chr.12: 56282395-56282395 on Build GRCh38
rs77708038 Chr.12: 56282448-56282448 on Build GRCh38
rs903081426 Chr.12: 56282548-56282548 on Build GRCh38
rs764848639 Chr.12: 56282514-56282514 on Build GRCh38
rs910879905 Chr.12: 56282432-56282432 on Build GRCh38
rs994475802 Chr.12: 56282718-56282718 on Build GRCh38
rs756424958 Chr.12: 56282455-56282455 on Build GRCh38
rs529056420 Chr.12: 56282621-56282621 on Build GRCh38
rs750840615 Chr.12: 56282552-56282552 on Build GRCh38
rs771987015 Chr.12: 56282633-56282633 on Build GRCh38
rs959906916 Chr.12: 56282777-56282777 on Build GRCh38
rs764686679 Chr.12: 56282419-56282419 on Build GRCh38
rs777769764 Chr.12: 56282652-56282652 on Build GRCh38
rs184765034 Chr.12: 56282788-56282788 on Build GRCh38
rs1042401100 Chr.12: 56282485-56282485 on Build GRCh38
rs759141031 Chr.12: 56282512-56282512 on Build GRCh38
rs370300444 Chr.12: 56282461-56282461 on Build GRCh38
rs768131425 Chr.12: 56282529-56282529 on Build GRCh38
rs558935334 Chr.12: 56282714-56282714 on Build GRCh38
rs769624488 Chr.12: 56282735-56282735 on Build GRCh38
rs772093150 Chr.12: 56282811-56282811 on Build GRCh38
rs753429392 Chr.12: 56282396-56282396 on Build GRCh38
rs749458351 Chr.12: 56282457-56282457 on Build GRCh38
rs567874104 Chr.12: 56282615-56282615 on Build GRCh38
rs116165297 Chr.12: 56282460-56282460 on Build GRCh38
rs537033737 Chr.12: 56282698-56282698 on Build GRCh38
rs766020907 Chr.12: 56282502-56282502 on Build GRCh38
rs878959925 Chr.12: 56282630-56282630 on Build GRCh38
rs188095046 Chr.12: 56282683-56282683 on Build GRCh38
rs757811179 Chr.12: 56282436-56282436 on Build GRCh38
rs199624776 Chr.12: 56282406-56282406 on Build GRCh38
rs746313850 Chr.12: 56282449-56282449 on Build GRCh38
rs780339659 Chr.12: 56282456-56282456 on Build GRCh38
rs1001475440 Chr.12: 56282725-56282725 on Build GRCh38
rs780512368 Chr.12: 56282571-56282571 on Build GRCh38
rs752245414 Chr.12: 56282515-56282515 on Build GRCh38
rs181980994 Chr.12: 56282697-56282697 on Build GRCh38
rs779271380 Chr.12: 56282627-56282627 on Build GRCh38
rs773009426 Chr.12: 56282481-56282481 on Build GRCh38
rs528162217 Chr.12: 56282495-56282495 on Build GRCh38
rs201525280 Chr.12: 56282430-56282430 on Build GRCh38
rs1026003197 Chr.12: 56282730-56282730 on Build GRCh38

View Full Product Details