Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 61708169-61708678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A2
Entrez Gene ID
338811
Gene Name
family with sequence similarity 19 member A2, C-C motif chemokine like
Gene Aliases
TAFA-2, TAFA2
UniGene
Hs.269745
Gene Chromosome Location
Chr.12: 61708248-62192839 on Build GRCh38
Transcripts
NM_178539.4 XM_006719370.2 XM_011538272.1 XM_011538274.1

SNP Detail

SNP ID Chromosome Location
rs966079304 Chr.12: 61708589-61708589 on Build GRCh38
rs116335576 Chr.12: 61708269-61708269 on Build GRCh38
rs1055550198 Chr.12: 61708581-61708582 on Build GRCh38
rs1056314282 Chr.12: 61708462-61708462 on Build GRCh38
rs751379764 Chr.12: 61708469-61708469 on Build GRCh38
rs553662335 Chr.12: 61708402-61708402 on Build GRCh38
rs903963403 Chr.12: 61708491-61708491 on Build GRCh38
rs1043820704 Chr.12: 61708456-61708456 on Build GRCh38
rs1027474607 Chr.12: 61708502-61708502 on Build GRCh38
rs1012026886 Chr.12: 61708501-61708501 on Build GRCh38
rs553663834 Chr.12: 61708630-61708631 on Build GRCh38
rs758568796 Chr.12: 61708299-61708299 on Build GRCh38
rs887841672 Chr.12: 61708513-61708513 on Build GRCh38
rs986222903 Chr.12: 61708365-61708366 on Build GRCh38
rs1033917814 Chr.12: 61708249-61708249 on Build GRCh38
rs946514650 Chr.12: 61708348-61708348 on Build GRCh38
rs939232722 Chr.12: 61708373-61708373 on Build GRCh38
rs779999822 Chr.12: 61708304-61708304 on Build GRCh38
rs547284787 Chr.12: 61708618-61708618 on Build GRCh38
rs945226551 Chr.12: 61708440-61708440 on Build GRCh38
rs938004941 Chr.12: 61708515-61708515 on Build GRCh38
rs747048496 Chr.12: 61708361-61708361 on Build GRCh38
rs920376221 Chr.12: 61708326-61708326 on Build GRCh38
rs1002645952 Chr.12: 61708238-61708238 on Build GRCh38
rs573275652 Chr.12: 61708467-61708467 on Build GRCh38
rs540176363 Chr.12: 61708377-61708377 on Build GRCh38
rs1043873772 Chr.12: 61708229-61708229 on Build GRCh38
rs913739212 Chr.12: 61708387-61708387 on Build GRCh38
rs895108761 Chr.12: 61708483-61708483 on Build GRCh38
rs971782957 Chr.12: 61708322-61708322 on Build GRCh38
rs932141138 Chr.12: 61708271-61708271 on Build GRCh38
rs910445490 Chr.12: 61708366-61708366 on Build GRCh38
rs914984110 Chr.12: 61708341-61708341 on Build GRCh38
rs976373180 Chr.12: 61708255-61708255 on Build GRCh38
rs976292102 Chr.12: 61708592-61708592 on Build GRCh38
rs1014950714 Chr.12: 61708208-61708208 on Build GRCh38
rs1029243994 Chr.12: 61708607-61708607 on Build GRCh38
rs1013816165 Chr.12: 61708442-61708442 on Build GRCh38
rs557296723 Chr.12: 61708203-61708203 on Build GRCh38
rs966672196 Chr.12: 61708233-61708233 on Build GRCh38
rs896703060 Chr.12: 61708582-61708582 on Build GRCh38
rs542215866 Chr.12: 61708619-61708619 on Build GRCh38
rs1004985816 Chr.12: 61708566-61708566 on Build GRCh38
rs781757251 Chr.12: 61708644-61708644 on Build GRCh38
rs866586034 Chr.12: 61708366-61708366 on Build GRCh38
rs1013727374 Chr.12: 61708288-61708288 on Build GRCh38
rs755561086 Chr.12: 61708582-61708582 on Build GRCh38

View Full Product Details