Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 62191123-62191628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A2
Entrez Gene ID
338811
Gene Name
family with sequence similarity 19 member A2, C-C motif chemokine like
Gene Aliases
TAFA-2, TAFA2
UniGene
Hs.269745
Gene Chromosome Location
Chr.12: 61708248-62192839 on Build GRCh38
Transcripts
NM_178539.4 XM_006719370.2 XM_011538272.1 XM_011538274.1

SNP Detail

SNP ID Chromosome Location
rs532934372 Chr.12: 62191207-62191207 on Build GRCh38
rs1050844651 Chr.12: 62191171-62191171 on Build GRCh38
rs896163506 Chr.12: 62191457-62191457 on Build GRCh38
rs943534403 Chr.12: 62191429-62191432 on Build GRCh38
rs909004430 Chr.12: 62191415-62191415 on Build GRCh38
rs1012741147 Chr.12: 62191600-62191600 on Build GRCh38
rs986191066 Chr.12: 62191322-62191322 on Build GRCh38
rs1043640192 Chr.12: 62191193-62191193 on Build GRCh38
rs942590896 Chr.12: 62191178-62191178 on Build GRCh38
rs997460696 Chr.12: 62191202-62191202 on Build GRCh38
rs561877802 Chr.12: 62191156-62191156 on Build GRCh38
rs904967793 Chr.12: 62191588-62191588 on Build GRCh38
rs746684543 Chr.12: 62191465-62191466 on Build GRCh38
rs372854834 Chr.12: 62191170-62191170 on Build GRCh38
rs781488586 Chr.12: 62191179-62191179 on Build GRCh38
rs766651393 Chr.12: 62191214-62191214 on Build GRCh38
rs1013314380 Chr.12: 62191469-62191469 on Build GRCh38
rs923298912 Chr.12: 62191295-62191295 on Build GRCh38
rs756136838 Chr.12: 62191388-62191388 on Build GRCh38
rs999582478 Chr.12: 62191201-62191201 on Build GRCh38
rs546327889 Chr.12: 62191302-62191302 on Build GRCh38
rs936687134 Chr.12: 62191519-62191519 on Build GRCh38
rs902477922 Chr.12: 62191517-62191517 on Build GRCh38
rs1006455466 Chr.12: 62191246-62191246 on Build GRCh38
rs984910884 Chr.12: 62191393-62191393 on Build GRCh38
rs569130840 Chr.12: 62191459-62191459 on Build GRCh38
rs569856081 Chr.12: 62191319-62191319 on Build GRCh38
rs1054276584 Chr.12: 62191557-62191557 on Build GRCh38
rs1001328670 Chr.12: 62191272-62191272 on Build GRCh38
rs981084263 Chr.12: 62191333-62191333 on Build GRCh38
rs1052493590 Chr.12: 62191423-62191423 on Build GRCh38
rs189349227 Chr.12: 62191379-62191379 on Build GRCh38
rs530591055 Chr.12: 62191166-62191166 on Build GRCh38
rs971331016 Chr.12: 62191326-62191326 on Build GRCh38
rs80246517 Chr.12: 62191176-62191176 on Build GRCh38
rs538245208 Chr.12: 62191590-62191590 on Build GRCh38
rs923507858 Chr.12: 62191466-62191466 on Build GRCh38
rs1042807270 Chr.12: 62191208-62191208 on Build GRCh38
rs555507875 Chr.12: 62191447-62191447 on Build GRCh38
rs962350816 Chr.12: 62191276-62191276 on Build GRCh38
rs1023776220 Chr.12: 62191234-62191234 on Build GRCh38
rs1044856392 Chr.12: 62191546-62191546 on Build GRCh38
rs918998270 Chr.12: 62191164-62191164 on Build GRCh38
rs969558465 Chr.12: 62191241-62191241 on Build GRCh38
rs954923787 Chr.12: 62191315-62191315 on Build GRCh38
rs777879204 Chr.12: 62191445-62191445 on Build GRCh38
rs911029700 Chr.12: 62191341-62191341 on Build GRCh38
rs756257771 Chr.12: 62191147-62191147 on Build GRCh38
rs902139571 Chr.12: 62191189-62191189 on Build GRCh38
rs567108781 Chr.12: 62191345-62191345 on Build GRCh38
rs935304485 Chr.12: 62191398-62191398 on Build GRCh38
rs972471604 Chr.12: 62191282-62191282 on Build GRCh38
rs896224414 Chr.12: 62191227-62191227 on Build GRCh38
rs903790339 Chr.12: 62191199-62191199 on Build GRCh38
rs972113813 Chr.12: 62191150-62191150 on Build GRCh38
rs2279563 Chr.12: 62191145-62191145 on Build GRCh38
rs971048534 Chr.12: 62191143-62191143 on Build GRCh38
rs1016506758 Chr.12: 62191268-62191268 on Build GRCh38
rs1013345143 Chr.12: 62191233-62191233 on Build GRCh38

View Full Product Details