Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 68248319-68248864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL22
Entrez Gene ID
50616
Gene Name
interleukin 22
Gene Aliases
IL-21, IL-22, IL-D110, IL-TIF, ILTIF, TIFIL-23, TIFa, zcyto18
UniGene
Hs.287369
Gene Chromosome Location
Chr.12: 68248245-68253501 on Build GRCh38
Transcripts
NM_020525.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161717 c.480G>C p.E160D Chr.12: 68248859-68248859 on Build GRCh38
5428806 c.499C>G p.L167V Chr.12: 68248840-68248840 on Build GRCh38
1677160 c.487G>T p.A163S Chr.12: 68248852-68248852 on Build GRCh38
1177245 c.496G>T p.E166* Chr.12: 68248843-68248843 on Build GRCh38
3812869 c.520C>G p.L174V Chr.12: 68248819-68248819 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752259949 Chr.12: 68248823-68248823 on Build GRCh38
rs772158791 Chr.12: 68248772-68248772 on Build GRCh38
rs576389057 Chr.12: 68248652-68248652 on Build GRCh38
rs1013781048 Chr.12: 68248600-68248600 on Build GRCh38
rs868357355 Chr.12: 68248604-68248604 on Build GRCh38
rs1043798784 Chr.12: 68248719-68248719 on Build GRCh38
rs892526651 Chr.12: 68248654-68248654 on Build GRCh38
rs868133096 Chr.12: 68248420-68248420 on Build GRCh38
rs923122655 Chr.12: 68248716-68248717 on Build GRCh38
rs897959591 Chr.12: 68248547-68248547 on Build GRCh38
rs184538934 Chr.12: 68248653-68248653 on Build GRCh38
rs1032266706 Chr.12: 68248674-68248674 on Build GRCh38
rs762305757 Chr.12: 68248786-68248786 on Build GRCh38
rs933441215 Chr.12: 68248468-68248469 on Build GRCh38
rs147812317 Chr.12: 68248825-68248825 on Build GRCh38
rs916975487 Chr.12: 68248575-68248575 on Build GRCh38
rs1050432679 Chr.12: 68248468-68248468 on Build GRCh38
rs1039600353 Chr.12: 68248625-68248625 on Build GRCh38
rs763683281 Chr.12: 68248826-68248826 on Build GRCh38
rs992543564 Chr.12: 68248558-68248558 on Build GRCh38
rs889146750 Chr.12: 68248504-68248504 on Build GRCh38
rs566349084 Chr.12: 68248402-68248406 on Build GRCh38
rs925081954 Chr.12: 68248400-68248400 on Build GRCh38
rs921228640 Chr.12: 68248630-68248630 on Build GRCh38
rs369510786 Chr.12: 68248764-68248764 on Build GRCh38
rs923371164 Chr.12: 68248411-68248411 on Build GRCh38
rs1009581952 Chr.12: 68248688-68248688 on Build GRCh38
rs771827145 Chr.12: 68248797-68248797 on Build GRCh38
rs909687030 Chr.12: 68248614-68248614 on Build GRCh38
rs138303944 Chr.12: 68248726-68248726 on Build GRCh38
rs1002377421 Chr.12: 68248514-68248514 on Build GRCh38
rs1022441682 Chr.12: 68248395-68248395 on Build GRCh38
rs753697592 Chr.12: 68248750-68248750 on Build GRCh38
rs188640573 Chr.12: 68248727-68248727 on Build GRCh38
rs541587485 Chr.12: 68248735-68248735 on Build GRCh38
rs373532288 Chr.12: 68248766-68248766 on Build GRCh38
rs933408928 Chr.12: 68248717-68248717 on Build GRCh38
rs376750729 Chr.12: 68248775-68248775 on Build GRCh38
rs1033938926 Chr.12: 68248520-68248520 on Build GRCh38
rs748766808 Chr.12: 68248549-68248549 on Build GRCh38
rs767038089 Chr.12: 68248817-68248817 on Build GRCh38
rs935152984 Chr.12: 68248401-68248402 on Build GRCh38
rs1009678168 Chr.12: 68248394-68248394 on Build GRCh38
rs143690611 Chr.12: 68248399-68248399 on Build GRCh38
rs780200539 Chr.12: 68248773-68248773 on Build GRCh38
rs777182679 Chr.12: 68248783-68248783 on Build GRCh38
rs755714425 Chr.12: 68248497-68248497 on Build GRCh38
rs867565387 Chr.12: 68248590-68248590 on Build GRCh38
rs879770639 Chr.12: 68248364-68248364 on Build GRCh38
rs956641531 Chr.12: 68248684-68248684 on Build GRCh38
rs192648178 Chr.12: 68248608-68248608 on Build GRCh38
rs536024095 Chr.12: 68248343-68248343 on Build GRCh38
rs199546412 Chr.12: 68248829-68248829 on Build GRCh38
rs941180526 Chr.12: 68248617-68248617 on Build GRCh38
rs188919450 Chr.12: 68248544-68248544 on Build GRCh38
rs369755821 Chr.12: 68248788-68248788 on Build GRCh38
rs769167674 Chr.12: 68248780-68248780 on Build GRCh38
rs1051130234 Chr.12: 68248637-68248637 on Build GRCh38
rs773429042 Chr.12: 68248792-68248792 on Build GRCh38
rs757218231 Chr.12: 68248632-68248632 on Build GRCh38
rs534093662 Chr.12: 68248548-68248548 on Build GRCh38

View Full Product Details