Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 68248735-68249236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL22
Entrez Gene ID
50616
Gene Name
interleukin 22
Gene Aliases
IL-21, IL-22, IL-D110, IL-TIF, ILTIF, TIFIL-23, TIFa, zcyto18
UniGene
Hs.287369
Gene Chromosome Location
Chr.12: 68248245-68253501 on Build GRCh38
Transcripts
NM_020525.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161717 c.480G>C p.E160D Chr.12: 68248859-68248859 on Build GRCh38
5428806 c.499C>G p.L167V Chr.12: 68248840-68248840 on Build GRCh38
1677160 c.487G>T p.A163S Chr.12: 68248852-68248852 on Build GRCh38
7372061 c.472A>G p.S158G Chr.12: 68248867-68248867 on Build GRCh38
1177245 c.496G>T p.E166* Chr.12: 68248843-68248843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772158791 Chr.12: 68248772-68248772 on Build GRCh38
rs752259949 Chr.12: 68248823-68248823 on Build GRCh38
rs748307855 Chr.12: 68248885-68248885 on Build GRCh38
rs746304596 Chr.12: 68248977-68248977 on Build GRCh38
rs945371405 Chr.12: 68249137-68249137 on Build GRCh38
rs1016683181 Chr.12: 68249058-68249058 on Build GRCh38
rs552006636 Chr.12: 68249054-68249054 on Build GRCh38
rs892140109 Chr.12: 68249139-68249139 on Build GRCh38
rs2227507 Chr.12: 68248867-68248867 on Build GRCh38
rs537415914 Chr.12: 68249152-68249152 on Build GRCh38
rs768793594 Chr.12: 68248872-68248872 on Build GRCh38
rs955396563 Chr.12: 68249127-68249127 on Build GRCh38
rs762305757 Chr.12: 68248786-68248786 on Build GRCh38
rs147812317 Chr.12: 68248825-68248825 on Build GRCh38
rs763683281 Chr.12: 68248826-68248826 on Build GRCh38
rs1013810331 Chr.12: 68249032-68249032 on Build GRCh38
rs1017432154 Chr.12: 68249205-68249205 on Build GRCh38
rs753373432 Chr.12: 68248919-68248919 on Build GRCh38
rs933947375 Chr.12: 68248866-68248866 on Build GRCh38
rs369510786 Chr.12: 68248764-68248764 on Build GRCh38
rs975632827 Chr.12: 68249083-68249083 on Build GRCh38
rs1055323575 Chr.12: 68249024-68249024 on Build GRCh38
rs956593265 Chr.12: 68248924-68248924 on Build GRCh38
rs917385725 Chr.12: 68249067-68249067 on Build GRCh38
rs546436547 Chr.12: 68248865-68248865 on Build GRCh38
rs760397239 Chr.12: 68248910-68248910 on Build GRCh38
rs550212092 Chr.12: 68248909-68248909 on Build GRCh38
rs547625379 Chr.12: 68249028-68249028 on Build GRCh38
rs771827145 Chr.12: 68248797-68248797 on Build GRCh38
rs769998678 Chr.12: 68248954-68248954 on Build GRCh38
rs1044618198 Chr.12: 68249093-68249093 on Build GRCh38
rs938171204 Chr.12: 68249022-68249022 on Build GRCh38
rs954723080 Chr.12: 68249021-68249021 on Build GRCh38
rs766365368 Chr.12: 68248894-68248894 on Build GRCh38
rs768868761 Chr.12: 68248894-68248895 on Build GRCh38
rs373532288 Chr.12: 68248766-68248766 on Build GRCh38
rs925354840 Chr.12: 68249159-68249159 on Build GRCh38
rs376750729 Chr.12: 68248775-68248775 on Build GRCh38
rs1030138817 Chr.12: 68249011-68249011 on Build GRCh38
rs781221249 Chr.12: 68248879-68248879 on Build GRCh38
rs184366105 Chr.12: 68249026-68249026 on Build GRCh38
rs143785595 Chr.12: 68249135-68249135 on Build GRCh38
rs2227506 Chr.12: 68249053-68249053 on Build GRCh38
rs767038089 Chr.12: 68248817-68248817 on Build GRCh38
rs191823946 Chr.12: 68248899-68248899 on Build GRCh38
rs938088450 Chr.12: 68249201-68249201 on Build GRCh38
rs535909820 Chr.12: 68248958-68248958 on Build GRCh38
rs780200539 Chr.12: 68248773-68248773 on Build GRCh38
rs962481713 Chr.12: 68249076-68249076 on Build GRCh38
rs904939951 Chr.12: 68249100-68249100 on Build GRCh38
rs903980938 Chr.12: 68248968-68248968 on Build GRCh38
rs374915113 Chr.12: 68248890-68248890 on Build GRCh38
rs1028132510 Chr.12: 68249121-68249121 on Build GRCh38
rs777182679 Chr.12: 68248783-68248783 on Build GRCh38
rs141353897 Chr.12: 68248852-68248852 on Build GRCh38
rs189165185 Chr.12: 68249037-68249037 on Build GRCh38
rs973356358 Chr.12: 68249034-68249034 on Build GRCh38
rs781766741 Chr.12: 68248976-68248976 on Build GRCh38
rs773656792 Chr.12: 68248897-68248897 on Build GRCh38
rs199546412 Chr.12: 68248829-68248829 on Build GRCh38
rs747223387 Chr.12: 68248868-68248868 on Build GRCh38
rs562101400 Chr.12: 68248927-68248927 on Build GRCh38
rs372203467 Chr.12: 68249131-68249131 on Build GRCh38
rs369755821 Chr.12: 68248788-68248788 on Build GRCh38
rs769167674 Chr.12: 68248780-68248780 on Build GRCh38
rs1009463265 Chr.12: 68249146-68249146 on Build GRCh38
rs773429042 Chr.12: 68248792-68248792 on Build GRCh38
rs771768909 Chr.12: 68248906-68248906 on Build GRCh38

View Full Product Details