Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69587387-69587897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT2
Entrez Gene ID
10576
Gene Name
chaperonin containing TCP1 subunit 2
Gene Aliases
99D8.1, CCT-beta, CCTB, HEL-S-100n, PRO1633, TCP-1-beta
UniGene
Hs.189772
Gene Chromosome Location
Chr.12: 69585428-69601577 on Build GRCh38
Transcripts
NM_001198842.1 NM_006431.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223566 c.172G>A p.D58N Chr.12: 69587532-69587532 on Build GRCh38
88518 c.172G>T p.D58Y Chr.12: 69587532-69587532 on Build GRCh38
1747218 c.256G>A p.D86N Chr.12: 69587616-69587616 on Build GRCh38
6623279 c.212_213insA p.N73fs*5 Chr.12: 69587572-69587573 on Build GRCh38
6290048 c.204C>T p.A68A Chr.12: 69587564-69587564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751957379 Chr.12: 69587651-69587652 on Build GRCh38
rs1016885653 Chr.12: 69587720-69587720 on Build GRCh38
rs1043319441 Chr.12: 69587426-69587427 on Build GRCh38
rs777399903 Chr.12: 69587517-69587517 on Build GRCh38
rs7136893 Chr.12: 69587706-69587706 on Build GRCh38
rs112593509 Chr.12: 69587617-69587617 on Build GRCh38
rs765322805 Chr.12: 69587571-69587571 on Build GRCh38
rs200570835 Chr.12: 69587593-69587593 on Build GRCh38
rs369156960 Chr.12: 69587496-69587496 on Build GRCh38
rs756967610 Chr.12: 69587473-69587473 on Build GRCh38
rs763244265 Chr.12: 69587520-69587522 on Build GRCh38
rs748553470 Chr.12: 69587740-69587740 on Build GRCh38
rs755822333 Chr.12: 69587497-69587497 on Build GRCh38
rs755270638 Chr.12: 69587603-69587603 on Build GRCh38
rs541090058 Chr.12: 69587839-69587839 on Build GRCh38
rs531299887 Chr.12: 69587773-69587773 on Build GRCh38
rs546551565 Chr.12: 69587772-69587772 on Build GRCh38
rs945103794 Chr.12: 69587795-69587795 on Build GRCh38
rs754036102 Chr.12: 69587463-69587463 on Build GRCh38
rs1002412664 Chr.12: 69587545-69587545 on Build GRCh38
rs775479503 Chr.12: 69587542-69587542 on Build GRCh38
rs1040690949 Chr.12: 69587514-69587514 on Build GRCh38
rs528348784 Chr.12: 69587544-69587544 on Build GRCh38
rs376214452 Chr.12: 69587645-69587645 on Build GRCh38
rs904126237 Chr.12: 69587840-69587840 on Build GRCh38
rs746810317 Chr.12: 69587659-69587659 on Build GRCh38
rs146023119 Chr.12: 69587539-69587539 on Build GRCh38
rs932374079 Chr.12: 69587854-69587854 on Build GRCh38
rs757336726 Chr.12: 69587468-69587468 on Build GRCh38
rs947614800 Chr.12: 69587755-69587755 on Build GRCh38
rs777424153 Chr.12: 69587607-69587607 on Build GRCh38
rs892108822 Chr.12: 69587480-69587480 on Build GRCh38
rs916052614 Chr.12: 69587754-69587754 on Build GRCh38
rs986515646 Chr.12: 69587789-69587789 on Build GRCh38
rs778745250 Chr.12: 69587530-69587530 on Build GRCh38
rs777027963 Chr.12: 69587666-69587666 on Build GRCh38
rs753447346 Chr.12: 69587612-69587612 on Build GRCh38
rs1005458511 Chr.12: 69587577-69587577 on Build GRCh38
rs777225038 Chr.12: 69587739-69587739 on Build GRCh38
rs1050768458 Chr.12: 69587477-69587477 on Build GRCh38
rs950679673 Chr.12: 69587707-69587707 on Build GRCh38
rs750443373 Chr.12: 69587575-69587575 on Build GRCh38
rs913574085 Chr.12: 69587790-69587790 on Build GRCh38
rs776932308 Chr.12: 69587547-69587547 on Build GRCh38
rs1040659214 Chr.12: 69587806-69587806 on Build GRCh38
rs758725500 Chr.12: 69587489-69587489 on Build GRCh38
rs780068015 Chr.12: 69587651-69587651 on Build GRCh38
rs534714078 Chr.12: 69587730-69587730 on Build GRCh38
rs1013593916 Chr.12: 69587687-69587687 on Build GRCh38
rs750370364 Chr.12: 69587743-69587743 on Build GRCh38
rs755877364 Chr.12: 69587652-69587653 on Build GRCh38
rs761649987 Chr.12: 69587524-69587524 on Build GRCh38
rs924827288 Chr.12: 69587771-69587771 on Build GRCh38
rs756827568 Chr.12: 69587627-69587627 on Build GRCh38
rs772110300 Chr.12: 69587650-69587650 on Build GRCh38
rs978980988 Chr.12: 69587765-69587765 on Build GRCh38
rs766702369 Chr.12: 69587646-69587648 on Build GRCh38
rs763567991 Chr.12: 69587580-69587580 on Build GRCh38
rs764393384 Chr.12: 69587456-69587456 on Build GRCh38
rs539374680 Chr.12: 69587851-69587851 on Build GRCh38
rs769084610 Chr.12: 69587716-69587716 on Build GRCh38
rs770424246 Chr.12: 69587528-69587528 on Build GRCh38
rs896590684 Chr.12: 69587564-69587564 on Build GRCh38
rs77151347 Chr.12: 69587486-69587486 on Build GRCh38
rs1043777952 Chr.12: 69587796-69587796 on Build GRCh38
rs750895311 Chr.12: 69587487-69587487 on Build GRCh38
rs780624649 Chr.12: 69587554-69587554 on Build GRCh38
rs749082867 Chr.12: 69587449-69587449 on Build GRCh38
rs182288953 Chr.12: 69587600-69587600 on Build GRCh38
rs778533150 Chr.12: 69587644-69587644 on Build GRCh38
rs996621911 Chr.12: 69587567-69587567 on Build GRCh38
rs760538203 Chr.12: 69587543-69587543 on Build GRCh38
rs938235974 Chr.12: 69587843-69587843 on Build GRCh38

View Full Product Details