Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91057659-91058154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KERA
Entrez Gene ID
11081
Gene Name
keratocan
Gene Aliases
CNA2, KTN, SLRR2B
UniGene
Hs.125750
Gene Chromosome Location
Chr.12: 91050491-91058354 on Build GRCh38
Transcripts
NM_007035.3

SNP Detail

SNP ID Chromosome Location
rs574955423 Chr.12: 91057786-91057787 on Build GRCh38
rs932126300 Chr.12: 91058061-91058061 on Build GRCh38
rs1012405885 Chr.12: 91057971-91057971 on Build GRCh38
rs372259494 Chr.12: 91057898-91057898 on Build GRCh38
rs920571698 Chr.12: 91058046-91058046 on Build GRCh38
rs922027402 Chr.12: 91057784-91057784 on Build GRCh38
rs187535489 Chr.12: 91057913-91057913 on Build GRCh38
rs1990550 Chr.12: 91057866-91057866 on Build GRCh38
rs1018835400 Chr.12: 91057687-91057687 on Build GRCh38
rs905652168 Chr.12: 91057896-91057896 on Build GRCh38
rs886287368 Chr.12: 91057975-91057975 on Build GRCh38
rs923996189 Chr.12: 91058036-91058036 on Build GRCh38
rs61184438 Chr.12: 91057987-91057987 on Build GRCh38
rs982352625 Chr.12: 91057752-91057752 on Build GRCh38
rs779861921 Chr.12: 91057983-91057983 on Build GRCh38
rs949762592 Chr.12: 91057892-91057892 on Build GRCh38
rs528418089 Chr.12: 91057942-91057942 on Build GRCh38
rs918150965 Chr.12: 91057887-91057887 on Build GRCh38
rs968096261 Chr.12: 91057797-91057797 on Build GRCh38
rs752451886 Chr.12: 91057684-91057684 on Build GRCh38
rs902399932 Chr.12: 91057879-91057879 on Build GRCh38
rs996613185 Chr.12: 91058008-91058008 on Build GRCh38
rs943876725 Chr.12: 91058106-91058106 on Build GRCh38
rs796714356 Chr.12: 91057823-91057823 on Build GRCh38
rs185783003 Chr.12: 91057843-91057843 on Build GRCh38
rs746712317 Chr.12: 91058004-91058004 on Build GRCh38
rs918199469 Chr.12: 91058074-91058074 on Build GRCh38
rs528434972 Chr.12: 91058105-91058105 on Build GRCh38
rs1044061937 Chr.12: 91057893-91057893 on Build GRCh38
rs932044139 Chr.12: 91057860-91057860 on Build GRCh38
rs1048954069 Chr.12: 91057965-91057965 on Build GRCh38
rs908090191 Chr.12: 91057764-91057764 on Build GRCh38
rs986173113 Chr.12: 91058095-91058095 on Build GRCh38
rs574127423 Chr.12: 91057710-91057710 on Build GRCh38
rs1040832717 Chr.12: 91058112-91058112 on Build GRCh38
rs754670133 Chr.12: 91058064-91058064 on Build GRCh38
rs570174903 Chr.12: 91058039-91058039 on Build GRCh38
rs911945019 Chr.12: 91058098-91058098 on Build GRCh38
rs979790953 Chr.12: 91058111-91058111 on Build GRCh38
rs982236951 Chr.12: 91058027-91058027 on Build GRCh38
rs57895264 Chr.12: 91057813-91057813 on Build GRCh38
rs34630139 Chr.12: 91057866-91057867 on Build GRCh38
rs957110186 Chr.12: 91058043-91058043 on Build GRCh38
rs879356207 Chr.12: 91057866-91057867 on Build GRCh38
rs192780633 Chr.12: 91058021-91058021 on Build GRCh38
rs979608798 Chr.12: 91057801-91057801 on Build GRCh38
rs2735332 Chr.12: 91057812-91057812 on Build GRCh38
rs549907410 Chr.12: 91058035-91058035 on Build GRCh38
rs1990549 Chr.12: 91057867-91057867 on Build GRCh38
rs545987551 Chr.12: 91057870-91057870 on Build GRCh38
rs974731615 Chr.12: 91057765-91057765 on Build GRCh38
rs992298390 Chr.12: 91058050-91058050 on Build GRCh38
rs1049607267 Chr.12: 91057863-91057863 on Build GRCh38
rs948329789 Chr.12: 91058110-91058110 on Build GRCh38
rs968126849 Chr.12: 91058033-91058033 on Build GRCh38
rs2735330 Chr.12: 91057872-91057872 on Build GRCh38
rs542111794 Chr.12: 91057943-91057943 on Build GRCh38

View Full Product Details