Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95960256-95960790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMDHD1
Entrez Gene ID
144193
Gene Name
amidohydrolase domain containing 1
Gene Aliases
HMFT1272
UniGene
Hs.424907
Gene Chromosome Location
Chr.12: 95943293-95968592 on Build GRCh38
Transcripts
NM_152435.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6520569 c.778C>A p.H260N Chr.12: 95960588-95960588 on Build GRCh38
944712 c.669A>G p.E223E Chr.12: 95960479-95960479 on Build GRCh38
2046465 c.691G>A p.V231I Chr.12: 95960501-95960501 on Build GRCh38
944713 c.754G>T p.D252Y Chr.12: 95960564-95960564 on Build GRCh38
1299981 c.621C>T p.I207I Chr.12: 95960431-95960431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779079130 Chr.12: 95960450-95960450 on Build GRCh38
rs757012045 Chr.12: 95960648-95960648 on Build GRCh38
rs922295494 Chr.12: 95960710-95960710 on Build GRCh38
rs770427304 Chr.12: 95960481-95960481 on Build GRCh38
rs745424544 Chr.12: 95960564-95960564 on Build GRCh38
rs759966715 Chr.12: 95960378-95960378 on Build GRCh38
rs759030798 Chr.12: 95960614-95960614 on Build GRCh38
rs765552693 Chr.12: 95960519-95960519 on Build GRCh38
rs867963427 Chr.12: 95960477-95960477 on Build GRCh38
rs926458953 Chr.12: 95960441-95960441 on Build GRCh38
rs369624479 Chr.12: 95960760-95960760 on Build GRCh38
rs143136011 Chr.12: 95960458-95960458 on Build GRCh38
rs184499535 Chr.12: 95960486-95960486 on Build GRCh38
rs969843167 Chr.12: 95960306-95960306 on Build GRCh38
rs1049519837 Chr.12: 95960750-95960750 on Build GRCh38
rs763146421 Chr.12: 95960620-95960620 on Build GRCh38
rs1001359829 Chr.12: 95960690-95960690 on Build GRCh38
rs956019941 Chr.12: 95960281-95960281 on Build GRCh38
rs367652571 Chr.12: 95960509-95960509 on Build GRCh38
rs989308877 Chr.12: 95960660-95960660 on Build GRCh38
rs375729822 Chr.12: 95960673-95960673 on Build GRCh38
rs777554650 Chr.12: 95960635-95960635 on Build GRCh38
rs138567370 Chr.12: 95960539-95960539 on Build GRCh38
rs565909257 Chr.12: 95960607-95960607 on Build GRCh38
rs759030748 Chr.12: 95960487-95960487 on Build GRCh38
rs372259162 Chr.12: 95960589-95960589 on Build GRCh38
rs551643279 Chr.12: 95960651-95960651 on Build GRCh38
rs772760819 Chr.12: 95960478-95960478 on Build GRCh38
rs768059078 Chr.12: 95960494-95960494 on Build GRCh38
rs376012943 Chr.12: 95960556-95960556 on Build GRCh38
rs373341997 Chr.12: 95960670-95960670 on Build GRCh38
rs545584707 Chr.12: 95960530-95960530 on Build GRCh38
rs113476184 Chr.12: 95960397-95960397 on Build GRCh38
rs769566351 Chr.12: 95960335-95960335 on Build GRCh38
rs762097837 Chr.12: 95960688-95960688 on Build GRCh38
rs779554858 Chr.12: 95960663-95960663 on Build GRCh38
rs753388239 Chr.12: 95960635-95960636 on Build GRCh38
rs980838325 Chr.12: 95960625-95960625 on Build GRCh38
rs1011293062 Chr.12: 95960286-95960286 on Build GRCh38
rs763627683 Chr.12: 95960474-95960474 on Build GRCh38
rs781307546 Chr.12: 95960531-95960531 on Build GRCh38
rs750632341 Chr.12: 95960615-95960615 on Build GRCh38
rs754025263 Chr.12: 95960517-95960517 on Build GRCh38
rs143223993 Chr.12: 95960501-95960501 on Build GRCh38
rs748912127 Chr.12: 95960480-95960480 on Build GRCh38
rs572290731 Chr.12: 95960373-95960373 on Build GRCh38
rs764265690 Chr.12: 95960438-95960438 on Build GRCh38
rs12369577 Chr.12: 95960650-95960650 on Build GRCh38
rs775947828 Chr.12: 95960496-95960496 on Build GRCh38
rs767138394 Chr.12: 95960637-95960637 on Build GRCh38
rs147106014 Chr.12: 95960528-95960528 on Build GRCh38
rs2043608 Chr.12: 95960677-95960677 on Build GRCh38
rs200703607 Chr.12: 95960601-95960601 on Build GRCh38
rs537041796 Chr.12: 95960684-95960684 on Build GRCh38
rs1035477862 Chr.12: 95960764-95960764 on Build GRCh38
rs756140324 Chr.12: 95960630-95960630 on Build GRCh38
rs757309974 Chr.12: 95960444-95960444 on Build GRCh38
rs761189940 Chr.12: 95960602-95960602 on Build GRCh38
rs772888600 Chr.12: 95960365-95960365 on Build GRCh38
rs1021881984 Chr.12: 95960305-95960305 on Build GRCh38
rs368544076 Chr.12: 95960568-95960568 on Build GRCh38
rs773040508 Chr.12: 95960443-95960443 on Build GRCh38
rs749029278 Chr.12: 95960545-95960545 on Build GRCh38
rs112246451 Chr.12: 95960631-95960631 on Build GRCh38
rs769590831 Chr.12: 95960474-95960474 on Build GRCh38
rs761291441 Chr.12: 95960670-95960670 on Build GRCh38
rs888185722 Chr.12: 95960752-95960752 on Build GRCh38
rs746949067 Chr.12: 95960454-95960454 on Build GRCh38
rs773894056 Chr.12: 95960592-95960592 on Build GRCh38
rs376460465 Chr.12: 95960608-95960608 on Build GRCh38
rs768906861 Chr.12: 95960603-95960603 on Build GRCh38
rs746497039 Chr.12: 95960667-95960667 on Build GRCh38
rs774847951 Chr.12: 95960375-95960375 on Build GRCh38
rs936484950 Chr.12: 95960447-95960447 on Build GRCh38
rs752581438 Chr.12: 95960628-95960628 on Build GRCh38
rs747931752 Chr.12: 95960469-95960469 on Build GRCh38
rs756481811 Chr.12: 95960424-95960424 on Build GRCh38
rs199577038 Chr.12: 95960555-95960555 on Build GRCh38
rs201755546 Chr.12: 95960429-95960429 on Build GRCh38
rs144127287 Chr.12: 95960544-95960544 on Build GRCh38
rs775887555 Chr.12: 95960569-95960569 on Build GRCh38
rs781264650 Chr.12: 95960460-95960460 on Build GRCh38
rs768004085 Chr.12: 95960389-95960389 on Build GRCh38
rs1038430859 Chr.12: 95960526-95960526 on Build GRCh38
rs560341531 Chr.12: 95960315-95960315 on Build GRCh38
rs932285114 Chr.12: 95960738-95960738 on Build GRCh38
rs147349485 Chr.12: 95960623-95960623 on Build GRCh38

View Full Product Details