Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6637215-6637719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC11
Entrez Gene ID
55735
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Gene Aliases
dJ126A5.1
UniGene
Hs.462640
Gene Chromosome Location
Chr.1: 6634168-6701906 on Build GRCh38
Transcripts
NM_018198.3 XM_017001745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2239872 c.1448T>A p.V483E Chr.1: 6637274-6637274 on Build GRCh38
6647187 c.1349G>A p.R450Q Chr.1: 6637479-6637479 on Build GRCh38
2239873 c.1440C>T p.S480S Chr.1: 6637282-6637282 on Build GRCh38
4009526 c.1396A>G p.N466D Chr.1: 6637326-6637326 on Build GRCh38
3966683 c.1390A>G p.I464V Chr.1: 6637332-6637332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759805759 Chr.1: 6637282-6637282 on Build GRCh38
rs763333816 Chr.1: 6637500-6637500 on Build GRCh38
rs915766785 Chr.1: 6637546-6637546 on Build GRCh38
rs761017526 Chr.1: 6637532-6637532 on Build GRCh38
rs756511722 Chr.1: 6637416-6637416 on Build GRCh38
rs887969778 Chr.1: 6637602-6637602 on Build GRCh38
rs535424787 Chr.1: 6637648-6637648 on Build GRCh38
rs773171401 Chr.1: 6637548-6637548 on Build GRCh38
rs916442712 Chr.1: 6637410-6637410 on Build GRCh38
rs747974583 Chr.1: 6637457-6637457 on Build GRCh38
rs762187544 Chr.1: 6637516-6637516 on Build GRCh38
rs527889556 Chr.1: 6637594-6637594 on Build GRCh38
rs767407742 Chr.1: 6637273-6637273 on Build GRCh38
rs546373070 Chr.1: 6637598-6637598 on Build GRCh38
rs770896848 Chr.1: 6637473-6637473 on Build GRCh38
rs901455760 Chr.1: 6637633-6637633 on Build GRCh38
rs766799308 Chr.1: 6637540-6637540 on Build GRCh38
rs146590118 Chr.1: 6637362-6637362 on Build GRCh38
rs753425246 Chr.1: 6637543-6637543 on Build GRCh38
rs780928418 Chr.1: 6637438-6637438 on Build GRCh38
rs772065181 Chr.1: 6637465-6637465 on Build GRCh38
rs765345795 Chr.1: 6637289-6637289 on Build GRCh38
rs768788746 Chr.1: 6637514-6637514 on Build GRCh38
rs935404368 Chr.1: 6637241-6637241 on Build GRCh38
rs199968865 Chr.1: 6637479-6637479 on Build GRCh38
rs978929072 Chr.1: 6637578-6637578 on Build GRCh38
rs752005155 Chr.1: 6637390-6637390 on Build GRCh38
rs764663149 Chr.1: 6637294-6637294 on Build GRCh38
rs756203458 Chr.1: 6637356-6637356 on Build GRCh38
rs560440198 Chr.1: 6637593-6637593 on Build GRCh38
rs369452881 Chr.1: 6637293-6637293 on Build GRCh38
rs763100403 Chr.1: 6637527-6637530 on Build GRCh38
rs768208104 Chr.1: 6637354-6637354 on Build GRCh38
rs753549668 Chr.1: 6637431-6637431 on Build GRCh38
rs774512991 Chr.1: 6637515-6637515 on Build GRCh38
rs767726774 Chr.1: 6637409-6637409 on Build GRCh38
rs144647823 Chr.1: 6637315-6637315 on Build GRCh38
rs919208368 Chr.1: 6637520-6637520 on Build GRCh38
rs200102617 Chr.1: 6637439-6637439 on Build GRCh38
rs923349898 Chr.1: 6637565-6637565 on Build GRCh38
rs757548391 Chr.1: 6637325-6637325 on Build GRCh38
rs778704509 Chr.1: 6637355-6637355 on Build GRCh38
rs545503599 Chr.1: 6637697-6637697 on Build GRCh38
rs202046613 Chr.1: 6637281-6637281 on Build GRCh38
rs768039589 Chr.1: 6637528-6637528 on Build GRCh38
rs573944451 Chr.1: 6637329-6637329 on Build GRCh38
rs889958762 Chr.1: 6637651-6637651 on Build GRCh38
rs761492954 Chr.1: 6637271-6637271 on Build GRCh38
rs759624916 Chr.1: 6637363-6637363 on Build GRCh38
rs781667741 Chr.1: 6637328-6637328 on Build GRCh38
rs750943024 Chr.1: 6637529-6637529 on Build GRCh38
rs779501058 Chr.1: 6637343-6637343 on Build GRCh38
rs779661036 Chr.1: 6637430-6637430 on Build GRCh38
rs925603292 Chr.1: 6637579-6637579 on Build GRCh38
rs771701102 Chr.1: 6637358-6637358 on Build GRCh38
rs942237468 Chr.1: 6637603-6637603 on Build GRCh38
rs1040870297 Chr.1: 6637307-6637307 on Build GRCh38
rs763101433 Chr.1: 6637376-6637376 on Build GRCh38
rs923959601 Chr.1: 6637235-6637235 on Build GRCh38
rs373179186 Chr.1: 6637345-6637345 on Build GRCh38
rs200927563 Chr.1: 6637470-6637470 on Build GRCh38
rs1006063102 Chr.1: 6637427-6637427 on Build GRCh38
rs976152052 Chr.1: 6637542-6637542 on Build GRCh38
rs752844444 Chr.1: 6637292-6637292 on Build GRCh38
rs770169117 Chr.1: 6637364-6637364 on Build GRCh38
rs750831694 Chr.1: 6637413-6637413 on Build GRCh38
rs148060356 Chr.1: 6637330-6637330 on Build GRCh38
rs568187852 Chr.1: 6637629-6637629 on Build GRCh38
rs138013045 Chr.1: 6637261-6637261 on Build GRCh38
rs959845488 Chr.1: 6637521-6637521 on Build GRCh38
rs775892167 Chr.1: 6637371-6637371 on Build GRCh38
rs562821219 Chr.1: 6637384-6637384 on Build GRCh38
rs762385218 Chr.1: 6637408-6637408 on Build GRCh38
rs767722137 Chr.1: 6637349-6637349 on Build GRCh38
rs896784868 Chr.1: 6637650-6637650 on Build GRCh38
rs866464257 Chr.1: 6637309-6637309 on Build GRCh38
rs994235388 Chr.1: 6637683-6637683 on Build GRCh38

View Full Product Details