Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8860908-8861405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO1
Entrez Gene ID
2023
Gene Name
enolase 1
Gene Aliases
ENO1L1, HEL-S-17, MPB1, NNE, PPH
UniGene
Hs.517145
Gene Chromosome Location
Chr.1: 8861000-8879092 on Build GRCh38
Transcripts
NM_001201483.1 NM_001428.3 XM_006710433.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4577632 c.1305A>G p.*435* Chr.1: 8861360-8861360 on Build GRCh38
3671837 c.1272C>A p.A424A Chr.1: 8861393-8861393 on Build GRCh38
4577633 c.1299C>A p.A433A Chr.1: 8861366-8861366 on Build GRCh38
1582258 c.1273G>A p.G425S Chr.1: 8861392-8861392 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs971056387 Chr.1: 8861232-8861232 on Build GRCh38
rs371299158 Chr.1: 8861352-8861352 on Build GRCh38
rs549636657 Chr.1: 8861218-8861218 on Build GRCh38
rs1065737 Chr.1: 8861109-8861109 on Build GRCh38
rs762352421 Chr.1: 8861354-8861354 on Build GRCh38
rs991786286 Chr.1: 8860939-8860939 on Build GRCh38
rs900001513 Chr.1: 8861126-8861126 on Build GRCh38
rs1041283907 Chr.1: 8861336-8861336 on Build GRCh38
rs1041690198 Chr.1: 8861060-8861060 on Build GRCh38
rs571533485 Chr.1: 8861226-8861226 on Build GRCh38
rs779444933 Chr.1: 8861337-8861338 on Build GRCh38
rs913591522 Chr.1: 8860943-8860943 on Build GRCh38
rs759132258 Chr.1: 8861104-8861104 on Build GRCh38
rs933414679 Chr.1: 8861149-8861149 on Build GRCh38
rs943190387 Chr.1: 8861071-8861071 on Build GRCh38
rs914678781 Chr.1: 8861285-8861285 on Build GRCh38
rs527378969 Chr.1: 8861087-8861087 on Build GRCh38
rs763278355 Chr.1: 8861351-8861351 on Build GRCh38
rs966421835 Chr.1: 8861342-8861342 on Build GRCh38
rs562450138 Chr.1: 8861277-8861277 on Build GRCh38
rs577533899 Chr.1: 8860988-8860988 on Build GRCh38
rs1065722 Chr.1: 8861372-8861372 on Build GRCh38
rs770132290 Chr.1: 8861346-8861346 on Build GRCh38
rs1059149 Chr.1: 8861097-8861097 on Build GRCh38
rs35715680 Chr.1: 8861184-8861185 on Build GRCh38
rs3179161 Chr.1: 8861001-8861001 on Build GRCh38
rs1004300534 Chr.1: 8861223-8861223 on Build GRCh38
rs576650903 Chr.1: 8861047-8861047 on Build GRCh38
rs773036999 Chr.1: 8861338-8861338 on Build GRCh38
rs756538264 Chr.1: 8861373-8861373 on Build GRCh38
rs781326126 Chr.1: 8860935-8860935 on Build GRCh38
rs1059176 Chr.1: 8861074-8861074 on Build GRCh38
rs373651010 Chr.1: 8861039-8861039 on Build GRCh38
rs139671695 Chr.1: 8861366-8861366 on Build GRCh38
rs1006859178 Chr.1: 8861233-8861233 on Build GRCh38
rs906191457 Chr.1: 8861385-8861385 on Build GRCh38
rs775783886 Chr.1: 8861349-8861349 on Build GRCh38
rs3199777 Chr.1: 8861116-8861116 on Build GRCh38
rs778409470 Chr.1: 8861322-8861322 on Build GRCh38
rs937448461 Chr.1: 8860995-8860995 on Build GRCh38
rs924632713 Chr.1: 8861321-8861321 on Build GRCh38
rs11544498 Chr.1: 8861309-8861309 on Build GRCh38
rs754420950 Chr.1: 8861320-8861320 on Build GRCh38
rs748077878 Chr.1: 8861331-8861331 on Build GRCh38
rs28999102 Chr.1: 8861135-8861135 on Build GRCh38
rs1041648884 Chr.1: 8861082-8861082 on Build GRCh38
rs1048517338 Chr.1: 8861347-8861347 on Build GRCh38
rs3206710 Chr.1: 8861061-8861061 on Build GRCh38
rs371183978 Chr.1: 8861219-8861219 on Build GRCh38
rs113945549 Chr.1: 8861008-8861008 on Build GRCh38
rs944263410 Chr.1: 8861080-8861080 on Build GRCh38
rs11544504 Chr.1: 8861236-8861236 on Build GRCh38
rs778887391 Chr.1: 8861255-8861255 on Build GRCh38
rs977827292 Chr.1: 8861304-8861304 on Build GRCh38
rs369375352 Chr.1: 8861337-8861337 on Build GRCh38
rs544057356 Chr.1: 8861235-8861235 on Build GRCh38
rs1803537 Chr.1: 8861077-8861077 on Build GRCh38
rs531428896 Chr.1: 8861163-8861163 on Build GRCh38
rs1065725 Chr.1: 8861343-8861343 on Build GRCh38
rs556920303 Chr.1: 8860946-8860946 on Build GRCh38
rs3206709 Chr.1: 8861152-8861152 on Build GRCh38
rs959180287 Chr.1: 8861276-8861276 on Build GRCh38
rs138375821 Chr.1: 8861360-8861360 on Build GRCh38
rs1038919002 Chr.1: 8861122-8861122 on Build GRCh38
rs968755717 Chr.1: 8861286-8861286 on Build GRCh38
rs770966365 Chr.1: 8861070-8861070 on Build GRCh38
rs544900822 Chr.1: 8860937-8860937 on Build GRCh38
rs3207089 Chr.1: 8861305-8861305 on Build GRCh38
rs3206703 Chr.1: 8861238-8861238 on Build GRCh38
rs374794608 Chr.1: 8861353-8861353 on Build GRCh38
rs559946012 Chr.1: 8861056-8861056 on Build GRCh38
rs3211543 Chr.1: 8861270-8861270 on Build GRCh38

View Full Product Details