Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8861245-8861755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO1
Entrez Gene ID
2023
Gene Name
enolase 1
Gene Aliases
ENO1L1, HEL-S-17, MPB1, NNE, PPH
UniGene
Hs.517145
Gene Chromosome Location
Chr.1: 8861000-8879092 on Build GRCh38
Transcripts
NM_001201483.1 NM_001428.3 XM_006710433.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4577632 c.1305A>G p.*435* Chr.1: 8861360-8861360 on Build GRCh38
3671837 c.1272C>A p.A424A Chr.1: 8861393-8861393 on Build GRCh38
4837789 c.1240G>A p.E414K Chr.1: 8861425-8861425 on Build GRCh38
5928072 c.1236-6C>T p.? Chr.1: 8861435-8861435 on Build GRCh38
4577633 c.1299C>A p.A433A Chr.1: 8861366-8861366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371299158 Chr.1: 8861352-8861352 on Build GRCh38
rs896002280 Chr.1: 8861537-8861537 on Build GRCh38
rs74050923 Chr.1: 8861523-8861523 on Build GRCh38
rs762352421 Chr.1: 8861354-8861354 on Build GRCh38
rs542291665 Chr.1: 8861715-8861715 on Build GRCh38
rs773394417 Chr.1: 8861463-8861464 on Build GRCh38
rs895065485 Chr.1: 8861694-8861694 on Build GRCh38
rs1041283907 Chr.1: 8861336-8861336 on Build GRCh38
rs201317089 Chr.1: 8861427-8861427 on Build GRCh38
rs777638154 Chr.1: 8861411-8861411 on Build GRCh38
rs771142338 Chr.1: 8861480-8861480 on Build GRCh38
rs1000533487 Chr.1: 8861566-8861566 on Build GRCh38
rs748565413 Chr.1: 8861399-8861404 on Build GRCh38
rs764855645 Chr.1: 8861480-8861480 on Build GRCh38
rs779444933 Chr.1: 8861337-8861338 on Build GRCh38
rs775022376 Chr.1: 8861401-8861401 on Build GRCh38
rs761185272 Chr.1: 8861457-8861457 on Build GRCh38
rs774472416 Chr.1: 8861439-8861439 on Build GRCh38
rs914678781 Chr.1: 8861285-8861285 on Build GRCh38
rs754648631 Chr.1: 8861393-8861393 on Build GRCh38
rs966421835 Chr.1: 8861342-8861342 on Build GRCh38
rs763278355 Chr.1: 8861351-8861351 on Build GRCh38
rs562450138 Chr.1: 8861277-8861277 on Build GRCh38
rs771949097 Chr.1: 8861392-8861392 on Build GRCh38
rs910053543 Chr.1: 8861538-8861538 on Build GRCh38
rs369746357 Chr.1: 8861395-8861395 on Build GRCh38
rs1065722 Chr.1: 8861372-8861372 on Build GRCh38
rs770132290 Chr.1: 8861346-8861346 on Build GRCh38
rs184739809 Chr.1: 8861456-8861456 on Build GRCh38
rs769042336 Chr.1: 8861434-8861434 on Build GRCh38
rs377267127 Chr.1: 8861462-8861462 on Build GRCh38
rs773036999 Chr.1: 8861338-8861338 on Build GRCh38
rs756538264 Chr.1: 8861373-8861373 on Build GRCh38
rs977132248 Chr.1: 8861510-8861510 on Build GRCh38
rs902331600 Chr.1: 8861718-8861718 on Build GRCh38
rs1020706527 Chr.1: 8861541-8861541 on Build GRCh38
rs1036769700 Chr.1: 8861550-8861550 on Build GRCh38
rs1010825781 Chr.1: 8861703-8861703 on Build GRCh38
rs1047824121 Chr.1: 8861414-8861414 on Build GRCh38
rs140836859 Chr.1: 8861684-8861684 on Build GRCh38
rs139671695 Chr.1: 8861366-8861366 on Build GRCh38
rs747658180 Chr.1: 8861402-8861402 on Build GRCh38
rs1032034930 Chr.1: 8861571-8861571 on Build GRCh38
rs906191457 Chr.1: 8861385-8861385 on Build GRCh38
rs775783886 Chr.1: 8861349-8861349 on Build GRCh38
rs78888703 Chr.1: 8861593-8861593 on Build GRCh38
rs906124172 Chr.1: 8861502-8861502 on Build GRCh38
rs778409470 Chr.1: 8861322-8861322 on Build GRCh38
rs567833503 Chr.1: 8861459-8861459 on Build GRCh38
rs974668405 Chr.1: 8861616-8861616 on Build GRCh38
rs924632713 Chr.1: 8861321-8861321 on Build GRCh38
rs11544498 Chr.1: 8861309-8861309 on Build GRCh38
rs965386204 Chr.1: 8861474-8861474 on Build GRCh38
rs879148876 Chr.1: 8861432-8861432 on Build GRCh38
rs373459987 Chr.1: 8861403-8861403 on Build GRCh38
rs754420950 Chr.1: 8861320-8861320 on Build GRCh38
rs766734256 Chr.1: 8861389-8861389 on Build GRCh38
rs748077878 Chr.1: 8861331-8861331 on Build GRCh38
rs772510631 Chr.1: 8861463-8861463 on Build GRCh38
rs1048517338 Chr.1: 8861347-8861347 on Build GRCh38
rs776586770 Chr.1: 8861418-8861418 on Build GRCh38
rs906215297 Chr.1: 8861584-8861584 on Build GRCh38
rs375678220 Chr.1: 8861534-8861534 on Build GRCh38
rs578199667 Chr.1: 8861526-8861526 on Build GRCh38
rs369375352 Chr.1: 8861337-8861337 on Build GRCh38
rs977827292 Chr.1: 8861304-8861304 on Build GRCh38
rs1065725 Chr.1: 8861343-8861343 on Build GRCh38
rs760894950 Chr.1: 8861470-8861471 on Build GRCh38
rs959180287 Chr.1: 8861276-8861276 on Build GRCh38
rs138375821 Chr.1: 8861360-8861360 on Build GRCh38
rs539010442 Chr.1: 8861546-8861546 on Build GRCh38
rs907946275 Chr.1: 8861702-8861702 on Build GRCh38
rs923075254 Chr.1: 8861622-8861622 on Build GRCh38
rs375928157 Chr.1: 8861448-8861448 on Build GRCh38
rs968755717 Chr.1: 8861286-8861286 on Build GRCh38
rs3207089 Chr.1: 8861305-8861305 on Build GRCh38
rs188800098 Chr.1: 8861463-8861463 on Build GRCh38
rs2071413 Chr.1: 8861676-8861676 on Build GRCh38
rs761860558 Chr.1: 8861444-8861444 on Build GRCh38
rs374794608 Chr.1: 8861353-8861353 on Build GRCh38
rs921530512 Chr.1: 8861512-8861512 on Build GRCh38
rs3211543 Chr.1: 8861270-8861270 on Build GRCh38
rs193006639 Chr.1: 8861417-8861417 on Build GRCh38
rs746966528 Chr.1: 8861412-8861412 on Build GRCh38

View Full Product Details