Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9848550-9849046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNBIP1
Entrez Gene ID
56998
Gene Name
catenin beta interacting protein 1
Gene Aliases
ICAT
UniGene
Hs.463759
Gene Chromosome Location
Chr.1: 9848276-9910322 on Build GRCh38
Transcripts
NM_001012329.1 NM_020248.2 XM_017001853.1 XM_017001854.1 XM_017001855.1

SNP Detail

SNP ID Chromosome Location
rs912937619 Chr.1: 9848961-9848961 on Build GRCh38
rs959170614 Chr.1: 9848874-9848874 on Build GRCh38
rs564356090 Chr.1: 9848962-9848962 on Build GRCh38
rs949383523 Chr.1: 9848649-9848649 on Build GRCh38
rs901263190 Chr.1: 9848860-9848860 on Build GRCh38
rs947681220 Chr.1: 9848701-9848701 on Build GRCh38
rs573140403 Chr.1: 9848963-9848963 on Build GRCh38
rs1007855726 Chr.1: 9848755-9848755 on Build GRCh38
rs917943475 Chr.1: 9848644-9848644 on Build GRCh38
rs575173430 Chr.1: 9848851-9848851 on Build GRCh38
rs77251883 Chr.1: 9848846-9848846 on Build GRCh38
rs577299796 Chr.1: 9849012-9849012 on Build GRCh38
rs1015310495 Chr.1: 9848572-9848572 on Build GRCh38
rs545510134 Chr.1: 9848884-9848884 on Build GRCh38
rs540553925 Chr.1: 9849018-9849018 on Build GRCh38
rs911474419 Chr.1: 9848977-9848977 on Build GRCh38
rs1015672710 Chr.1: 9848771-9848771 on Build GRCh38
rs961506655 Chr.1: 9848599-9848599 on Build GRCh38
rs973807147 Chr.1: 9848834-9848834 on Build GRCh38
rs978154935 Chr.1: 9848886-9848886 on Build GRCh38
rs540792914 Chr.1: 9848817-9848817 on Build GRCh38
rs1051971194 Chr.1: 9848951-9848951 on Build GRCh38
rs1016576207 Chr.1: 9848913-9848913 on Build GRCh38
rs990750634 Chr.1: 9848877-9848877 on Build GRCh38
rs889991152 Chr.1: 9848827-9848827 on Build GRCh38
rs1042872324 Chr.1: 9848665-9848665 on Build GRCh38
rs556820730 Chr.1: 9848848-9848848 on Build GRCh38
rs923928389 Chr.1: 9848889-9848889 on Build GRCh38
rs889950464 Chr.1: 9848590-9848590 on Build GRCh38
rs1053914470 Chr.1: 9848684-9848684 on Build GRCh38
rs951072098 Chr.1: 9848969-9848969 on Build GRCh38
rs1017498648 Chr.1: 9848638-9848655 on Build GRCh38
rs537732058 Chr.1: 9848617-9848618 on Build GRCh38
rs936746336 Chr.1: 9848673-9848673 on Build GRCh38
rs937942577 Chr.1: 9848623-9848623 on Build GRCh38
rs868650809 Chr.1: 9848800-9848800 on Build GRCh38
rs573536520 Chr.1: 9848672-9848672 on Build GRCh38
rs1006164349 Chr.1: 9848897-9848897 on Build GRCh38
rs114793321 Chr.1: 9848634-9848634 on Build GRCh38
rs749684252 Chr.1: 9848979-9848979 on Build GRCh38
rs1041197162 Chr.1: 9848852-9848852 on Build GRCh38
rs1030515702 Chr.1: 9848702-9848702 on Build GRCh38
rs559367859 Chr.1: 9848895-9848895 on Build GRCh38
rs535004472 Chr.1: 9848700-9848700 on Build GRCh38
rs961188722 Chr.1: 9848792-9848792 on Build GRCh38
rs938405540 Chr.1: 9848693-9848694 on Build GRCh38
rs778220764 Chr.1: 9848893-9848893 on Build GRCh38
rs114241722 Chr.1: 9848625-9848625 on Build GRCh38
rs574697103 Chr.1: 9848828-9848828 on Build GRCh38
rs7225 Chr.1: 9848622-9848622 on Build GRCh38
rs896798750 Chr.1: 9849016-9849016 on Build GRCh38
rs775613743 Chr.1: 9848948-9848948 on Build GRCh38
rs942929486 Chr.1: 9849001-9849002 on Build GRCh38
rs922121202 Chr.1: 9848941-9848941 on Build GRCh38
rs1007010877 Chr.1: 9848594-9848594 on Build GRCh38
rs969976790 Chr.1: 9848936-9848936 on Build GRCh38
rs552816528 Chr.1: 9848647-9848647 on Build GRCh38
rs553123465 Chr.1: 9848732-9848732 on Build GRCh38
rs1005538018 Chr.1: 9848958-9848958 on Build GRCh38
rs557377563 Chr.1: 9848581-9848581 on Build GRCh38
rs949416129 Chr.1: 9848881-9848881 on Build GRCh38
rs1053779726 Chr.1: 9848896-9848896 on Build GRCh38
rs890754862 Chr.1: 9848957-9848957 on Build GRCh38
rs748441419 Chr.1: 9848718-9848718 on Build GRCh38
rs115660003 Chr.1: 9848698-9848698 on Build GRCh38
rs756546856 Chr.1: 9848816-9848816 on Build GRCh38
rs917804267 Chr.1: 9848878-9848878 on Build GRCh38
rs979063849 Chr.1: 9848971-9848971 on Build GRCh38
rs973973407 Chr.1: 9848601-9848601 on Build GRCh38
rs890794750 Chr.1: 9848724-9848724 on Build GRCh38

View Full Product Details