Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9877816-9878317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNBIP1
Entrez Gene ID
56998
Gene Name
catenin beta interacting protein 1
Gene Aliases
ICAT
UniGene
Hs.463759
Gene Chromosome Location
Chr.1: 9848276-9910322 on Build GRCh38
Transcripts
NM_001012329.1 NM_020248.2 XM_017001853.1 XM_017001854.1 XM_017001855.1

SNP Detail

SNP ID Chromosome Location
rs951963374 Chr.1: 9878118-9878118 on Build GRCh38
rs950175517 Chr.1: 9877946-9877946 on Build GRCh38
rs142054057 Chr.1: 9878026-9878026 on Build GRCh38
rs1054594200 Chr.1: 9878177-9878177 on Build GRCh38
rs992244983 Chr.1: 9878045-9878045 on Build GRCh38
rs776884328 Chr.1: 9877898-9877898 on Build GRCh38
rs1027523169 Chr.1: 9878265-9878265 on Build GRCh38
rs1043972691 Chr.1: 9878084-9878084 on Build GRCh38
rs951952915 Chr.1: 9878267-9878267 on Build GRCh38
rs1018694593 Chr.1: 9878067-9878067 on Build GRCh38
rs1010993748 Chr.1: 9878022-9878022 on Build GRCh38
rs566291333 Chr.1: 9877986-9877986 on Build GRCh38
rs769929361 Chr.1: 9877851-9877851 on Build GRCh38
rs934830617 Chr.1: 9878171-9878171 on Build GRCh38
rs1001986413 Chr.1: 9878224-9878224 on Build GRCh38
rs993339030 Chr.1: 9878263-9878263 on Build GRCh38
rs768725019 Chr.1: 9877841-9877841 on Build GRCh38
rs17034420 Chr.1: 9878107-9878107 on Build GRCh38
rs960440017 Chr.1: 9878041-9878041 on Build GRCh38
rs1050414744 Chr.1: 9878227-9878227 on Build GRCh38
rs151147798 Chr.1: 9878165-9878165 on Build GRCh38
rs925471433 Chr.1: 9878098-9878098 on Build GRCh38
rs962489095 Chr.1: 9877987-9877987 on Build GRCh38
rs1045773509 Chr.1: 9877964-9877964 on Build GRCh38
rs12075754 Chr.1: 9878057-9878057 on Build GRCh38
rs540114202 Chr.1: 9877846-9877846 on Build GRCh38
rs61161175 Chr.1: 9878246-9878246 on Build GRCh38
rs888815255 Chr.1: 9878228-9878228 on Build GRCh38
rs186553906 Chr.1: 9877881-9877881 on Build GRCh38
rs893347305 Chr.1: 9878011-9878011 on Build GRCh38
rs558521707 Chr.1: 9877890-9877890 on Build GRCh38
rs116812680 Chr.1: 9878250-9878250 on Build GRCh38
rs991807215 Chr.1: 9878125-9878125 on Build GRCh38
rs901916522 Chr.1: 9878124-9878124 on Build GRCh38
rs962560083 Chr.1: 9878213-9878213 on Build GRCh38
rs1004092149 Chr.1: 9877968-9877968 on Build GRCh38
rs997144284 Chr.1: 9878197-9878197 on Build GRCh38
rs71583826 Chr.1: 9877842-9877843 on Build GRCh38
rs1054651542 Chr.1: 9878006-9878006 on Build GRCh38
rs1018724343 Chr.1: 9878247-9878247 on Build GRCh38
rs920511094 Chr.1: 9878110-9878110 on Build GRCh38
rs945638274 Chr.1: 9878065-9878065 on Build GRCh38
rs960742294 Chr.1: 9878231-9878231 on Build GRCh38
rs766931242 Chr.1: 9878132-9878132 on Build GRCh38
rs189448656 Chr.1: 9878232-9878232 on Build GRCh38
rs572491162 Chr.1: 9878167-9878167 on Build GRCh38
rs541526460 Chr.1: 9878185-9878185 on Build GRCh38
rs551980776 Chr.1: 9878068-9878068 on Build GRCh38
rs961887820 Chr.1: 9878069-9878069 on Build GRCh38
rs533601783 Chr.1: 9877993-9877993 on Build GRCh38
rs1028690222 Chr.1: 9878025-9878025 on Build GRCh38
rs950041957 Chr.1: 9878152-9878152 on Build GRCh38
rs573616478 Chr.1: 9878091-9878091 on Build GRCh38

View Full Product Details