Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101723440-101723937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHPT1
Entrez Gene ID
56994
Gene Name
choline phosphotransferase 1
Gene Aliases
CPT, CPT1
UniGene
Hs.293077
Gene Chromosome Location
Chr.12: 101697639-101752038 on Build GRCh38
Transcripts
NM_020244.2 XM_011538574.1 XM_011538575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2063579 c.1062A>C p.A354A Chr.12: 101723844-101723844 on Build GRCh38
6234015 c.1065+5G>T p.? Chr.12: 101723852-101723852 on Build GRCh38
430171 c.973C>T p.Q325* Chr.12: 101723755-101723755 on Build GRCh38
6628577 c.1038C>T p.D346D Chr.12: 101723820-101723820 on Build GRCh38
3667074 c.968A>G p.Y323C Chr.12: 101723750-101723750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780129088 Chr.12: 101723566-101723566 on Build GRCh38
rs761803831 Chr.12: 101723701-101723701 on Build GRCh38
rs753017500 Chr.12: 101723770-101723770 on Build GRCh38
rs756408672 Chr.12: 101723775-101723775 on Build GRCh38
rs373712055 Chr.12: 101723852-101723852 on Build GRCh38
rs11082 Chr.12: 101723811-101723811 on Build GRCh38
rs971654599 Chr.12: 101723619-101723619 on Build GRCh38
rs149394701 Chr.12: 101723473-101723474 on Build GRCh38
rs754143006 Chr.12: 101723886-101723886 on Build GRCh38
rs776461364 Chr.12: 101723625-101723629 on Build GRCh38
rs79716906 Chr.12: 101723803-101723803 on Build GRCh38
rs759516890 Chr.12: 101723850-101723850 on Build GRCh38
rs765290675 Chr.12: 101723709-101723709 on Build GRCh38
rs756286764 Chr.12: 101723883-101723883 on Build GRCh38
rs535029486 Chr.12: 101723575-101723575 on Build GRCh38
rs866964354 Chr.12: 101723787-101723787 on Build GRCh38
rs900548306 Chr.12: 101723662-101723662 on Build GRCh38
rs933276315 Chr.12: 101723835-101723835 on Build GRCh38
rs757710976 Chr.12: 101723786-101723786 on Build GRCh38
rs572035898 Chr.12: 101723768-101723768 on Build GRCh38
rs764915335 Chr.12: 101723702-101723705 on Build GRCh38
rs1012925370 Chr.12: 101723588-101723588 on Build GRCh38
rs569832616 Chr.12: 101723663-101723663 on Build GRCh38
rs1042440140 Chr.12: 101723508-101723508 on Build GRCh38
rs555795326 Chr.12: 101723636-101723636 on Build GRCh38
rs368918351 Chr.12: 101723584-101723584 on Build GRCh38
rs745309692 Chr.12: 101723799-101723799 on Build GRCh38
rs541777969 Chr.12: 101723716-101723716 on Build GRCh38
rs761717958 Chr.12: 101723825-101723825 on Build GRCh38
rs764029502 Chr.12: 101723694-101723694 on Build GRCh38
rs959344323 Chr.12: 101723907-101723907 on Build GRCh38
rs757929652 Chr.12: 101723742-101723742 on Build GRCh38
rs1038885500 Chr.12: 101723475-101723475 on Build GRCh38
rs141024430 Chr.12: 101723780-101723780 on Build GRCh38
rs779405162 Chr.12: 101723798-101723798 on Build GRCh38
rs1034783975 Chr.12: 101723864-101723864 on Build GRCh38
rs756911163 Chr.12: 101723673-101723674 on Build GRCh38
rs902496400 Chr.12: 101723544-101723544 on Build GRCh38
rs561553917 Chr.12: 101723763-101723763 on Build GRCh38
rs1048017700 Chr.12: 101723882-101723882 on Build GRCh38
rs1040473612 Chr.12: 101723590-101723590 on Build GRCh38
rs180901994 Chr.12: 101723470-101723470 on Build GRCh38
rs755094453 Chr.12: 101723761-101723761 on Build GRCh38
rs765202598 Chr.12: 101723833-101723833 on Build GRCh38
rs746714890 Chr.12: 101723814-101723814 on Build GRCh38
rs1023200736 Chr.12: 101723592-101723592 on Build GRCh38
rs115007859 Chr.12: 101723821-101723821 on Build GRCh38
rs779486456 Chr.12: 101723872-101723895 on Build GRCh38
rs756588891 Chr.12: 101723473-101723473 on Build GRCh38
rs768456280 Chr.12: 101723820-101723820 on Build GRCh38
rs997728906 Chr.12: 101723537-101723537 on Build GRCh38
rs776680337 Chr.12: 101723697-101723697 on Build GRCh38
rs773272605 Chr.12: 101723845-101723845 on Build GRCh38
rs577799309 Chr.12: 101723585-101723585 on Build GRCh38
rs1028571092 Chr.12: 101723670-101723670 on Build GRCh38
rs1047585759 Chr.12: 101723545-101723545 on Build GRCh38
rs752828705 Chr.12: 101723853-101723853 on Build GRCh38
rs368481855 Chr.12: 101723894-101723894 on Build GRCh38
rs550003918 Chr.12: 101723884-101723884 on Build GRCh38
rs537286227 Chr.12: 101723607-101723607 on Build GRCh38
rs142506108 Chr.12: 101723793-101723793 on Build GRCh38
rs1006475306 Chr.12: 101723913-101723913 on Build GRCh38
rs186321698 Chr.12: 101723570-101723570 on Build GRCh38
rs750040817 Chr.12: 101723711-101723711 on Build GRCh38
rs751634924 Chr.12: 101723731-101723731 on Build GRCh38

View Full Product Details