Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 105370945-105371480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf75
Entrez Gene ID
387882
Gene Name
chromosome 12 open reading frame 75
Gene Aliases
AGD3, OCC-1, OCC1
UniGene
Hs.368938
Gene Chromosome Location
Chr.12: 105330636-105371518 on Build GRCh38
Transcripts
NM_001145199.1

SNP Detail

SNP ID Chromosome Location
rs779376994 Chr.12: 105371283-105371283 on Build GRCh38
rs142018451 Chr.12: 105371392-105371392 on Build GRCh38
rs368567335 Chr.12: 105370986-105370986 on Build GRCh38
rs186290074 Chr.12: 105371269-105371269 on Build GRCh38
rs757448805 Chr.12: 105371249-105371249 on Build GRCh38
rs531812891 Chr.12: 105371267-105371267 on Build GRCh38
rs918638105 Chr.12: 105371085-105371085 on Build GRCh38
rs749804855 Chr.12: 105371342-105371342 on Build GRCh38
rs941529832 Chr.12: 105371077-105371077 on Build GRCh38
rs974173912 Chr.12: 105371348-105371348 on Build GRCh38
rs929982303 Chr.12: 105371108-105371108 on Build GRCh38
rs962860043 Chr.12: 105371344-105371344 on Build GRCh38
rs6797 Chr.12: 105371206-105371206 on Build GRCh38
rs911783635 Chr.12: 105371084-105371084 on Build GRCh38
rs535071139 Chr.12: 105371184-105371184 on Build GRCh38
rs150683511 Chr.12: 105371393-105371393 on Build GRCh38
rs992641431 Chr.12: 105371451-105371451 on Build GRCh38
rs568554797 Chr.12: 105371287-105371287 on Build GRCh38
rs930305764 Chr.12: 105371134-105371134 on Build GRCh38
rs895736942 Chr.12: 105371136-105371136 on Build GRCh38
rs944676291 Chr.12: 105371098-105371098 on Build GRCh38
rs1054212234 Chr.12: 105371247-105371247 on Build GRCh38
rs1001685858 Chr.12: 105371195-105371195 on Build GRCh38
rs143768120 Chr.12: 105371270-105371270 on Build GRCh38
rs1020962866 Chr.12: 105371256-105371256 on Build GRCh38
rs552464757 Chr.12: 105371107-105371107 on Build GRCh38
rs73383218 Chr.12: 105371026-105371026 on Build GRCh38
rs918691981 Chr.12: 105371359-105371359 on Build GRCh38
rs950028364 Chr.12: 105371145-105371145 on Build GRCh38
rs1020323743 Chr.12: 105370987-105370987 on Build GRCh38
rs553104978 Chr.12: 105371391-105371391 on Build GRCh38
rs547892889 Chr.12: 105371345-105371345 on Build GRCh38
rs566177199 Chr.12: 105371175-105371175 on Build GRCh38
rs546860187 Chr.12: 105371082-105371082 on Build GRCh38
rs966276510 Chr.12: 105371349-105371349 on Build GRCh38
rs375146675 Chr.12: 105371248-105371248 on Build GRCh38
rs1000020684 Chr.12: 105371246-105371246 on Build GRCh38
rs905732111 Chr.12: 105371217-105371217 on Build GRCh38
rs751838017 Chr.12: 105371215-105371215 on Build GRCh38
rs986044737 Chr.12: 105371005-105371005 on Build GRCh38
rs748995669 Chr.12: 105371288-105371288 on Build GRCh38
rs566103461 Chr.12: 105371376-105371376 on Build GRCh38
rs971973205 Chr.12: 105371353-105371353 on Build GRCh38
rs904962854 Chr.12: 105371192-105371192 on Build GRCh38
rs565013683 Chr.12: 105371421-105371421 on Build GRCh38
rs978780135 Chr.12: 105371021-105371021 on Build GRCh38
rs930272985 Chr.12: 105371405-105371405 on Build GRCh38
rs1061749 Chr.12: 105371319-105371319 on Build GRCh38
rs1048815143 Chr.12: 105371192-105371192 on Build GRCh38
rs953189808 Chr.12: 105371071-105371071 on Build GRCh38

View Full Product Details